Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 22946017-22946480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC3
Entrez Gene ID
219681
Gene Name
armadillo repeat containing 3
Gene Aliases
CT81, KU-CT-1
UniGene
Hs.659807
Gene Chromosome Location
Chr.10: 22928024-23038527 on Build GRCh38
Transcripts
NM_001282745.1 NM_001282746.1 NM_001282747.1 NM_173081.4 XM_005252380.2 XM_005252381.2 XM_011519350.2 XM_017015831.1 XM_017015832.1 XM_017015833.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6065488 c.56C>A p.P19Q Chr.10: 22946151-22946151 on Build GRCh38
5759652 c.166+7_166+8insT p.? Chr.10: 22946268-22946269 on Build GRCh38
5678756 c.131A>T p.K44I Chr.10: 22946226-22946226 on Build GRCh38
5678755 c.131A>T p.K44I Chr.10: 22946226-22946226 on Build GRCh38
3436574 c.148T>A p.Y50N Chr.10: 22946243-22946243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758024069 Chr.10: 22946261-22946261 on Build GRCh38
rs745613551 Chr.10: 22946095-22946096 on Build GRCh38
rs541181054 Chr.10: 22946384-22946384 on Build GRCh38
rs778733776 Chr.10: 22946247-22946247 on Build GRCh38
rs371887647 Chr.10: 22946270-22946270 on Build GRCh38
rs775427317 Chr.10: 22946269-22946269 on Build GRCh38
rs562018374 Chr.10: 22946110-22946110 on Build GRCh38
rs750373852 Chr.10: 22946194-22946194 on Build GRCh38
rs775803616 Chr.10: 22946297-22946297 on Build GRCh38
rs761910798 Chr.10: 22946160-22946160 on Build GRCh38
rs182361558 Chr.10: 22946362-22946362 on Build GRCh38
rs1029748955 Chr.10: 22946107-22946107 on Build GRCh38
rs772372004 Chr.10: 22946174-22946174 on Build GRCh38
rs961925541 Chr.10: 22946175-22946175 on Build GRCh38
rs367649259 Chr.10: 22946259-22946259 on Build GRCh38
rs937383365 Chr.10: 22946394-22946394 on Build GRCh38
rs756062248 Chr.10: 22946219-22946219 on Build GRCh38
rs994060165 Chr.10: 22946062-22946068 on Build GRCh38
rs368018823 Chr.10: 22946266-22946266 on Build GRCh38
rs754765906 Chr.10: 22946228-22946228 on Build GRCh38
rs1006317785 Chr.10: 22946112-22946112 on Build GRCh38
rs111970965 Chr.10: 22946422-22946422 on Build GRCh38
rs112647805 Chr.10: 22946111-22946111 on Build GRCh38
rs968045590 Chr.10: 22946320-22946320 on Build GRCh38
rs751729830 Chr.10: 22946158-22946158 on Build GRCh38
rs1024882733 Chr.10: 22946330-22946330 on Build GRCh38
rs767601531 Chr.10: 22946181-22946181 on Build GRCh38
rs764313006 Chr.10: 22946153-22946153 on Build GRCh38
rs1018059362 Chr.10: 22946229-22946229 on Build GRCh38
rs753695218 Chr.10: 22946222-22946222 on Build GRCh38
rs746421248 Chr.10: 22946267-22946267 on Build GRCh38
rs565587591 Chr.10: 22946268-22946269 on Build GRCh38
rs374957753 Chr.10: 22946118-22946118 on Build GRCh38
rs747764467 Chr.10: 22946256-22946256 on Build GRCh38
rs1013823771 Chr.10: 22946311-22946311 on Build GRCh38
rs187449128 Chr.10: 22946424-22946424 on Build GRCh38
rs751023830 Chr.10: 22946377-22946377 on Build GRCh38
rs887738778 Chr.10: 22946094-22946094 on Build GRCh38
rs980616378 Chr.10: 22946366-22946366 on Build GRCh38
rs1015397581 Chr.10: 22946157-22946157 on Build GRCh38
rs981690694 Chr.10: 22946341-22946341 on Build GRCh38

View Full Product Details