Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 22961604-22962128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC3
Entrez Gene ID
219681
Gene Name
armadillo repeat containing 3
Gene Aliases
CT81, KU-CT-1
UniGene
Hs.659807
Gene Chromosome Location
Chr.10: 22928024-23038527 on Build GRCh38
Transcripts
NM_001282745.1 NM_001282746.1 NM_001282747.1 NM_173081.4 XM_005252380.2 XM_005252381.2 XM_011519350.2 XM_017015831.1 XM_017015832.1 XM_017015833.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
274047 c.715A>C p.K239Q Chr.10: 22962061-22962061 on Build GRCh38
4165455 c.680T>G p.M227R Chr.10: 22962026-22962026 on Build GRCh38
4437519 c.713_714insT p.K239fs*1 Chr.10: 22962059-22962060 on Build GRCh38
227552 c.605C>T p.S202L Chr.10: 22961951-22961951 on Build GRCh38
4165454 c.680T>G p.M227R Chr.10: 22962026-22962026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374224435 Chr.10: 22962025-22962025 on Build GRCh38
rs186415940 Chr.10: 22962063-22962063 on Build GRCh38
rs973938395 Chr.10: 22962043-22962043 on Build GRCh38
rs781026238 Chr.10: 22962083-22962083 on Build GRCh38
rs987672266 Chr.10: 22961681-22961681 on Build GRCh38
rs766270838 Chr.10: 22961843-22961843 on Build GRCh38
rs770710346 Chr.10: 22961975-22961975 on Build GRCh38
rs943512276 Chr.10: 22961779-22961779 on Build GRCh38
rs751461064 Chr.10: 22962072-22962072 on Build GRCh38
rs775628110 Chr.10: 22961695-22961695 on Build GRCh38
rs772716427 Chr.10: 22962004-22962004 on Build GRCh38
rs768059647 Chr.10: 22961892-22961892 on Build GRCh38
rs779698923 Chr.10: 22961850-22961850 on Build GRCh38
rs552233648 Chr.10: 22961678-22961678 on Build GRCh38
rs143893027 Chr.10: 22961940-22961940 on Build GRCh38
rs966986117 Chr.10: 22961905-22961905 on Build GRCh38
rs780253470 Chr.10: 22961835-22961835 on Build GRCh38
rs878950850 Chr.10: 22961716-22961718 on Build GRCh38
rs995739980 Chr.10: 22961884-22961884 on Build GRCh38
rs975570785 Chr.10: 22961680-22961680 on Build GRCh38
rs373582466 Chr.10: 22961912-22961912 on Build GRCh38
rs1029397537 Chr.10: 22961921-22961921 on Build GRCh38
rs764764866 Chr.10: 22961888-22961888 on Build GRCh38
rs182810480 Chr.10: 22961638-22961638 on Build GRCh38
rs924852593 Chr.10: 22961768-22961768 on Build GRCh38
rs559103578 Chr.10: 22961927-22961927 on Build GRCh38
rs778743270 Chr.10: 22961838-22961838 on Build GRCh38
rs922270192 Chr.10: 22961642-22961642 on Build GRCh38
rs768445121 Chr.10: 22961982-22961986 on Build GRCh38
rs530843797 Chr.10: 22961987-22961987 on Build GRCh38
rs149454604 Chr.10: 22961920-22961920 on Build GRCh38
rs1021320309 Chr.10: 22962020-22962020 on Build GRCh38
rs568886267 Chr.10: 22961706-22961706 on Build GRCh38
rs775133484 Chr.10: 22961992-22961992 on Build GRCh38
rs968845267 Chr.10: 22961724-22961724 on Build GRCh38
rs746733675 Chr.10: 22961864-22961864 on Build GRCh38
rs773804605 Chr.10: 22962006-22962006 on Build GRCh38
rs963734973 Chr.10: 22961674-22961674 on Build GRCh38
rs1003573140 Chr.10: 22961855-22961855 on Build GRCh38
rs200269169 Chr.10: 22961935-22961935 on Build GRCh38
rs764554689 Chr.10: 22961841-22961841 on Build GRCh38
rs35798093 Chr.10: 22961897-22961897 on Build GRCh38
rs977734810 Chr.10: 22961759-22961759 on Build GRCh38
rs920640133 Chr.10: 22961826-22961826 on Build GRCh38
rs761040358 Chr.10: 22961764-22961764 on Build GRCh38
rs376421363 Chr.10: 22961865-22961865 on Build GRCh38
rs757043972 Chr.10: 22962080-22962080 on Build GRCh38
rs765304042 Chr.10: 22962027-22962027 on Build GRCh38
rs577220653 Chr.10: 22962037-22962037 on Build GRCh38
rs200768638 Chr.10: 22961885-22961885 on Build GRCh38
rs201134940 Chr.10: 22961896-22961896 on Build GRCh38
rs548339594 Chr.10: 22961735-22961735 on Build GRCh38
rs748050100 Chr.10: 22961839-22961839 on Build GRCh38
rs765894329 Chr.10: 22961959-22961959 on Build GRCh38
rs771907177 Chr.10: 22961850-22961850 on Build GRCh38
rs769245936 Chr.10: 22961986-22961986 on Build GRCh38
rs74121683 Chr.10: 22961978-22961978 on Build GRCh38
rs754355253 Chr.10: 22961859-22961859 on Build GRCh38
rs985112609 Chr.10: 22961944-22961944 on Build GRCh38
rs745510825 Chr.10: 22961982-22961982 on Build GRCh38
rs762484589 Chr.10: 22962002-22962002 on Build GRCh38
rs531457076 Chr.10: 22961734-22961734 on Build GRCh38
rs370375510 Chr.10: 22962058-22962058 on Build GRCh38
rs766731236 Chr.10: 22962015-22962015 on Build GRCh38
rs762895439 Chr.10: 22961716-22961716 on Build GRCh38
rs924894705 Chr.10: 22962044-22962044 on Build GRCh38
rs992862996 Chr.10: 22962086-22962086 on Build GRCh38
rs749339956 Chr.10: 22961837-22961837 on Build GRCh38
rs138681890 Chr.10: 22962030-22962030 on Build GRCh38
rs929376583 Chr.10: 22961836-22961836 on Build GRCh38
rs752782595 Chr.10: 22962028-22962028 on Build GRCh38
rs754047537 Chr.10: 22961914-22961914 on Build GRCh38
rs758990975 Chr.10: 22961886-22961886 on Build GRCh38
rs760975316 Chr.10: 22962013-22962013 on Build GRCh38
rs78062147 Chr.10: 22961890-22961890 on Build GRCh38
rs996580496 Chr.10: 22962019-22962019 on Build GRCh38
rs910831070 Chr.10: 22961697-22961697 on Build GRCh38
rs933693240 Chr.10: 22961677-22961677 on Build GRCh38
rs749154106 Chr.10: 22961933-22961946 on Build GRCh38
rs777621623 Chr.10: 22961952-22961952 on Build GRCh38
rs1037884165 Chr.10: 22961817-22961817 on Build GRCh38
rs754060123 Chr.10: 22962024-22962024 on Build GRCh38
rs762394643 Chr.10: 22961891-22961891 on Build GRCh38

View Full Product Details