Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 23320649-23321156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf67
Entrez Gene ID
256815
Gene Name
chromosome 10 open reading frame 67
Gene Aliases
C10orf115, LINC01552, bA215C7.4
UniGene
Hs.132442
Gene Chromosome Location
Chr.10: 23202851-23345216 on Build GRCh38
Transcripts
NM_153714.2 XM_011519440.1 XM_011519441.1 XM_011519442.2 XM_011519444.2 XM_011519445.2 XM_011519446.1 XM_017016029.1 XM_017016030.1 XM_017016031.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5589864 c.347C>T p.S116F Chr.10: 23320793-23320793 on Build GRCh38
5370465 c.498C>T p.S166S Chr.10: 23320792-23320792 on Build GRCh38
5370466 c.348C>T p.S116S Chr.10: 23320792-23320792 on Build GRCh38
917087 c.529G>C p.V177L Chr.10: 23320761-23320761 on Build GRCh38
5428754 c.368A>G p.D123G Chr.10: 23320772-23320772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781070349 Chr.10: 23320924-23320924 on Build GRCh38
rs758152123 Chr.10: 23320752-23320752 on Build GRCh38
rs561373851 Chr.10: 23320895-23320896 on Build GRCh38
rs763296661 Chr.10: 23320813-23320813 on Build GRCh38
rs771106932 Chr.10: 23321024-23321024 on Build GRCh38
rs748249693 Chr.10: 23320799-23320799 on Build GRCh38
rs756607064 Chr.10: 23320753-23320753 on Build GRCh38
rs942241775 Chr.10: 23320689-23320689 on Build GRCh38
rs111466499 Chr.10: 23320902-23320903 on Build GRCh38
rs769817249 Chr.10: 23320804-23320804 on Build GRCh38
rs567244048 Chr.10: 23320912-23320912 on Build GRCh38
rs1019178044 Chr.10: 23321078-23321078 on Build GRCh38
rs781062423 Chr.10: 23320798-23320798 on Build GRCh38
rs944881433 Chr.10: 23320873-23320873 on Build GRCh38
rs377398355 Chr.10: 23321129-23321129 on Build GRCh38
rs372849908 Chr.10: 23320714-23320714 on Build GRCh38
rs746737371 Chr.10: 23320847-23320847 on Build GRCh38
rs753393294 Chr.10: 23320740-23320740 on Build GRCh38
rs530405026 Chr.10: 23320816-23320816 on Build GRCh38
rs573730734 Chr.10: 23321025-23321025 on Build GRCh38
rs923576238 Chr.10: 23320832-23320832 on Build GRCh38
rs558961610 Chr.10: 23320992-23320992 on Build GRCh38
rs759744842 Chr.10: 23321038-23321038 on Build GRCh38
rs367624805 Chr.10: 23320693-23320693 on Build GRCh38
rs375305150 Chr.10: 23320747-23320747 on Build GRCh38
rs760139478 Chr.10: 23320723-23320723 on Build GRCh38
rs756784171 Chr.10: 23320741-23320741 on Build GRCh38
rs1011476092 Chr.10: 23321042-23321042 on Build GRCh38
rs536498137 Chr.10: 23320921-23320921 on Build GRCh38
rs1004762076 Chr.10: 23320673-23320673 on Build GRCh38
rs528353836 Chr.10: 23320683-23320683 on Build GRCh38
rs369549102 Chr.10: 23320846-23320846 on Build GRCh38
rs766887953 Chr.10: 23320718-23320718 on Build GRCh38
rs183966569 Chr.10: 23321098-23321098 on Build GRCh38
rs768209883 Chr.10: 23320900-23320900 on Build GRCh38
rs762909303 Chr.10: 23321023-23321023 on Build GRCh38
rs912060844 Chr.10: 23320867-23320867 on Build GRCh38
rs538916747 Chr.10: 23320969-23320969 on Build GRCh38
rs903621054 Chr.10: 23321015-23321015 on Build GRCh38
rs909500160 Chr.10: 23320955-23320955 on Build GRCh38
rs754379469 Chr.10: 23320996-23320996 on Build GRCh38
rs779572122 Chr.10: 23320839-23320839 on Build GRCh38
rs770051889 Chr.10: 23320702-23320702 on Build GRCh38
rs763450072 Chr.10: 23320724-23320724 on Build GRCh38
rs922010625 Chr.10: 23320920-23320920 on Build GRCh38
rs960539033 Chr.10: 23320694-23320694 on Build GRCh38
rs909434107 Chr.10: 23320687-23320687 on Build GRCh38
rs181359212 Chr.10: 23320788-23320788 on Build GRCh38
rs1000532755 Chr.10: 23321019-23321019 on Build GRCh38
rs886415041 Chr.10: 23320947-23320947 on Build GRCh38
rs768642919 Chr.10: 23320791-23320791 on Build GRCh38
rs1023839077 Chr.10: 23321002-23321002 on Build GRCh38
rs1002594028 Chr.10: 23320886-23320886 on Build GRCh38
rs750123026 Chr.10: 23320751-23320751 on Build GRCh38
rs1035833232 Chr.10: 23320890-23320890 on Build GRCh38
rs773627213 Chr.10: 23320711-23320711 on Build GRCh38
rs773539146 Chr.10: 23320805-23320805 on Build GRCh38
rs752069925 Chr.10: 23320719-23320719 on Build GRCh38
rs967137439 Chr.10: 23320862-23320862 on Build GRCh38
rs11013386 Chr.10: 23320968-23320968 on Build GRCh38
rs919348162 Chr.10: 23320706-23320706 on Build GRCh38
rs553106691 Chr.10: 23320929-23320929 on Build GRCh38
rs767758320 Chr.10: 23321083-23321083 on Build GRCh38

View Full Product Details