Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 24583602-24584151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP21
Entrez Gene ID
57584
Gene Name
Rho GTPase activating protein 21
Gene Aliases
ARHGAP10
UniGene
Hs.524195
Gene Chromosome Location
Chr.10: 24583609-24725421 on Build GRCh38
Transcripts
NM_020824.3 XM_005252542.3 XM_005252544.4 XM_011519602.2 XM_011519603.2 XM_011519604.2 XM_011519605.2 XM_011519606.2 XM_011519607.2 XM_017016459.1 XM_017016460.1 XM_017016461.1

SNP Detail

SNP ID Chromosome Location
rs952786869 Chr.10: 24583920-24583920 on Build GRCh38
rs10327 Chr.10: 24584061-24584061 on Build GRCh38
rs921154245 Chr.10: 24583990-24583990 on Build GRCh38
rs192704606 Chr.10: 24584056-24584056 on Build GRCh38
rs932450494 Chr.10: 24583737-24583737 on Build GRCh38
rs557299864 Chr.10: 24583825-24583825 on Build GRCh38
rs576664400 Chr.10: 24583643-24583643 on Build GRCh38
rs117654517 Chr.10: 24583993-24583993 on Build GRCh38
rs1025161356 Chr.10: 24583860-24583861 on Build GRCh38
rs3184324 Chr.10: 24583994-24583994 on Build GRCh38
rs3184312 Chr.10: 24584046-24584046 on Build GRCh38
rs1004409968 Chr.10: 24584071-24584071 on Build GRCh38
rs1043295265 Chr.10: 24583742-24583742 on Build GRCh38
rs878924808 Chr.10: 24583640-24583640 on Build GRCh38
rs555641259 Chr.10: 24583766-24583766 on Build GRCh38
rs905418985 Chr.10: 24584058-24584059 on Build GRCh38
rs983268577 Chr.10: 24583923-24583924 on Build GRCh38
rs908656996 Chr.10: 24583960-24583960 on Build GRCh38
rs999016319 Chr.10: 24583804-24583804 on Build GRCh38
rs112035075 Chr.10: 24583830-24583830 on Build GRCh38
rs529091107 Chr.10: 24584036-24584036 on Build GRCh38
rs3184337 Chr.10: 24583975-24583975 on Build GRCh38
rs3184309 Chr.10: 24584057-24584057 on Build GRCh38
rs3184330 Chr.10: 24583986-24583986 on Build GRCh38
rs575732951 Chr.10: 24583877-24583877 on Build GRCh38
rs1027574171 Chr.10: 24583892-24583892 on Build GRCh38
rs1040970930 Chr.10: 24583715-24583715 on Build GRCh38
rs548978153 Chr.10: 24584053-24584053 on Build GRCh38
rs3184307 Chr.10: 24584086-24584086 on Build GRCh38
rs3184310 Chr.10: 24584048-24584048 on Build GRCh38
rs3184313 Chr.10: 24584039-24584039 on Build GRCh38
rs3184308 Chr.10: 24584081-24584081 on Build GRCh38
rs749561442 Chr.10: 24583760-24583760 on Build GRCh38
rs908539517 Chr.10: 24583658-24583658 on Build GRCh38
rs969628885 Chr.10: 24583862-24583862 on Build GRCh38
rs188146861 Chr.10: 24584031-24584031 on Build GRCh38
rs535055328 Chr.10: 24583656-24583656 on Build GRCh38
rs944058982 Chr.10: 24583674-24583674 on Build GRCh38
rs1002347407 Chr.10: 24583878-24583879 on Build GRCh38
rs965354368 Chr.10: 24583973-24583973 on Build GRCh38
rs976767776 Chr.10: 24583974-24583974 on Build GRCh38
rs3210916 Chr.10: 24583931-24583931 on Build GRCh38
rs765159491 Chr.10: 24583999-24583999 on Build GRCh38
rs1002871978 Chr.10: 24584059-24584059 on Build GRCh38
rs3184321 Chr.10: 24584000-24584000 on Build GRCh38
rs1053265475 Chr.10: 24584017-24584017 on Build GRCh38
rs751933390 Chr.10: 24583692-24583692 on Build GRCh38
rs3184348 Chr.10: 24583883-24583883 on Build GRCh38
rs1127930 Chr.10: 24583700-24583700 on Build GRCh38
rs543596193 Chr.10: 24584002-24584002 on Build GRCh38
rs753295763 Chr.10: 24584022-24584022 on Build GRCh38
rs368935276 Chr.10: 24583757-24583757 on Build GRCh38
rs145297844 Chr.10: 24583716-24583716 on Build GRCh38
rs1013903205 Chr.10: 24583848-24583848 on Build GRCh38
rs3184353 Chr.10: 24583863-24583863 on Build GRCh38
rs931402558 Chr.10: 24583647-24583647 on Build GRCh38
rs3184359 Chr.10: 24583829-24583829 on Build GRCh38
rs952984167 Chr.10: 24584062-24584062 on Build GRCh38

View Full Product Details