Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 24585626-24586124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP21
Entrez Gene ID
57584
Gene Name
Rho GTPase activating protein 21
Gene Aliases
ARHGAP10
UniGene
Hs.524195
Gene Chromosome Location
Chr.10: 24583609-24725421 on Build GRCh38
Transcripts
NM_020824.3 XM_005252542.3 XM_005252544.4 XM_011519602.2 XM_011519603.2 XM_011519604.2 XM_011519605.2 XM_011519606.2 XM_011519607.2 XM_017016459.1 XM_017016460.1 XM_017016461.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5703544 c.4630G>T p.D1544Y Chr.10: 24585656-24585656 on Build GRCh38
4982152 c.4273G>C p.E1425Q Chr.10: 24586013-24586013 on Build GRCh38
2133568 c.4491G>A p.T1497T Chr.10: 24585795-24585795 on Build GRCh38
2133569 c.4378G>C p.E1460Q Chr.10: 24585908-24585908 on Build GRCh38
4548468 c.4478G>T p.G1493V Chr.10: 24585808-24585808 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61854256 Chr.10: 24586021-24586021 on Build GRCh38
rs200304974 Chr.10: 24585904-24585904 on Build GRCh38
rs141727692 Chr.10: 24585743-24585743 on Build GRCh38
rs777570601 Chr.10: 24585648-24585648 on Build GRCh38
rs750955643 Chr.10: 24585728-24585728 on Build GRCh38
rs778593945 Chr.10: 24585856-24585856 on Build GRCh38
rs752154046 Chr.10: 24585834-24585834 on Build GRCh38
rs556966750 Chr.10: 24585747-24585747 on Build GRCh38
rs752349033 Chr.10: 24585745-24585745 on Build GRCh38
rs767642142 Chr.10: 24586045-24586045 on Build GRCh38
rs775758205 Chr.10: 24585694-24585694 on Build GRCh38
rs959052780 Chr.10: 24586020-24586022 on Build GRCh38
rs751585755 Chr.10: 24585968-24585968 on Build GRCh38
rs779848429 Chr.10: 24585957-24585957 on Build GRCh38
rs897652746 Chr.10: 24585858-24585858 on Build GRCh38
rs765823375 Chr.10: 24585722-24585722 on Build GRCh38
rs771394963 Chr.10: 24586017-24586017 on Build GRCh38
rs61735170 Chr.10: 24585899-24585899 on Build GRCh38
rs3210911 Chr.10: 24585971-24585971 on Build GRCh38
rs761397100 Chr.10: 24586029-24586029 on Build GRCh38
rs200136147 Chr.10: 24585708-24585708 on Build GRCh38
rs765015998 Chr.10: 24585942-24585942 on Build GRCh38
rs762349483 Chr.10: 24585721-24585721 on Build GRCh38
rs370382688 Chr.10: 24585655-24585655 on Build GRCh38
rs138692601 Chr.10: 24585832-24585832 on Build GRCh38
rs201814092 Chr.10: 24586041-24586041 on Build GRCh38
rs774810878 Chr.10: 24586018-24586018 on Build GRCh38
rs765739709 Chr.10: 24585813-24585813 on Build GRCh38
rs745777741 Chr.10: 24585755-24585756 on Build GRCh38
rs775462639 Chr.10: 24585781-24585781 on Build GRCh38
rs370000857 Chr.10: 24585862-24585862 on Build GRCh38
rs774981213 Chr.10: 24585914-24585914 on Build GRCh38
rs751495726 Chr.10: 24586042-24586042 on Build GRCh38
rs144805251 Chr.10: 24585796-24585796 on Build GRCh38
rs769009600 Chr.10: 24585795-24585795 on Build GRCh38
rs761483526 Chr.10: 24585936-24585936 on Build GRCh38
rs1056215158 Chr.10: 24585794-24585794 on Build GRCh38
rs774828148 Chr.10: 24585966-24585967 on Build GRCh38
rs569426222 Chr.10: 24585835-24585835 on Build GRCh38
rs184846029 Chr.10: 24585666-24585666 on Build GRCh38
rs61732945 Chr.10: 24586048-24586048 on Build GRCh38
rs760056699 Chr.10: 24586019-24586019 on Build GRCh38
rs764513682 Chr.10: 24585706-24585706 on Build GRCh38
rs753437547 Chr.10: 24585934-24585934 on Build GRCh38
rs1032427124 Chr.10: 24585889-24585889 on Build GRCh38
rs758268178 Chr.10: 24585951-24585951 on Build GRCh38
rs776553614 Chr.10: 24585751-24585753 on Build GRCh38
rs777478309 Chr.10: 24585753-24585753 on Build GRCh38
rs755000024 Chr.10: 24586043-24586043 on Build GRCh38
rs61735172 Chr.10: 24586065-24586065 on Build GRCh38
rs772083096 Chr.10: 24585681-24585681 on Build GRCh38
rs377732314 Chr.10: 24585857-24585857 on Build GRCh38
rs575521938 Chr.10: 24586078-24586078 on Build GRCh38
rs769917488 Chr.10: 24585999-24585999 on Build GRCh38
rs772786279 Chr.10: 24585958-24585958 on Build GRCh38
rs368758881 Chr.10: 24585890-24585890 on Build GRCh38
rs745839476 Chr.10: 24585777-24585777 on Build GRCh38
rs3210912 Chr.10: 24585969-24585969 on Build GRCh38
rs201341804 Chr.10: 24585974-24585974 on Build GRCh38
rs770150135 Chr.10: 24585880-24585880 on Build GRCh38
rs749095417 Chr.10: 24585754-24585754 on Build GRCh38
rs753556425 Chr.10: 24585838-24585838 on Build GRCh38
rs553103965 Chr.10: 24585970-24585970 on Build GRCh38
rs749603047 Chr.10: 24586013-24586013 on Build GRCh38
rs373721050 Chr.10: 24585872-24585872 on Build GRCh38
rs1020523653 Chr.10: 24585932-24585932 on Build GRCh38
rs745950085 Chr.10: 24585679-24585679 on Build GRCh38
rs781531928 Chr.10: 24585876-24585876 on Build GRCh38
rs757022071 Chr.10: 24585843-24585843 on Build GRCh38
rs772981446 Chr.10: 24586033-24586033 on Build GRCh38
rs567238229 Chr.10: 24585882-24585882 on Build GRCh38
rs61758701 Chr.10: 24585780-24585780 on Build GRCh38
rs749565719 Chr.10: 24586066-24586066 on Build GRCh38
rs762622331 Chr.10: 24586034-24586034 on Build GRCh38
rs778907197 Chr.10: 24585766-24585766 on Build GRCh38
rs757103634 Chr.10: 24585765-24585765 on Build GRCh38
rs746785139 Chr.10: 24586047-24586047 on Build GRCh38
rs764779961 Chr.10: 24586030-24586030 on Build GRCh38
rs768114070 Chr.10: 24586020-24586020 on Build GRCh38
rs768921548 Chr.10: 24585873-24585873 on Build GRCh38
rs756275555 Chr.10: 24586060-24586060 on Build GRCh38
rs747185272 Chr.10: 24585784-24585784 on Build GRCh38
rs760828147 Chr.10: 24585697-24585697 on Build GRCh38
rs778142537 Chr.10: 24586002-24586002 on Build GRCh38
rs372987359 Chr.10: 24585816-24585816 on Build GRCh38
rs763455374 Chr.10: 24585883-24585883 on Build GRCh38
rs201172274 Chr.10: 24585939-24585939 on Build GRCh38
rs763636012 Chr.10: 24585923-24585923 on Build GRCh38
rs754482786 Chr.10: 24585730-24585730 on Build GRCh38
rs780120544 Chr.10: 24585871-24585871 on Build GRCh38
rs370970328 Chr.10: 24585949-24585949 on Build GRCh38
rs61735178 Chr.10: 24585742-24585742 on Build GRCh38
rs957865037 Chr.10: 24585719-24585719 on Build GRCh38
rs373586817 Chr.10: 24585818-24585818 on Build GRCh38
rs748433077 Chr.10: 24585879-24585879 on Build GRCh38
rs1011776588 Chr.10: 24585891-24585891 on Build GRCh38
rs573043562 Chr.10: 24585973-24585973 on Build GRCh38
rs769816744 Chr.10: 24585918-24585920 on Build GRCh38
rs928748345 Chr.10: 24585776-24585776 on Build GRCh38
rs770860414 Chr.10: 24585656-24585656 on Build GRCh38
rs750334535 Chr.10: 24585861-24585861 on Build GRCh38
rs763396585 Chr.10: 24585827-24585827 on Build GRCh38
rs545239109 Chr.10: 24586023-24586023 on Build GRCh38
rs61735171 Chr.10: 24586012-24586012 on Build GRCh38
rs3893301 Chr.10: 24585711-24585711 on Build GRCh38
rs1018504397 Chr.10: 24585651-24585651 on Build GRCh38

View Full Product Details