Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 24589156-24589674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP21
Entrez Gene ID
57584
Gene Name
Rho GTPase activating protein 21
Gene Aliases
ARHGAP10
UniGene
Hs.524195
Gene Chromosome Location
Chr.10: 24583609-24725421 on Build GRCh38
Transcripts
NM_020824.3 XM_005252542.3 XM_005252544.4 XM_011519602.2 XM_011519603.2 XM_011519604.2 XM_011519605.2 XM_011519606.2 XM_011519607.2 XM_017016459.1 XM_017016460.1 XM_017016461.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
373984 c.4156A>G p.T1386A Chr.10: 24589294-24589294 on Build GRCh38
917150 c.4175C>A p.S1392Y Chr.10: 24589275-24589275 on Build GRCh38
6128906 c.4177A>G p.K1393E Chr.10: 24589273-24589273 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747599551 Chr.10: 24589291-24589291 on Build GRCh38
rs893553784 Chr.10: 24589329-24589329 on Build GRCh38
rs749344810 Chr.10: 24589582-24589582 on Build GRCh38
rs7478260 Chr.10: 24589200-24589200 on Build GRCh38
rs561459884 Chr.10: 24589572-24589572 on Build GRCh38
rs571042694 Chr.10: 24589228-24589228 on Build GRCh38
rs946588463 Chr.10: 24589651-24589651 on Build GRCh38
rs759489199 Chr.10: 24589229-24589229 on Build GRCh38
rs767554029 Chr.10: 24589231-24589231 on Build GRCh38
rs753963932 Chr.10: 24589247-24589247 on Build GRCh38
rs950101122 Chr.10: 24589183-24589183 on Build GRCh38
rs1001063886 Chr.10: 24589599-24589599 on Build GRCh38
rs766650790 Chr.10: 24589239-24589239 on Build GRCh38
rs892608530 Chr.10: 24589603-24589603 on Build GRCh38
rs143719758 Chr.10: 24589401-24589401 on Build GRCh38
rs575054584 Chr.10: 24589371-24589371 on Build GRCh38
rs577515932 Chr.10: 24589422-24589422 on Build GRCh38
rs764285525 Chr.10: 24589241-24589241 on Build GRCh38
rs371350723 Chr.10: 24589286-24589286 on Build GRCh38
rs199832079 Chr.10: 24589222-24589222 on Build GRCh38
rs562306389 Chr.10: 24589432-24589432 on Build GRCh38
rs748826962 Chr.10: 24589317-24589317 on Build GRCh38
rs368602349 Chr.10: 24589225-24589225 on Build GRCh38
rs374821928 Chr.10: 24589254-24589254 on Build GRCh38
rs774204952 Chr.10: 24589344-24589344 on Build GRCh38
rs779186887 Chr.10: 24589268-24589268 on Build GRCh38
rs137952198 Chr.10: 24589221-24589221 on Build GRCh38
rs1017090851 Chr.10: 24589542-24589542 on Build GRCh38
rs371581583 Chr.10: 24589242-24589242 on Build GRCh38
rs962752041 Chr.10: 24589547-24589547 on Build GRCh38
rs770721070 Chr.10: 24589335-24589335 on Build GRCh38
rs367939109 Chr.10: 24589274-24589274 on Build GRCh38
rs927864312 Chr.10: 24589580-24589580 on Build GRCh38
rs987960516 Chr.10: 24589587-24589587 on Build GRCh38
rs541494358 Chr.10: 24589571-24589571 on Build GRCh38
rs991960910 Chr.10: 24589552-24589552 on Build GRCh38
rs1025454164 Chr.10: 24589623-24589623 on Build GRCh38
rs534429336 Chr.10: 24589377-24589377 on Build GRCh38
rs1013769846 Chr.10: 24589604-24589604 on Build GRCh38
rs915100677 Chr.10: 24589625-24589625 on Build GRCh38
rs564555557 Chr.10: 24589372-24589373 on Build GRCh38
rs769135627 Chr.10: 24589296-24589296 on Build GRCh38
rs1023064188 Chr.10: 24589553-24589553 on Build GRCh38
rs904235368 Chr.10: 24589586-24589593 on Build GRCh38
rs539999883 Chr.10: 24589220-24589220 on Build GRCh38
rs777877568 Chr.10: 24589534-24589534 on Build GRCh38
rs772812769 Chr.10: 24589298-24589298 on Build GRCh38
rs1042706314 Chr.10: 24589585-24589585 on Build GRCh38
rs756120393 Chr.10: 24589237-24589237 on Build GRCh38
rs760966223 Chr.10: 24589232-24589233 on Build GRCh38
rs968278241 Chr.10: 24589203-24589203 on Build GRCh38
rs367703958 Chr.10: 24589362-24589362 on Build GRCh38
rs10159665 Chr.10: 24589570-24589570 on Build GRCh38
rs1039352829 Chr.10: 24589393-24589393 on Build GRCh38
rs370748087 Chr.10: 24589245-24589245 on Build GRCh38
rs931223347 Chr.10: 24589506-24589506 on Build GRCh38
rs1041174787 Chr.10: 24589575-24589575 on Build GRCh38
rs969768480 Chr.10: 24589465-24589465 on Build GRCh38
rs887361121 Chr.10: 24589481-24589481 on Build GRCh38
rs192357110 Chr.10: 24589584-24589584 on Build GRCh38
rs919810172 Chr.10: 24589505-24589505 on Build GRCh38
rs186866784 Chr.10: 24589423-24589423 on Build GRCh38

View Full Product Details