Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 26502822-26503331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBB1IP
Entrez Gene ID
54518
Gene Name
amyloid beta precursor protein binding family B member 1 interacting protein
Gene Aliases
INAG1, PREL1, RARP1, RIAM
UniGene
Hs.310421
Gene Chromosome Location
Chr.10: 26438324-26567879 on Build GRCh38
Transcripts
NM_019043.3 XM_006717451.2 XM_011519514.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4548711 c.454G>A p.E152K Chr.10: 26503197-26503197 on Build GRCh38
4550962 c.514G>A p.E172K Chr.10: 26503257-26503257 on Build GRCh38
7329842 c.454-1G>C p.? Chr.10: 26503196-26503196 on Build GRCh38
4144611 c.457G>T p.E153* Chr.10: 26503200-26503200 on Build GRCh38
389812 c.498G>C p.A166A Chr.10: 26503241-26503241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970292311 Chr.10: 26503052-26503052 on Build GRCh38
rs547814198 Chr.10: 26502973-26502973 on Build GRCh38
rs890422594 Chr.10: 26503075-26503075 on Build GRCh38
rs760190593 Chr.10: 26503185-26503186 on Build GRCh38
rs117705625 Chr.10: 26503241-26503241 on Build GRCh38
rs201024500 Chr.10: 26503171-26503171 on Build GRCh38
rs983461003 Chr.10: 26502859-26502859 on Build GRCh38
rs763733371 Chr.10: 26503303-26503303 on Build GRCh38
rs891443261 Chr.10: 26503006-26503006 on Build GRCh38
rs1008910764 Chr.10: 26503108-26503108 on Build GRCh38
rs770112760 Chr.10: 26503264-26503264 on Build GRCh38
rs757019430 Chr.10: 26503213-26503213 on Build GRCh38
rs113442503 Chr.10: 26503215-26503215 on Build GRCh38
rs1052736320 Chr.10: 26503001-26503001 on Build GRCh38
rs189390747 Chr.10: 26503150-26503150 on Build GRCh38
rs560536763 Chr.10: 26502871-26502871 on Build GRCh38
rs773603193 Chr.10: 26503161-26503161 on Build GRCh38
rs1007902183 Chr.10: 26502996-26502996 on Build GRCh38
rs141086430 Chr.10: 26503064-26503064 on Build GRCh38
rs759103707 Chr.10: 26503164-26503164 on Build GRCh38
rs763593686 Chr.10: 26503281-26503281 on Build GRCh38
rs765707416 Chr.10: 26503154-26503154 on Build GRCh38
rs939940843 Chr.10: 26502862-26502862 on Build GRCh38
rs995356972 Chr.10: 26503122-26503122 on Build GRCh38
rs761585909 Chr.10: 26503110-26503110 on Build GRCh38
rs61758872 Chr.10: 26503220-26503220 on Build GRCh38
rs907849215 Chr.10: 26502861-26502861 on Build GRCh38
rs112496589 Chr.10: 26503041-26503041 on Build GRCh38
rs532215937 Chr.10: 26502874-26502874 on Build GRCh38
rs371510843 Chr.10: 26503145-26503145 on Build GRCh38
rs555032115 Chr.10: 26503288-26503288 on Build GRCh38
rs538523075 Chr.10: 26503251-26503251 on Build GRCh38
rs926930309 Chr.10: 26502864-26502864 on Build GRCh38
rs753625491 Chr.10: 26503181-26503181 on Build GRCh38
rs531622607 Chr.10: 26502942-26502942 on Build GRCh38
rs767202528 Chr.10: 26503165-26503165 on Build GRCh38
rs932473645 Chr.10: 26502944-26502944 on Build GRCh38
rs760290851 Chr.10: 26503297-26503297 on Build GRCh38
rs1015374728 Chr.10: 26503109-26503109 on Build GRCh38
rs149680034 Chr.10: 26502868-26502868 on Build GRCh38
rs868260118 Chr.10: 26503194-26503194 on Build GRCh38
rs370433845 Chr.10: 26503148-26503148 on Build GRCh38
rs199955004 Chr.10: 26503283-26503283 on Build GRCh38
rs547791562 Chr.10: 26502896-26502896 on Build GRCh38
rs747115059 Chr.10: 26503226-26503226 on Build GRCh38
rs113416020 Chr.10: 26503035-26503035 on Build GRCh38
rs145472585 Chr.10: 26502986-26502986 on Build GRCh38
rs567415168 Chr.10: 26502916-26502916 on Build GRCh38
rs117439316 Chr.10: 26502949-26502949 on Build GRCh38
rs768828226 Chr.10: 26503238-26503238 on Build GRCh38
rs762116430 Chr.10: 26503151-26503151 on Build GRCh38
rs752362366 Chr.10: 26503170-26503170 on Build GRCh38
rs1049539343 Chr.10: 26502975-26502975 on Build GRCh38
rs139379193 Chr.10: 26503259-26503259 on Build GRCh38
rs562685048 Chr.10: 26502933-26502933 on Build GRCh38
rs969543324 Chr.10: 26503156-26503156 on Build GRCh38
rs912987384 Chr.10: 26502924-26502924 on Build GRCh38
rs780157280 Chr.10: 26503225-26503225 on Build GRCh38
rs773764941 Chr.10: 26503275-26503275 on Build GRCh38

View Full Product Details