Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 27196690-27197193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACBD5
Entrez Gene ID
91452
Gene Name
acyl-CoA binding domain containing 5
Gene Aliases
-
UniGene
Hs.530597
Gene Chromosome Location
Chr.10: 27182624-27248674 on Build GRCh38
Transcripts
NM_001042473.3 NM_001271512.3 NM_001301251.1 NM_001301252.1 NM_001301253.1 NM_001301254.1 NM_145698.4 XM_006717528.3 XM_006717530.3 XM_006717531.2 XM_006717535.2 XM_017016884.1 XM_017016885.1 XM_017016886.1 XM_017016887.1 XM_017016888.1 XM_017016889.1 XM_017016890.1 XM_017016891.1 XM_017016892.1 XM_017016893.1 XM_017016894.1 XM_017016895.1 XM_017016896.1 XM_017016897.1 XM_017016898.1 XM_017016899.1 XM_017016900.1 XM_017016901.1 XM_017016902.1 XM_017016903.1 XM_017016904.1 XM_017016905.1 XM_017016906.1 XM_017016907.1 XM_017016908.1

SNP Detail

SNP ID Chromosome Location
rs142191359 Chr.10: 27197049-27197050 on Build GRCh38
rs896401562 Chr.10: 27196728-27196728 on Build GRCh38
rs58351748 Chr.10: 27196785-27196785 on Build GRCh38
rs758991531 Chr.10: 27197016-27197019 on Build GRCh38
rs754129843 Chr.10: 27197094-27197094 on Build GRCh38
rs756991490 Chr.10: 27197097-27197097 on Build GRCh38
rs957472302 Chr.10: 27196923-27196923 on Build GRCh38
rs765043505 Chr.10: 27197142-27197142 on Build GRCh38
rs562538847 Chr.10: 27196950-27196950 on Build GRCh38
rs777042908 Chr.10: 27196766-27196766 on Build GRCh38
rs994000304 Chr.10: 27196851-27196851 on Build GRCh38
rs763585379 Chr.10: 27196937-27196937 on Build GRCh38
rs998667133 Chr.10: 27196878-27196878 on Build GRCh38
rs764555246 Chr.10: 27197050-27197050 on Build GRCh38
rs1031957532 Chr.10: 27196904-27196904 on Build GRCh38
rs765656824 Chr.10: 27197157-27197157 on Build GRCh38
rs1027660727 Chr.10: 27196944-27196944 on Build GRCh38
rs976098668 Chr.10: 27197006-27197006 on Build GRCh38
rs952047530 Chr.10: 27196947-27196947 on Build GRCh38
rs983630209 Chr.10: 27197051-27197051 on Build GRCh38
rs937700286 Chr.10: 27197123-27197123 on Build GRCh38
rs577069442 Chr.10: 27196817-27196817 on Build GRCh38
rs12356269 Chr.10: 27196972-27196972 on Build GRCh38
rs990255042 Chr.10: 27196966-27196966 on Build GRCh38
rs753496558 Chr.10: 27197011-27197011 on Build GRCh38
rs763722858 Chr.10: 27196979-27196979 on Build GRCh38
rs979185553 Chr.10: 27197054-27197054 on Build GRCh38
rs773806609 Chr.10: 27196916-27196916 on Build GRCh38
rs966680513 Chr.10: 27197008-27197008 on Build GRCh38
rs762440079 Chr.10: 27197140-27197140 on Build GRCh38
rs182263760 Chr.10: 27196978-27196978 on Build GRCh38
rs1055051033 Chr.10: 27196724-27196724 on Build GRCh38
rs545915475 Chr.10: 27196818-27196818 on Build GRCh38
rs763577436 Chr.10: 27197048-27197049 on Build GRCh38
rs76683151 Chr.10: 27196770-27196770 on Build GRCh38
rs924986979 Chr.10: 27197064-27197064 on Build GRCh38
rs976304116 Chr.10: 27197016-27197020 on Build GRCh38

View Full Product Details