Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 27812466-27812976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC4
Entrez Gene ID
55130
Gene Name
armadillo repeat containing 4
Gene Aliases
CILD23
UniGene
Hs.127530
Gene Chromosome Location
Chr.10: 27796355-27999050 on Build GRCh38
Transcripts
NM_001290020.1 NM_001290021.1 NM_001312689.1 NM_018076.4 XM_011519527.1 XM_011519528.1 XM_011519533.2 XM_011519534.2 XM_011519535.2 XM_011519537.2 XM_017016369.1 XM_017016370.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
343172 c.3028C>A p.L1010M Chr.10: 27812619-27812619 on Build GRCh38
3436979 c.3124A>G p.R1042G Chr.10: 27812523-27812523 on Build GRCh38
3436980 c.3094C>T p.R1032C Chr.10: 27812553-27812553 on Build GRCh38
1176781 c.3134G>T p.*1045L Chr.10: 27812513-27812513 on Build GRCh38
348234 c.3089A>T p.N1030I Chr.10: 27812558-27812558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767489618 Chr.10: 27812534-27812534 on Build GRCh38
rs554115182 Chr.10: 27812580-27812580 on Build GRCh38
rs12248984 Chr.10: 27812821-27812821 on Build GRCh38
rs769683129 Chr.10: 27812498-27812498 on Build GRCh38
rs1013973913 Chr.10: 27812852-27812852 on Build GRCh38
rs1008305700 Chr.10: 27812770-27812770 on Build GRCh38
rs770927145 Chr.10: 27812518-27812518 on Build GRCh38
rs542704930 Chr.10: 27812684-27812684 on Build GRCh38
rs184054538 Chr.10: 27812510-27812510 on Build GRCh38
rs200934236 Chr.10: 27812539-27812539 on Build GRCh38
rs546928912 Chr.10: 27812578-27812578 on Build GRCh38
rs747727153 Chr.10: 27812491-27812491 on Build GRCh38
rs547801110 Chr.10: 27812941-27812941 on Build GRCh38
rs764233304 Chr.10: 27812597-27812597 on Build GRCh38
rs1031541937 Chr.10: 27812874-27812874 on Build GRCh38
rs3737184 Chr.10: 27812526-27812526 on Build GRCh38
rs745364784 Chr.10: 27812553-27812553 on Build GRCh38
rs759357387 Chr.10: 27812589-27812589 on Build GRCh38
rs1047853802 Chr.10: 27812728-27812728 on Build GRCh38
rs375622560 Chr.10: 27812552-27812552 on Build GRCh38
rs757762137 Chr.10: 27812543-27812543 on Build GRCh38
rs764460220 Chr.10: 27812541-27812541 on Build GRCh38
rs942323929 Chr.10: 27812735-27812735 on Build GRCh38
rs375535760 Chr.10: 27812637-27812637 on Build GRCh38
rs780400239 Chr.10: 27812865-27812865 on Build GRCh38
rs145919015 Chr.10: 27812514-27812514 on Build GRCh38
rs180998698 Chr.10: 27812749-27812749 on Build GRCh38
rs964719495 Chr.10: 27812683-27812683 on Build GRCh38
rs867897827 Chr.10: 27812586-27812586 on Build GRCh38
rs957238905 Chr.10: 27812571-27812571 on Build GRCh38
rs769070494 Chr.10: 27812938-27812938 on Build GRCh38
rs757155470 Chr.10: 27812673-27812673 on Build GRCh38
rs763015694 Chr.10: 27812517-27812517 on Build GRCh38
rs556416045 Chr.10: 27812548-27812548 on Build GRCh38
rs998695382 Chr.10: 27812868-27812868 on Build GRCh38
rs1046493638 Chr.10: 27812859-27812859 on Build GRCh38
rs774587750 Chr.10: 27812522-27812522 on Build GRCh38
rs909378916 Chr.10: 27812731-27812731 on Build GRCh38
rs1031364988 Chr.10: 27812771-27812771 on Build GRCh38
rs777629169 Chr.10: 27812657-27812657 on Build GRCh38
rs1025464178 Chr.10: 27812544-27812544 on Build GRCh38
rs866397170 Chr.10: 27812617-27812617 on Build GRCh38
rs754157507 Chr.10: 27812542-27812542 on Build GRCh38
rs185279155 Chr.10: 27812928-27812928 on Build GRCh38
rs61843168 Chr.10: 27812754-27812754 on Build GRCh38
rs753002909 Chr.10: 27812535-27812535 on Build GRCh38
rs1055284196 Chr.10: 27812745-27812745 on Build GRCh38
rs138588942 Chr.10: 27812561-27812561 on Build GRCh38
rs968174774 Chr.10: 27812864-27812864 on Build GRCh38
rs1022558400 Chr.10: 27812849-27812849 on Build GRCh38
rs762221801 Chr.10: 27812618-27812618 on Build GRCh38
rs892708100 Chr.10: 27812782-27812782 on Build GRCh38
rs750988855 Chr.10: 27812631-27812631 on Build GRCh38
rs926736331 Chr.10: 27812513-27812513 on Build GRCh38
rs754214367 Chr.10: 27812603-27812603 on Build GRCh38
rs747499395 Chr.10: 27812922-27812922 on Build GRCh38
rs767086472 Chr.10: 27812651-27812651 on Build GRCh38
rs758020238 Chr.10: 27812792-27812792 on Build GRCh38
rs536964995 Chr.10: 27812885-27812885 on Build GRCh38
rs975870334 Chr.10: 27812685-27812685 on Build GRCh38
rs765557199 Chr.10: 27812630-27812630 on Build GRCh38
rs1034179870 Chr.10: 27812879-27812879 on Build GRCh38
rs769530979 Chr.10: 27812567-27812567 on Build GRCh38
rs774495532 Chr.10: 27812584-27812584 on Build GRCh38
rs143968716 Chr.10: 27812920-27812920 on Build GRCh38
rs922991487 Chr.10: 27812711-27812711 on Build GRCh38
rs567161839 Chr.10: 27812722-27812722 on Build GRCh38
rs996693185 Chr.10: 27812595-27812595 on Build GRCh38
rs543579260 Chr.10: 27812853-27812853 on Build GRCh38
rs781294210 Chr.10: 27812644-27812646 on Build GRCh38
rs775378809 Chr.10: 27812593-27812593 on Build GRCh38
rs758783044 Chr.10: 27812634-27812634 on Build GRCh38
rs772239137 Chr.10: 27812591-27812591 on Build GRCh38
rs572398951 Chr.10: 27812653-27812653 on Build GRCh38
rs749034282 Chr.10: 27812579-27812579 on Build GRCh38
rs748946300 Chr.10: 27812662-27812662 on Build GRCh38
rs968226713 Chr.10: 27812492-27812492 on Build GRCh38
rs992557787 Chr.10: 27812949-27812949 on Build GRCh38
rs960847386 Chr.10: 27812887-27812887 on Build GRCh38
rs757325706 Chr.10: 27812638-27812640 on Build GRCh38
rs981225478 Chr.10: 27812494-27812494 on Build GRCh38
rs747861861 Chr.10: 27812565-27812565 on Build GRCh38
rs760987566 Chr.10: 27812596-27812596 on Build GRCh38

View Full Product Details