Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 27935037-27935567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC4
Entrez Gene ID
55130
Gene Name
armadillo repeat containing 4
Gene Aliases
CILD23
UniGene
Hs.127530
Gene Chromosome Location
Chr.10: 27796355-27999050 on Build GRCh38
Transcripts
NM_001290020.1 NM_001290021.1 NM_001312689.1 NM_018076.4 XM_011519527.1 XM_011519528.1 XM_011519533.2 XM_011519534.2 XM_011519535.2 XM_011519537.2 XM_017016368.1 XM_017016369.1 XM_017016370.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1183623 c.2351G>T p.C784F Chr.10: 27935154-27935154 on Build GRCh38
1183625 c.2357A>C p.E786A Chr.10: 27935148-27935148 on Build GRCh38
4897334 c.873A>C p.T291T Chr.10: 27935207-27935207 on Build GRCh38
4861614 c.916G>A p.G306R Chr.10: 27935164-27935164 on Build GRCh38
7356443 c.2342G>T p.G781V Chr.10: 27935163-27935163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758289540 Chr.10: 27935211-27935211 on Build GRCh38
rs757057849 Chr.10: 27935097-27935097 on Build GRCh38
rs747260565 Chr.10: 27935124-27935124 on Build GRCh38
rs755099287 Chr.10: 27935275-27935275 on Build GRCh38
rs936157972 Chr.10: 27935434-27935434 on Build GRCh38
rs760325470 Chr.10: 27935247-27935247 on Build GRCh38
rs200299616 Chr.10: 27935245-27935245 on Build GRCh38
rs1041878021 Chr.10: 27935170-27935170 on Build GRCh38
rs35280610 Chr.10: 27935125-27935125 on Build GRCh38
rs750178697 Chr.10: 27935531-27935531 on Build GRCh38
rs771413885 Chr.10: 27935296-27935296 on Build GRCh38
rs201117549 Chr.10: 27935264-27935264 on Build GRCh38
rs911615349 Chr.10: 27935403-27935403 on Build GRCh38
rs145619075 Chr.10: 27935163-27935163 on Build GRCh38
rs201498403 Chr.10: 27935221-27935221 on Build GRCh38
rs777719165 Chr.10: 27935079-27935079 on Build GRCh38
rs778017230 Chr.10: 27935290-27935290 on Build GRCh38
rs972636695 Chr.10: 27935414-27935414 on Build GRCh38
rs536988483 Chr.10: 27935301-27935301 on Build GRCh38
rs1049280156 Chr.10: 27935478-27935478 on Build GRCh38
rs749731316 Chr.10: 27935292-27935292 on Build GRCh38
rs762152739 Chr.10: 27935142-27935142 on Build GRCh38
rs184602729 Chr.10: 27935300-27935300 on Build GRCh38
rs925725505 Chr.10: 27935067-27935067 on Build GRCh38
rs778743746 Chr.10: 27935189-27935189 on Build GRCh38
rs752423499 Chr.10: 27935069-27935069 on Build GRCh38
rs113146637 Chr.10: 27935412-27935412 on Build GRCh38
rs926640509 Chr.10: 27935527-27935527 on Build GRCh38
rs750518822 Chr.10: 27935065-27935065 on Build GRCh38
rs1024289085 Chr.10: 27935538-27935538 on Build GRCh38
rs922861532 Chr.10: 27935340-27935340 on Build GRCh38
rs112483026 Chr.10: 27935234-27935234 on Build GRCh38
rs894528996 Chr.10: 27935527-27935527 on Build GRCh38
rs143059966 Chr.10: 27935233-27935233 on Build GRCh38
rs573975924 Chr.10: 27935400-27935400 on Build GRCh38
rs563921348 Chr.10: 27935526-27935526 on Build GRCh38
rs35242712 Chr.10: 27935146-27935146 on Build GRCh38
rs778777989 Chr.10: 27935102-27935102 on Build GRCh38
rs376908802 Chr.10: 27935145-27935145 on Build GRCh38
rs200311436 Chr.10: 27935265-27935265 on Build GRCh38
rs946308925 Chr.10: 27935380-27935380 on Build GRCh38
rs748352808 Chr.10: 27935280-27935280 on Build GRCh38
rs144407870 Chr.10: 27935253-27935253 on Build GRCh38
rs554294331 Chr.10: 27935272-27935272 on Build GRCh38
rs766361375 Chr.10: 27935271-27935271 on Build GRCh38
rs117515566 Chr.10: 27935199-27935199 on Build GRCh38
rs767169905 Chr.10: 27935158-27935158 on Build GRCh38
rs780112130 Chr.10: 27935219-27935219 on Build GRCh38
rs985909964 Chr.10: 27935394-27935394 on Build GRCh38
rs772185029 Chr.10: 27935115-27935115 on Build GRCh38
rs781286186 Chr.10: 27935277-27935277 on Build GRCh38
rs975722163 Chr.10: 27935306-27935306 on Build GRCh38
rs867758259 Chr.10: 27935075-27935075 on Build GRCh38
rs267602461 Chr.10: 27935074-27935074 on Build GRCh38
rs753480648 Chr.10: 27935252-27935252 on Build GRCh38
rs553449553 Chr.10: 27935407-27935407 on Build GRCh38
rs763662864 Chr.10: 27935248-27935248 on Build GRCh38
rs745784537 Chr.10: 27935105-27935105 on Build GRCh38
rs368538204 Chr.10: 27935257-27935257 on Build GRCh38
rs7093861 Chr.10: 27935420-27935420 on Build GRCh38
rs571264531 Chr.10: 27935225-27935225 on Build GRCh38
rs761475632 Chr.10: 27935261-27935261 on Build GRCh38
rs755790914 Chr.10: 27935072-27935072 on Build GRCh38
rs1010426754 Chr.10: 27935169-27935169 on Build GRCh38
rs148311329 Chr.10: 27935355-27935355 on Build GRCh38
rs961909404 Chr.10: 27935249-27935249 on Build GRCh38
rs765276317 Chr.10: 27935445-27935445 on Build GRCh38
rs768719701 Chr.10: 27935222-27935222 on Build GRCh38
rs757039976 Chr.10: 27935183-27935183 on Build GRCh38
rs558555708 Chr.10: 27935343-27935343 on Build GRCh38
rs753521915 Chr.10: 27935083-27935083 on Build GRCh38
rs878855047 Chr.10: 27935229-27935229 on Build GRCh38
rs763561367 Chr.10: 27935235-27935235 on Build GRCh38
rs541040117 Chr.10: 27935365-27935365 on Build GRCh38
rs1055889060 Chr.10: 27935526-27935527 on Build GRCh38
rs767114873 Chr.10: 27935062-27935062 on Build GRCh38
rs780057616 Chr.10: 27935116-27935116 on Build GRCh38
rs886041523 Chr.10: 27935099-27935099 on Build GRCh38
rs763471776 Chr.10: 27935151-27935151 on Build GRCh38
rs756295807 Chr.10: 27935284-27935284 on Build GRCh38
rs138219785 Chr.10: 27935171-27935171 on Build GRCh38
rs776992489 Chr.10: 27935136-27935136 on Build GRCh38
rs763894304 Chr.10: 27935166-27935166 on Build GRCh38

View Full Product Details