Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 27939883-27940418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC4
Entrez Gene ID
55130
Gene Name
armadillo repeat containing 4
Gene Aliases
CILD23
UniGene
Hs.127530
Gene Chromosome Location
Chr.10: 27796355-27999050 on Build GRCh38
Transcripts
NM_001290020.1 NM_001290021.1 NM_001312689.1 NM_018076.4 XM_011519527.1 XM_011519528.1 XM_011519533.2 XM_011519534.2 XM_011519535.2 XM_011519537.2 XM_017016368.1 XM_017016369.1 XM_017016370.1 XM_017016371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6604917 c.2059G>A p.E687K Chr.10: 27939935-27939935 on Build GRCh38
3437005 c.2019G>A p.R673R Chr.10: 27939975-27939975 on Build GRCh38
4406394 c.2022C>T p.I674I Chr.10: 27939972-27939972 on Build GRCh38
3437006 c.1987G>A p.E663K Chr.10: 27940007-27940007 on Build GRCh38
6304463 c.2088C>T p.A696A Chr.10: 27939906-27939906 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764698031 Chr.10: 27940372-27940372 on Build GRCh38
rs939722468 Chr.10: 27940051-27940051 on Build GRCh38
rs760463853 Chr.10: 27940088-27940088 on Build GRCh38
rs777280534 Chr.10: 27939941-27939941 on Build GRCh38
rs927020993 Chr.10: 27940330-27940330 on Build GRCh38
rs114515937 Chr.10: 27940043-27940043 on Build GRCh38
rs566342248 Chr.10: 27940277-27940277 on Build GRCh38
rs187023198 Chr.10: 27940033-27940033 on Build GRCh38
rs968836627 Chr.10: 27939964-27939964 on Build GRCh38
rs756813598 Chr.10: 27940044-27940044 on Build GRCh38
rs35472668 Chr.10: 27939997-27939997 on Build GRCh38
rs148613172 Chr.10: 27940259-27940260 on Build GRCh38
rs191449244 Chr.10: 27940360-27940360 on Build GRCh38
rs756833107 Chr.10: 27940247-27940250 on Build GRCh38
rs1044507105 Chr.10: 27940133-27940133 on Build GRCh38
rs965347288 Chr.10: 27940303-27940303 on Build GRCh38
rs775104383 Chr.10: 27939981-27939981 on Build GRCh38
rs748857448 Chr.10: 27939955-27939955 on Build GRCh38
rs931096545 Chr.10: 27940120-27940120 on Build GRCh38
rs772137537 Chr.10: 27940055-27940056 on Build GRCh38
rs748077762 Chr.10: 27939913-27939922 on Build GRCh38
rs556371991 Chr.10: 27939914-27939914 on Build GRCh38
rs762051082 Chr.10: 27939998-27939998 on Build GRCh38
rs530431839 Chr.10: 27940323-27940324 on Build GRCh38
rs1022012517 Chr.10: 27939963-27939963 on Build GRCh38
rs745562035 Chr.10: 27940059-27940059 on Build GRCh38
rs560508452 Chr.10: 27940183-27940184 on Build GRCh38
rs938811413 Chr.10: 27940180-27940180 on Build GRCh38
rs1052986901 Chr.10: 27940101-27940101 on Build GRCh38
rs533778251 Chr.10: 27940322-27940322 on Build GRCh38
rs776290212 Chr.10: 27940285-27940285 on Build GRCh38
rs755772185 Chr.10: 27940383-27940384 on Build GRCh38
rs760664232 Chr.10: 27939989-27939989 on Build GRCh38
rs145519493 Chr.10: 27940261-27940262 on Build GRCh38
rs778581107 Chr.10: 27939960-27939960 on Build GRCh38
rs572132398 Chr.10: 27940020-27940020 on Build GRCh38
rs905546295 Chr.10: 27940158-27940158 on Build GRCh38
rs1013877314 Chr.10: 27940362-27940362 on Build GRCh38
rs755324201 Chr.10: 27940031-27940031 on Build GRCh38
rs7893462 Chr.10: 27939936-27939936 on Build GRCh38
rs60857634 Chr.10: 27940181-27940181 on Build GRCh38
rs1015786773 Chr.10: 27940193-27940194 on Build GRCh38
rs961841523 Chr.10: 27940195-27940195 on Build GRCh38
rs572924819 Chr.10: 27940238-27940239 on Build GRCh38
rs745483471 Chr.10: 27939974-27939974 on Build GRCh38
rs140338410 Chr.10: 27939915-27939915 on Build GRCh38
rs765611160 Chr.10: 27939908-27939908 on Build GRCh38
rs1014239106 Chr.10: 27940193-27940193 on Build GRCh38
rs58108383 Chr.10: 27940262-27940263 on Build GRCh38
rs746366706 Chr.10: 27940255-27940281 on Build GRCh38
rs892061520 Chr.10: 27940192-27940192 on Build GRCh38
rs536574661 Chr.10: 27940391-27940391 on Build GRCh38
rs771920114 Chr.10: 27939980-27939980 on Build GRCh38
rs943882859 Chr.10: 27940100-27940100 on Build GRCh38
rs995637954 Chr.10: 27940221-27940221 on Build GRCh38
rs10658729 Chr.10: 27940265-27940266 on Build GRCh38
rs961279309 Chr.10: 27940323-27940323 on Build GRCh38
rs532572855 Chr.10: 27940209-27940209 on Build GRCh38
rs368642943 Chr.10: 27940026-27940026 on Build GRCh38
rs752140248 Chr.10: 27939923-27939923 on Build GRCh38
rs758055274 Chr.10: 27940173-27940174 on Build GRCh38
rs1008006731 Chr.10: 27940314-27940314 on Build GRCh38
rs201801775 Chr.10: 27940261-27940261 on Build GRCh38
rs549995505 Chr.10: 27940082-27940082 on Build GRCh38
rs904496502 Chr.10: 27940263-27940263 on Build GRCh38
rs953921353 Chr.10: 27940301-27940301 on Build GRCh38
rs1010454703 Chr.10: 27940242-27940242 on Build GRCh38
rs765376476 Chr.10: 27940007-27940007 on Build GRCh38
rs74937547 Chr.10: 27940281-27940281 on Build GRCh38
rs751814479 Chr.10: 27940027-27940027 on Build GRCh38
rs267602462 Chr.10: 27939975-27939975 on Build GRCh38
rs1029474029 Chr.10: 27940292-27940292 on Build GRCh38
rs770427120 Chr.10: 27939956-27939956 on Build GRCh38
rs375053618 Chr.10: 27940045-27940045 on Build GRCh38
rs985430994 Chr.10: 27940296-27940297 on Build GRCh38
rs747821786 Chr.10: 27940145-27940145 on Build GRCh38
rs777594102 Chr.10: 27940176-27940179 on Build GRCh38
rs895453171 Chr.10: 27940360-27940361 on Build GRCh38
rs548089738 Chr.10: 27940271-27940271 on Build GRCh38
rs145103626 Chr.10: 27940194-27940195 on Build GRCh38
rs919545760 Chr.10: 27940108-27940108 on Build GRCh38
rs973037949 Chr.10: 27940353-27940353 on Build GRCh38
rs10658728 Chr.10: 27940257-27940258 on Build GRCh38
rs1035929134 Chr.10: 27939911-27939911 on Build GRCh38
rs547434035 Chr.10: 27939952-27939952 on Build GRCh38

View Full Product Details