Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 35009115-35009620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL2
Entrez Gene ID
8453
Gene Name
cullin 2
Gene Aliases
-
UniGene
Hs.82919
Gene Chromosome Location
Chr.10: 35008551-35126899 on Build GRCh38
Transcripts
NM_001198777.1 NM_001198778.1 NM_001198779.1 NM_001324375.1 NM_001324376.1 NM_003591.3 XM_011519743.1 XM_011519744.1 XM_011519745.1 XM_011519747.1

SNP Detail

SNP ID Chromosome Location
rs908245392 Chr.10: 35009466-35009466 on Build GRCh38
rs909117271 Chr.10: 35009157-35009157 on Build GRCh38
rs902941128 Chr.10: 35009267-35009267 on Build GRCh38
rs1009997342 Chr.10: 35009220-35009221 on Build GRCh38
rs1035399689 Chr.10: 35009330-35009330 on Build GRCh38
rs930015268 Chr.10: 35009189-35009189 on Build GRCh38
rs919849853 Chr.10: 35009355-35009355 on Build GRCh38
rs201027415 Chr.10: 35009515-35009515 on Build GRCh38
rs938185006 Chr.10: 35009494-35009494 on Build GRCh38
rs12263295 Chr.10: 35009314-35009314 on Build GRCh38
rs1019451681 Chr.10: 35009238-35009238 on Build GRCh38
rs954760920 Chr.10: 35009228-35009228 on Build GRCh38
rs371645683 Chr.10: 35009183-35009184 on Build GRCh38
rs12263376 Chr.10: 35009436-35009436 on Build GRCh38
rs918699669 Chr.10: 35009187-35009187 on Build GRCh38
rs1046780722 Chr.10: 35009184-35009204 on Build GRCh38
rs528553981 Chr.10: 35009160-35009160 on Build GRCh38
rs1026097656 Chr.10: 35009418-35009418 on Build GRCh38
rs890495510 Chr.10: 35009578-35009578 on Build GRCh38
rs947091065 Chr.10: 35009242-35009242 on Build GRCh38
rs1046536784 Chr.10: 35009200-35009203 on Build GRCh38
rs894069239 Chr.10: 35009542-35009542 on Build GRCh38
rs893772154 Chr.10: 35009214-35009214 on Build GRCh38
rs942858601 Chr.10: 35009514-35009514 on Build GRCh38
rs893363299 Chr.10: 35009233-35009233 on Build GRCh38
rs1021028185 Chr.10: 35009226-35009229 on Build GRCh38
rs533201417 Chr.10: 35009439-35009439 on Build GRCh38
rs975318242 Chr.10: 35009309-35009309 on Build GRCh38
rs185382012 Chr.10: 35009280-35009280 on Build GRCh38
rs755457474 Chr.10: 35009528-35009528 on Build GRCh38
rs975555288 Chr.10: 35009545-35009545 on Build GRCh38
rs942580746 Chr.10: 35009153-35009153 on Build GRCh38
rs544343001 Chr.10: 35009394-35009394 on Build GRCh38
rs905328902 Chr.10: 35009493-35009493 on Build GRCh38
rs912519663 Chr.10: 35009186-35009223 on Build GRCh38
rs181523690 Chr.10: 35009185-35009185 on Build GRCh38
rs1020956178 Chr.10: 35009570-35009570 on Build GRCh38
rs575711208 Chr.10: 35009326-35009326 on Build GRCh38
rs1040006901 Chr.10: 35009222-35009222 on Build GRCh38
rs912636906 Chr.10: 35009510-35009510 on Build GRCh38
rs951408838 Chr.10: 35009463-35009463 on Build GRCh38
rs540372788 Chr.10: 35009560-35009560 on Build GRCh38
rs962714642 Chr.10: 35009159-35009159 on Build GRCh38
rs12263386 Chr.10: 35009566-35009566 on Build GRCh38
rs942826019 Chr.10: 35009217-35009217 on Build GRCh38
rs1016149991 Chr.10: 35009376-35009376 on Build GRCh38
rs1041738803 Chr.10: 35009246-35009246 on Build GRCh38
rs564512159 Chr.10: 35009434-35009434 on Build GRCh38
rs974106081 Chr.10: 35009396-35009396 on Build GRCh38
rs1046168224 Chr.10: 35009487-35009487 on Build GRCh38
rs58058299 Chr.10: 35009201-35009202 on Build GRCh38
rs546580382 Chr.10: 35009486-35009486 on Build GRCh38
rs1033007202 Chr.10: 35009204-35009204 on Build GRCh38
rs912773150 Chr.10: 35009203-35009203 on Build GRCh38
rs59269668 Chr.10: 35009513-35009514 on Build GRCh38
rs202174023 Chr.10: 35009200-35009200 on Build GRCh38
rs560449269 Chr.10: 35009496-35009496 on Build GRCh38
rs35427253 Chr.10: 35009211-35009212 on Build GRCh38
rs371440945 Chr.10: 35009361-35009361 on Build GRCh38
rs930976629 Chr.10: 35009184-35009202 on Build GRCh38
rs1051178983 Chr.10: 35009202-35009202 on Build GRCh38
rs61841046 Chr.10: 35009342-35009342 on Build GRCh38
rs919600883 Chr.10: 35009181-35009182 on Build GRCh38
rs397975596 Chr.10: 35009514-35009515 on Build GRCh38
rs200538637 Chr.10: 35009201-35009201 on Build GRCh38
rs912425948 Chr.10: 35009148-35009148 on Build GRCh38
rs3065352 Chr.10: 35009221-35009222 on Build GRCh38
rs1002455307 Chr.10: 35009202-35009209 on Build GRCh38
rs894338094 Chr.10: 35009333-35009333 on Build GRCh38

View Full Product Details