Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 35127364-35127810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREM
Entrez Gene ID
1390
Gene Name
cAMP responsive element modulator
Gene Aliases
CREM-2, ICER, hCREM-2
UniGene
Hs.200250
Gene Chromosome Location
Chr.10: 35126841-35212958 on Build GRCh38
Transcripts
NM_001267562.1 NM_001267563.1 NM_001881.3 NM_181571.2 NM_183013.2 NM_183060.2 NR_051973.1 NR_051974.1 NR_051975.1 XM_006717378.3 XM_006717379.3 XM_006717382.3 XM_006717383.3 XM_006717387.3 XM_011519316.2 XM_011519324.2 XM_011519325.2 XM_011519327.2 XM_017015716.1 XM_017015717.1 XM_017015718.1 XM_017015719.1 XM_017015720.1 XM_017015721.1 XM_017015722.1 XM_017015723.1 XM_017015724.1 XM_017015725.1 XM_017015726.1 XM_017015727.1 XM_017015728.1 XM_017015729.1 XM_017015731.1 XM_017015732.1 XM_017015733.1 XM_017015734.1 XM_017015735.1 XM_017015736.1 XM_017015737.1

SNP Detail

SNP ID Chromosome Location
rs193196210 Chr.10: 35127527-35127527 on Build GRCh38
rs12764516 Chr.10: 35127559-35127559 on Build GRCh38
rs1030182782 Chr.10: 35127385-35127385 on Build GRCh38
rs922492889 Chr.10: 35127461-35127461 on Build GRCh38
rs958326078 Chr.10: 35127662-35127662 on Build GRCh38
rs375293438 Chr.10: 35127682-35127682 on Build GRCh38
rs991217729 Chr.10: 35127663-35127663 on Build GRCh38
rs1011732509 Chr.10: 35127589-35127589 on Build GRCh38
rs971919033 Chr.10: 35127700-35127700 on Build GRCh38
rs994817704 Chr.10: 35127494-35127494 on Build GRCh38
rs942646247 Chr.10: 35127753-35127753 on Build GRCh38
rs948533069 Chr.10: 35127388-35127388 on Build GRCh38
rs188368112 Chr.10: 35127422-35127422 on Build GRCh38
rs770439688 Chr.10: 35127782-35127782 on Build GRCh38
rs1038566034 Chr.10: 35127732-35127732 on Build GRCh38
rs925771272 Chr.10: 35127591-35127591 on Build GRCh38
rs1020981062 Chr.10: 35127590-35127590 on Build GRCh38
rs1052823786 Chr.10: 35127408-35127408 on Build GRCh38
rs933618037 Chr.10: 35127643-35127643 on Build GRCh38
rs961688882 Chr.10: 35127483-35127483 on Build GRCh38
rs1018461240 Chr.10: 35127394-35127394 on Build GRCh38
rs1051093982 Chr.10: 35127523-35127523 on Build GRCh38
rs139926892 Chr.10: 35127744-35127744 on Build GRCh38
rs12764861 Chr.10: 35127546-35127546 on Build GRCh38
rs570607986 Chr.10: 35127431-35127431 on Build GRCh38
rs937086486 Chr.10: 35127454-35127454 on Build GRCh38
rs933259854 Chr.10: 35127723-35127723 on Build GRCh38
rs550758457 Chr.10: 35127413-35127413 on Build GRCh38
rs12764854 Chr.10: 35127533-35127533 on Build GRCh38
rs893206059 Chr.10: 35127764-35127764 on Build GRCh38
rs1007098236 Chr.10: 35127392-35127392 on Build GRCh38
rs967476286 Chr.10: 35127432-35127432 on Build GRCh38
rs539575006 Chr.10: 35127505-35127505 on Build GRCh38
rs928428509 Chr.10: 35127440-35127440 on Build GRCh38
rs183916397 Chr.10: 35127579-35127579 on Build GRCh38
rs535416716 Chr.10: 35127592-35127592 on Build GRCh38
rs997958551 Chr.10: 35127416-35127416 on Build GRCh38
rs957484954 Chr.10: 35127396-35127396 on Build GRCh38
rs12765038 Chr.10: 35127585-35127585 on Build GRCh38
rs891147390 Chr.10: 35127747-35127747 on Build GRCh38
rs575506820 Chr.10: 35127675-35127675 on Build GRCh38
rs1036702483 Chr.10: 35127504-35127504 on Build GRCh38
rs1017608409 Chr.10: 35127465-35127465 on Build GRCh38
rs1049515112 Chr.10: 35127649-35127649 on Build GRCh38
rs930005516 Chr.10: 35127521-35127521 on Build GRCh38
rs941850605 Chr.10: 35127496-35127496 on Build GRCh38
rs1053427616 Chr.10: 35127628-35127628 on Build GRCh38
rs12765063 Chr.10: 35127641-35127641 on Build GRCh38
rs976724734 Chr.10: 35127430-35127430 on Build GRCh38
rs981550931 Chr.10: 35127604-35127604 on Build GRCh38
rs955438006 Chr.10: 35127390-35127390 on Build GRCh38
rs537262976 Chr.10: 35127384-35127384 on Build GRCh38
rs909052593 Chr.10: 35127489-35127489 on Build GRCh38
rs919201035 Chr.10: 35127703-35127703 on Build GRCh38
rs981114476 Chr.10: 35127438-35127438 on Build GRCh38
rs916635226 Chr.10: 35127553-35127553 on Build GRCh38
rs1005713131 Chr.10: 35127731-35127732 on Build GRCh38
rs1049483272 Chr.10: 35127418-35127418 on Build GRCh38
rs777421038 Chr.10: 35127725-35127725 on Build GRCh38
rs552675188 Chr.10: 35127506-35127506 on Build GRCh38
rs1055439477 Chr.10: 35127457-35127457 on Build GRCh38
rs1039837279 Chr.10: 35127756-35127756 on Build GRCh38
rs941888839 Chr.10: 35127691-35127691 on Build GRCh38
rs1035209986 Chr.10: 35127765-35127765 on Build GRCh38
rs780608497 Chr.10: 35127562-35127562 on Build GRCh38

View Full Product Details