Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 35148326-35148837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREM
Entrez Gene ID
1390
Gene Name
cAMP responsive element modulator
Gene Aliases
CREM-2, ICER, hCREM-2
UniGene
Hs.200250
Gene Chromosome Location
Chr.10: 35126841-35212958 on Build GRCh38
Transcripts
NM_001267562.1 NM_001267563.1 NM_001881.3 NM_181571.2 NM_183011.1 NM_183012.1 NM_183013.2 NM_183060.2 NR_051973.1 NR_051974.1 NR_051975.1 XM_006717378.3 XM_006717379.3 XM_006717382.3 XM_006717383.3 XM_006717387.3 XM_011519316.2 XM_011519324.2 XM_011519325.2 XM_011519327.2 XM_017015716.1 XM_017015717.1 XM_017015718.1 XM_017015719.1 XM_017015720.1 XM_017015721.1 XM_017015722.1 XM_017015723.1 XM_017015724.1 XM_017015725.1 XM_017015726.1 XM_017015727.1 XM_017015728.1 XM_017015729.1 XM_017015730.1 XM_017015731.1 XM_017015732.1 XM_017015733.1 XM_017015734.1 XM_017015735.1 XM_017015736.1 XM_017015737.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4468570 c.154C>T p.P52S Chr.10: 35148477-35148477 on Build GRCh38
5632840 c.13A>G p.T5A Chr.10: 35148384-35148384 on Build GRCh38
3437451 c.153C>T p.I51I Chr.10: 35148476-35148476 on Build GRCh38
5501053 c.25C>A p.Q9K Chr.10: 35148396-35148396 on Build GRCh38
5501054 c.73C>A p.Q25K Chr.10: 35148396-35148396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048677810 Chr.10: 35148661-35148661 on Build GRCh38
rs372536835 Chr.10: 35148390-35148390 on Build GRCh38
rs146537690 Chr.10: 35148689-35148689 on Build GRCh38
rs747859431 Chr.10: 35148405-35148405 on Build GRCh38
rs763720077 Chr.10: 35148685-35148685 on Build GRCh38
rs749188389 Chr.10: 35148418-35148418 on Build GRCh38
rs765632130 Chr.10: 35148511-35148511 on Build GRCh38
rs761043447 Chr.10: 35148490-35148490 on Build GRCh38
rs769118584 Chr.10: 35148796-35148796 on Build GRCh38
rs949445153 Chr.10: 35148550-35148550 on Build GRCh38
rs996142176 Chr.10: 35148787-35148787 on Build GRCh38
rs753887560 Chr.10: 35148504-35148504 on Build GRCh38
rs529569867 Chr.10: 35148751-35148751 on Build GRCh38
rs144195292 Chr.10: 35148595-35148595 on Build GRCh38
rs372937921 Chr.10: 35148523-35148523 on Build GRCh38
rs963758729 Chr.10: 35148476-35148476 on Build GRCh38
rs901778601 Chr.10: 35148354-35148354 on Build GRCh38
rs746010683 Chr.10: 35148629-35148629 on Build GRCh38
rs749052615 Chr.10: 35148481-35148482 on Build GRCh38
rs927003233 Chr.10: 35148638-35148638 on Build GRCh38
rs769292046 Chr.10: 35148408-35148408 on Build GRCh38
rs758752754 Chr.10: 35148516-35148516 on Build GRCh38
rs191495090 Chr.10: 35148508-35148508 on Build GRCh38
rs187417913 Chr.10: 35148387-35148387 on Build GRCh38
rs756240114 Chr.10: 35148368-35148368 on Build GRCh38
rs775841250 Chr.10: 35148460-35148460 on Build GRCh38
rs765287917 Chr.10: 35148442-35148442 on Build GRCh38
rs377329761 Chr.10: 35148747-35148747 on Build GRCh38
rs576903713 Chr.10: 35148409-35148409 on Build GRCh38
rs893007328 Chr.10: 35148668-35148668 on Build GRCh38
rs377066017 Chr.10: 35148531-35148531 on Build GRCh38
rs372126215 Chr.10: 35148451-35148451 on Build GRCh38
rs546079469 Chr.10: 35148440-35148440 on Build GRCh38
rs940412922 Chr.10: 35148690-35148690 on Build GRCh38
rs530953645 Chr.10: 35148521-35148521 on Build GRCh38
rs777286096 Chr.10: 35148529-35148529 on Build GRCh38
rs770121371 Chr.10: 35148652-35148652 on Build GRCh38
rs764325669 Chr.10: 35148372-35148372 on Build GRCh38
rs963788998 Chr.10: 35148780-35148780 on Build GRCh38
rs146676068 Chr.10: 35148447-35148447 on Build GRCh38
rs975123236 Chr.10: 35148783-35148783 on Build GRCh38
rs377042226 Chr.10: 35148422-35148422 on Build GRCh38
rs369013471 Chr.10: 35148517-35148517 on Build GRCh38
rs376014027 Chr.10: 35148489-35148489 on Build GRCh38
rs996477986 Chr.10: 35148401-35148401 on Build GRCh38
rs750988776 Chr.10: 35148512-35148512 on Build GRCh38
rs547288161 Chr.10: 35148650-35148650 on Build GRCh38
rs368835528 Chr.10: 35148373-35148373 on Build GRCh38
rs759370583 Chr.10: 35148450-35148450 on Build GRCh38
rs1000209558 Chr.10: 35148807-35148807 on Build GRCh38
rs1011360928 Chr.10: 35148675-35148675 on Build GRCh38
rs544299294 Chr.10: 35148579-35148579 on Build GRCh38
rs34227693 Chr.10: 35148400-35148400 on Build GRCh38
rs369290785 Chr.10: 35148446-35148446 on Build GRCh38
rs919665725 Chr.10: 35148789-35148789 on Build GRCh38
rs762602452 Chr.10: 35148655-35148655 on Build GRCh38
rs182779938 Chr.10: 35148364-35148364 on Build GRCh38
rs564145610 Chr.10: 35148592-35148592 on Build GRCh38
rs764201167 Chr.10: 35148494-35148494 on Build GRCh38
rs901661290 Chr.10: 35148776-35148776 on Build GRCh38
rs146013089 Chr.10: 35148644-35148645 on Build GRCh38
rs752652323 Chr.10: 35148634-35148634 on Build GRCh38
rs775818985 Chr.10: 35148674-35148674 on Build GRCh38
rs780803968 Chr.10: 35148403-35148403 on Build GRCh38
rs775879845 Chr.10: 35148439-35148441 on Build GRCh38
rs757719946 Chr.10: 35148380-35148380 on Build GRCh38
rs754364248 Chr.10: 35148378-35148378 on Build GRCh38
rs1007906588 Chr.10: 35148714-35148714 on Build GRCh38
rs549345224 Chr.10: 35148797-35148797 on Build GRCh38

View Full Product Details