Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 35167402-35167864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREM
Entrez Gene ID
1390
Gene Name
cAMP responsive element modulator
Gene Aliases
CREM-2, ICER, hCREM-2
UniGene
Hs.200250
Gene Chromosome Location
Chr.10: 35126841-35212958 on Build GRCh38
Transcripts
NM_001267562.1 NM_001267563.1 NM_001267564.1 NM_001267565.1 NM_001881.3 NM_181571.2 NM_182771.1 NM_182772.1 NM_183011.1 NM_183012.1 NM_183013.2 NM_183060.2 NR_051973.1 NR_051974.1 NR_051975.1 XM_006717378.3 XM_006717379.3 XM_006717382.3 XM_006717383.3 XM_006717387.3 XM_011519316.2 XM_011519324.2 XM_011519325.2 XM_011519327.2 XM_017015716.1 XM_017015717.1 XM_017015718.1 XM_017015719.1 XM_017015720.1 XM_017015721.1 XM_017015722.1 XM_017015723.1 XM_017015724.1 XM_017015725.1 XM_017015726.1 XM_017015727.1 XM_017015728.1 XM_017015729.1 XM_017015730.1 XM_017015731.1 XM_017015732.1 XM_017015733.1 XM_017015734.1 XM_017015735.1 XM_017015736.1 XM_017015737.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5529211 c.56C>A p.S19Y Chr.10: 35167781-35167781 on Build GRCh38
6397732 c.16C>A p.H6N Chr.10: 35167741-35167741 on Build GRCh38
4816710 c.30C>T p.F10F Chr.10: 35167755-35167755 on Build GRCh38
1347688 c.35G>A p.R12H Chr.10: 35167760-35167760 on Build GRCh38
1675161 c.73G>T p.E25* Chr.10: 35167798-35167798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs917106729 Chr.10: 35167651-35167651 on Build GRCh38
rs373218984 Chr.10: 35167776-35167776 on Build GRCh38
rs114159711 Chr.10: 35167820-35167820 on Build GRCh38
rs747332085 Chr.10: 35167594-35167595 on Build GRCh38
rs894296556 Chr.10: 35167631-35167631 on Build GRCh38
rs761778762 Chr.10: 35167711-35167711 on Build GRCh38
rs921416560 Chr.10: 35167492-35167492 on Build GRCh38
rs910112365 Chr.10: 35167679-35167679 on Build GRCh38
rs746769445 Chr.10: 35167815-35167815 on Build GRCh38
rs115669429 Chr.10: 35167476-35167476 on Build GRCh38
rs939965439 Chr.10: 35167526-35167526 on Build GRCh38
rs545836462 Chr.10: 35167672-35167672 on Build GRCh38
rs939419005 Chr.10: 35167605-35167605 on Build GRCh38
rs773297047 Chr.10: 35167835-35167835 on Build GRCh38
rs760932537 Chr.10: 35167697-35167697 on Build GRCh38
rs982200802 Chr.10: 35167663-35167663 on Build GRCh38
rs1056988374 Chr.10: 35167630-35167631 on Build GRCh38
rs574538790 Chr.10: 35167490-35167490 on Build GRCh38
rs777545657 Chr.10: 35167703-35167703 on Build GRCh38
rs563467403 Chr.10: 35167565-35167565 on Build GRCh38
rs768546724 Chr.10: 35167819-35167819 on Build GRCh38
rs978661479 Chr.10: 35167599-35167599 on Build GRCh38
rs760251468 Chr.10: 35167837-35167838 on Build GRCh38
rs772570674 Chr.10: 35167836-35167837 on Build GRCh38
rs988626196 Chr.10: 35167644-35167644 on Build GRCh38
rs767562720 Chr.10: 35167820-35167821 on Build GRCh38
rs745733796 Chr.10: 35167771-35167771 on Build GRCh38
rs9783159 Chr.10: 35167461-35167461 on Build GRCh38
rs1031393791 Chr.10: 35167725-35167725 on Build GRCh38
rs984773399 Chr.10: 35167493-35167493 on Build GRCh38
rs140243641 Chr.10: 35167760-35167760 on Build GRCh38
rs756706885 Chr.10: 35167765-35167765 on Build GRCh38
rs543572164 Chr.10: 35167536-35167536 on Build GRCh38
rs778692142 Chr.10: 35167680-35167680 on Build GRCh38
rs775428299 Chr.10: 35167695-35167695 on Build GRCh38
rs750893395 Chr.10: 35167726-35167726 on Build GRCh38
rs997907064 Chr.10: 35167508-35167508 on Build GRCh38
rs528766180 Chr.10: 35167759-35167759 on Build GRCh38
rs761832555 Chr.10: 35167808-35167810 on Build GRCh38
rs938009012 Chr.10: 35167670-35167670 on Build GRCh38
rs76802222 Chr.10: 35167785-35167785 on Build GRCh38
rs562161172 Chr.10: 35167769-35167769 on Build GRCh38
rs370650746 Chr.10: 35167831-35167831 on Build GRCh38
rs1048426502 Chr.10: 35167671-35167671 on Build GRCh38
rs565426304 Chr.10: 35167507-35167507 on Build GRCh38
rs559322022 Chr.10: 35167736-35167736 on Build GRCh38
rs776954911 Chr.10: 35167707-35167707 on Build GRCh38
rs763460377 Chr.10: 35167734-35167734 on Build GRCh38
rs369808368 Chr.10: 35167750-35167750 on Build GRCh38
rs1033339063 Chr.10: 35167635-35167635 on Build GRCh38
rs779955748 Chr.10: 35167811-35167811 on Build GRCh38
rs778389369 Chr.10: 35167561-35167561 on Build GRCh38
rs888745613 Chr.10: 35167436-35167436 on Build GRCh38
rs548145963 Chr.10: 35167544-35167544 on Build GRCh38
rs745858589 Chr.10: 35167684-35167684 on Build GRCh38
rs909893357 Chr.10: 35167512-35167512 on Build GRCh38
rs765418452 Chr.10: 35167714-35167714 on Build GRCh38
rs370766716 Chr.10: 35167689-35167689 on Build GRCh38
rs41313822 Chr.10: 35167600-35167600 on Build GRCh38
rs955973107 Chr.10: 35167638-35167638 on Build GRCh38
rs774375230 Chr.10: 35167775-35167777 on Build GRCh38
rs768580073 Chr.10: 35167677-35167678 on Build GRCh38
rs768792377 Chr.10: 35167698-35167698 on Build GRCh38
rs949420016 Chr.10: 35167654-35167654 on Build GRCh38
rs111419679 Chr.10: 35167431-35167431 on Build GRCh38
rs771792531 Chr.10: 35167665-35167665 on Build GRCh38
rs528220450 Chr.10: 35167515-35167515 on Build GRCh38

View Full Product Details