Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 35608133-35608629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJD4
Entrez Gene ID
219770
Gene Name
gap junction protein delta 4
Gene Aliases
CX40.1
UniGene
Hs.638922
Gene Chromosome Location
Chr.10: 35605410-35608935 on Build GRCh38
Transcripts
NM_153368.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4144663 c.805C>A p.R269S Chr.10: 35608318-35608318 on Build GRCh38
684418 c.831C>G p.S277R Chr.10: 35608344-35608344 on Build GRCh38
40904 c.845C>T p.S282F Chr.10: 35608358-35608358 on Build GRCh38
5786389 c.771G>A p.R257R Chr.10: 35608284-35608284 on Build GRCh38
6671295 c.662G>A p.R221Q Chr.10: 35608175-35608175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200373511 Chr.10: 35608198-35608198 on Build GRCh38
rs991560017 Chr.10: 35608201-35608201 on Build GRCh38
rs761174554 Chr.10: 35608184-35608184 on Build GRCh38
rs946776113 Chr.10: 35608293-35608310 on Build GRCh38
rs772327530 Chr.10: 35608467-35608467 on Build GRCh38
rs768579100 Chr.10: 35608306-35608306 on Build GRCh38
rs956674591 Chr.10: 35608447-35608447 on Build GRCh38
rs867452720 Chr.10: 35608227-35608227 on Build GRCh38
rs183406069 Chr.10: 35608585-35608585 on Build GRCh38
rs748547746 Chr.10: 35608212-35608212 on Build GRCh38
rs537661394 Chr.10: 35608589-35608589 on Build GRCh38
rs529933444 Chr.10: 35608190-35608190 on Build GRCh38
rs369179387 Chr.10: 35608283-35608283 on Build GRCh38
rs777498735 Chr.10: 35608445-35608445 on Build GRCh38
rs199590260 Chr.10: 35608206-35608206 on Build GRCh38
rs111304561 Chr.10: 35608232-35608232 on Build GRCh38
rs770618498 Chr.10: 35608154-35608154 on Build GRCh38
rs904585171 Chr.10: 35608571-35608571 on Build GRCh38
rs780621371 Chr.10: 35608181-35608181 on Build GRCh38
rs775266410 Chr.10: 35608239-35608239 on Build GRCh38
rs200889483 Chr.10: 35608372-35608372 on Build GRCh38
rs34772501 Chr.10: 35608507-35608508 on Build GRCh38
rs548344551 Chr.10: 35608568-35608568 on Build GRCh38
rs560919389 Chr.10: 35608460-35608460 on Build GRCh38
rs776358447 Chr.10: 35608247-35608247 on Build GRCh38
rs767586928 Chr.10: 35608159-35608159 on Build GRCh38
rs1013046130 Chr.10: 35608387-35608387 on Build GRCh38
rs746521519 Chr.10: 35608305-35608305 on Build GRCh38
rs778713646 Chr.10: 35608225-35608225 on Build GRCh38
rs760523540 Chr.10: 35608171-35608171 on Build GRCh38
rs956833628 Chr.10: 35608237-35608237 on Build GRCh38
rs774403435 Chr.10: 35608155-35608155 on Build GRCh38
rs369250060 Chr.10: 35608555-35608555 on Build GRCh38
rs867282957 Chr.10: 35608248-35608248 on Build GRCh38
rs565706094 Chr.10: 35608557-35608557 on Build GRCh38
rs372982907 Chr.10: 35608392-35608392 on Build GRCh38
rs555177601 Chr.10: 35608234-35608234 on Build GRCh38
rs754458032 Chr.10: 35608525-35608525 on Build GRCh38
rs747267315 Chr.10: 35608184-35608184 on Build GRCh38
rs748991317 Chr.10: 35608189-35608189 on Build GRCh38
rs747005535 Chr.10: 35608243-35608243 on Build GRCh38
rs1031354856 Chr.10: 35608417-35608417 on Build GRCh38
rs754184758 Chr.10: 35608390-35608390 on Build GRCh38
rs766949119 Chr.10: 35608201-35608201 on Build GRCh38
rs996180458 Chr.10: 35608386-35608386 on Build GRCh38
rs77962491 Chr.10: 35608552-35608552 on Build GRCh38
rs376870967 Chr.10: 35608487-35608487 on Build GRCh38
rs759628221 Chr.10: 35608292-35608292 on Build GRCh38
rs745564408 Chr.10: 35608295-35608295 on Build GRCh38
rs552221917 Chr.10: 35608546-35608546 on Build GRCh38
rs537335234 Chr.10: 35608262-35608262 on Build GRCh38
rs147622465 Chr.10: 35608416-35608416 on Build GRCh38
rs781090050 Chr.10: 35608423-35608423 on Build GRCh38
rs769756898 Chr.10: 35608444-35608444 on Build GRCh38
rs764218355 Chr.10: 35608174-35608174 on Build GRCh38
rs377442192 Chr.10: 35608407-35608407 on Build GRCh38
rs535200408 Chr.10: 35608223-35608223 on Build GRCh38
rs765343383 Chr.10: 35608293-35608303 on Build GRCh38
rs764556009 Chr.10: 35608353-35608353 on Build GRCh38
rs538629092 Chr.10: 35608199-35608199 on Build GRCh38
rs773951943 Chr.10: 35608194-35608194 on Build GRCh38
rs141883026 Chr.10: 35608376-35608376 on Build GRCh38
rs952550869 Chr.10: 35608480-35608480 on Build GRCh38
rs867654897 Chr.10: 35608153-35608153 on Build GRCh38
rs528178597 Chr.10: 35608559-35608559 on Build GRCh38
rs751624521 Chr.10: 35608261-35608261 on Build GRCh38
rs768153609 Chr.10: 35608270-35608270 on Build GRCh38
rs765566462 Chr.10: 35608595-35608595 on Build GRCh38
rs903929444 Chr.10: 35608393-35608393 on Build GRCh38
rs752230784 Chr.10: 35608532-35608532 on Build GRCh38
rs1013427310 Chr.10: 35608446-35608446 on Build GRCh38
rs780644504 Chr.10: 35608531-35608531 on Build GRCh38
rs192362407 Chr.10: 35608277-35608277 on Build GRCh38
rs759132395 Chr.10: 35608196-35608196 on Build GRCh38
rs772453319 Chr.10: 35608580-35608580 on Build GRCh38
rs545809371 Chr.10: 35608366-35608366 on Build GRCh38
rs370286081 Chr.10: 35608459-35608459 on Build GRCh38
rs759509399 Chr.10: 35608156-35608156 on Build GRCh38
rs768135317 Chr.10: 35608355-35608355 on Build GRCh38
rs752785611 Chr.10: 35608514-35608514 on Build GRCh38
rs1057504440 Chr.10: 35608528-35608528 on Build GRCh38
rs779584150 Chr.10: 35608400-35608400 on Build GRCh38
rs779002375 Chr.10: 35608177-35608177 on Build GRCh38
rs142184341 Chr.10: 35608418-35608418 on Build GRCh38
rs763198460 Chr.10: 35608327-35608327 on Build GRCh38
rs760849098 Chr.10: 35608586-35608586 on Build GRCh38
rs778695135 Chr.10: 35608290-35608290 on Build GRCh38
rs777279417 Chr.10: 35608289-35608289 on Build GRCh38
rs764593459 Chr.10: 35608388-35608388 on Build GRCh38
rs752161045 Chr.10: 35608209-35608209 on Build GRCh38
rs749052188 Chr.10: 35608453-35608453 on Build GRCh38
rs531975885 Chr.10: 35608534-35608534 on Build GRCh38
rs112658635 Chr.10: 35608217-35608217 on Build GRCh38
rs374623236 Chr.10: 35608472-35608472 on Build GRCh38
rs777579109 Chr.10: 35608535-35608535 on Build GRCh38
rs757304072 Chr.10: 35608175-35608175 on Build GRCh38
rs777068787 Chr.10: 35608591-35608591 on Build GRCh38
rs773395724 Chr.10: 35608324-35608324 on Build GRCh38
rs371212610 Chr.10: 35608494-35608494 on Build GRCh38
rs529949064 Chr.10: 35608493-35608493 on Build GRCh38
rs759541237 Chr.10: 35608354-35608354 on Build GRCh38
rs1031235592 Chr.10: 35608404-35608404 on Build GRCh38
rs766524465 Chr.10: 35608260-35608260 on Build GRCh38
rs908936343 Chr.10: 35608475-35608475 on Build GRCh38
rs773485636 Chr.10: 35608250-35608250 on Build GRCh38
rs75803523 Chr.10: 35608197-35608197 on Build GRCh38
rs761980497 Chr.10: 35608202-35608202 on Build GRCh38
rs547212582 Chr.10: 35608163-35608163 on Build GRCh38
rs113486133 Chr.10: 35608564-35608564 on Build GRCh38
rs1037032678 Chr.10: 35608311-35608311 on Build GRCh38
rs968716512 Chr.10: 35608414-35608414 on Build GRCh38
rs866560750 Chr.10: 35608574-35608574 on Build GRCh38
rs765372882 Chr.10: 35608249-35608249 on Build GRCh38
rs906012374 Chr.10: 35608161-35608161 on Build GRCh38
rs563606358 Chr.10: 35608524-35608524 on Build GRCh38
rs138751121 Chr.10: 35608310-35608310 on Build GRCh38
rs770741306 Chr.10: 35608191-35608191 on Build GRCh38
rs1049692373 Chr.10: 35608329-35608329 on Build GRCh38
rs186885885 Chr.10: 35608256-35608256 on Build GRCh38
rs139809902 Chr.10: 35608350-35608350 on Build GRCh38
rs765464798 Chr.10: 35608204-35608204 on Build GRCh38
rs774433895 Chr.10: 35608352-35608352 on Build GRCh38
rs866262508 Chr.10: 35608375-35608375 on Build GRCh38
rs903879101 Chr.10: 35608567-35608567 on Build GRCh38
rs768505889 Chr.10: 35608246-35608246 on Build GRCh38
rs897994887 Chr.10: 35608164-35608164 on Build GRCh38
rs201736651 Chr.10: 35608178-35608178 on Build GRCh38
rs766047746 Chr.10: 35608522-35608522 on Build GRCh38
rs755271116 Chr.10: 35608211-35608211 on Build GRCh38
rs763507520 Chr.10: 35608296-35608296 on Build GRCh38
rs980465904 Chr.10: 35608274-35608274 on Build GRCh38
rs80247967 Chr.10: 35608597-35608597 on Build GRCh38
rs755990278 Chr.10: 35608533-35608533 on Build GRCh38
rs750835638 Chr.10: 35608601-35608601 on Build GRCh38
rs779575513 Chr.10: 35608300-35608300 on Build GRCh38
rs941665704 Chr.10: 35608308-35608308 on Build GRCh38
rs753467235 Chr.10: 35608593-35608593 on Build GRCh38
rs980395733 Chr.10: 35608471-35608471 on Build GRCh38
rs769127011 Chr.10: 35608187-35608187 on Build GRCh38
rs151211621 Chr.10: 35608180-35608180 on Build GRCh38

View Full Product Details