Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 43138344-43138851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSGALNACT2
Entrez Gene ID
55454
Gene Name
chondroitin sulfate N-acetylgalactosaminyltransferase 2
Gene Aliases
CHGN2, ChGn-2, GALNACT-2, GALNACT2, PRO0082, beta4GalNAcT
UniGene
Hs.657569
Gene Chromosome Location
Chr.10: 43138445-43185308 on Build GRCh38
Transcripts
NM_018590.4 NR_135077.1 XM_011539938.1 XM_017016388.1 XM_017016389.1 XM_017016390.1 XM_017016391.1

SNP Detail

SNP ID Chromosome Location
rs201533869 Chr.10: 43138680-43138680 on Build GRCh38
rs991085688 Chr.10: 43138725-43138725 on Build GRCh38
rs558331636 Chr.10: 43138464-43138464 on Build GRCh38
rs775386125 Chr.10: 43138408-43138408 on Build GRCh38
rs975613453 Chr.10: 43138557-43138557 on Build GRCh38
rs77538976 Chr.10: 43138822-43138822 on Build GRCh38
rs967613310 Chr.10: 43138534-43138534 on Build GRCh38
rs374518707 Chr.10: 43138450-43138450 on Build GRCh38
rs529142327 Chr.10: 43138499-43138499 on Build GRCh38
rs907607908 Chr.10: 43138623-43138623 on Build GRCh38
rs1020924626 Chr.10: 43138371-43138371 on Build GRCh38
rs559541784 Chr.10: 43138624-43138624 on Build GRCh38
rs536216904 Chr.10: 43138467-43138468 on Build GRCh38
rs762778073 Chr.10: 43138566-43138566 on Build GRCh38
rs1014670054 Chr.10: 43138699-43138699 on Build GRCh38
rs936375470 Chr.10: 43138447-43138447 on Build GRCh38
rs182112701 Chr.10: 43138816-43138816 on Build GRCh38
rs1038685397 Chr.10: 43138449-43138449 on Build GRCh38
rs979008534 Chr.10: 43138413-43138413 on Build GRCh38
rs918125770 Chr.10: 43138733-43138733 on Build GRCh38
rs145045898 Chr.10: 43138468-43138469 on Build GRCh38
rs1020771690 Chr.10: 43138525-43138525 on Build GRCh38
rs895842501 Chr.10: 43138697-43138697 on Build GRCh38
rs1014371710 Chr.10: 43138517-43138517 on Build GRCh38
rs540311919 Chr.10: 43138685-43138685 on Build GRCh38
rs549251465 Chr.10: 43138793-43138793 on Build GRCh38
rs966742682 Chr.10: 43138375-43138375 on Build GRCh38
rs575974998 Chr.10: 43138374-43138374 on Build GRCh38
rs148684525 Chr.10: 43138492-43138492 on Build GRCh38
rs970978403 Chr.10: 43138775-43138775 on Build GRCh38
rs530930650 Chr.10: 43138789-43138789 on Build GRCh38
rs964266780 Chr.10: 43138556-43138556 on Build GRCh38
rs764132167 Chr.10: 43138660-43138660 on Build GRCh38
rs1037655344 Chr.10: 43138470-43138470 on Build GRCh38
rs978894352 Chr.10: 43138539-43138539 on Build GRCh38
rs940386894 Chr.10: 43138640-43138640 on Build GRCh38
rs1020403779 Chr.10: 43138714-43138714 on Build GRCh38
rs1032498976 Chr.10: 43138379-43138379 on Build GRCh38
rs932961889 Chr.10: 43138603-43138603 on Build GRCh38
rs899087929 Chr.10: 43138480-43138480 on Build GRCh38
rs926207483 Chr.10: 43138415-43138415 on Build GRCh38
rs1034367423 Chr.10: 43138507-43138507 on Build GRCh38
rs1051561154 Chr.10: 43138607-43138607 on Build GRCh38
rs534751278 Chr.10: 43138817-43138817 on Build GRCh38
rs1005119352 Chr.10: 43138468-43138468 on Build GRCh38
rs543180005 Chr.10: 43138412-43138412 on Build GRCh38
rs371679632 Chr.10: 43138748-43138748 on Build GRCh38
rs1051937809 Chr.10: 43138467-43138467 on Build GRCh38
rs558270315 Chr.10: 43138648-43138648 on Build GRCh38
rs960283234 Chr.10: 43138508-43138508 on Build GRCh38
rs926090510 Chr.10: 43138548-43138548 on Build GRCh38
rs1055708579 Chr.10: 43138693-43138693 on Build GRCh38
rs921562532 Chr.10: 43138465-43138465 on Build GRCh38
rs569980076 Chr.10: 43138734-43138734 on Build GRCh38
rs533287040 Chr.10: 43138653-43138653 on Build GRCh38
rs538462132 Chr.10: 43138592-43138592 on Build GRCh38
rs1000929637 Chr.10: 43138377-43138377 on Build GRCh38
rs972366262 Chr.10: 43138787-43138787 on Build GRCh38
rs867215627 Chr.10: 43138612-43138612 on Build GRCh38
rs921629461 Chr.10: 43138582-43138582 on Build GRCh38
rs903275532 Chr.10: 43138722-43138722 on Build GRCh38

View Full Product Details