Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 43175814-43176319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSGALNACT2
Entrez Gene ID
55454
Gene Name
chondroitin sulfate N-acetylgalactosaminyltransferase 2
Gene Aliases
CHGN2, ChGn-2, GALNACT-2, GALNACT2, PRO0082, beta4GalNAcT
UniGene
Hs.657569
Gene Chromosome Location
Chr.10: 43138445-43185308 on Build GRCh38
Transcripts
NM_001319654.1 NM_001319656.1 NM_018590.4 NR_135077.1 XM_017016388.1 XM_017016389.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1347835 c.1316G>A p.R439H Chr.10: 43176012-43176012 on Build GRCh38
5574534 c.1297G>T p.G433* Chr.10: 43175993-43175993 on Build GRCh38
5027212 c.1271C>A p.S424Y Chr.10: 43175967-43175967 on Build GRCh38
5698160 c.1282C>T p.R428* Chr.10: 43175978-43175978 on Build GRCh38
5913017 c.1315C>T p.R439C Chr.10: 43176011-43176011 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs142222902 Chr.10: 43175887-43175887 on Build GRCh38
rs763444153 Chr.10: 43176039-43176039 on Build GRCh38
rs202069979 Chr.10: 43175911-43175911 on Build GRCh38
rs747899426 Chr.10: 43175932-43175932 on Build GRCh38
rs913302009 Chr.10: 43176243-43176243 on Build GRCh38
rs959897237 Chr.10: 43176225-43176225 on Build GRCh38
rs547211052 Chr.10: 43176283-43176283 on Build GRCh38
rs901347970 Chr.10: 43176087-43176087 on Build GRCh38
rs761828809 Chr.10: 43176020-43176020 on Build GRCh38
rs745869057 Chr.10: 43176077-43176077 on Build GRCh38
rs549390508 Chr.10: 43176164-43176164 on Build GRCh38
rs80035255 Chr.10: 43175945-43175945 on Build GRCh38
rs753852596 Chr.10: 43175907-43175907 on Build GRCh38
rs775146962 Chr.10: 43175978-43175978 on Build GRCh38
rs777250633 Chr.10: 43176058-43176058 on Build GRCh38
rs1004535934 Chr.10: 43176133-43176133 on Build GRCh38
rs562729829 Chr.10: 43176093-43176093 on Build GRCh38
rs768969174 Chr.10: 43176144-43176144 on Build GRCh38
rs373735593 Chr.10: 43175934-43175935 on Build GRCh38
rs771928849 Chr.10: 43175970-43175970 on Build GRCh38
rs768741535 Chr.10: 43176010-43176010 on Build GRCh38
rs557905301 Chr.10: 43176048-43176048 on Build GRCh38
rs886219112 Chr.10: 43176121-43176121 on Build GRCh38
rs753411642 Chr.10: 43176062-43176062 on Build GRCh38
rs543364865 Chr.10: 43176158-43176158 on Build GRCh38
rs371897213 Chr.10: 43176201-43176201 on Build GRCh38
rs398013310 Chr.10: 43175943-43175943 on Build GRCh38
rs967888895 Chr.10: 43176244-43176244 on Build GRCh38
rs545418745 Chr.10: 43175932-43175932 on Build GRCh38
rs534822228 Chr.10: 43175931-43175932 on Build GRCh38
rs377210956 Chr.10: 43176029-43176029 on Build GRCh38
rs763502942 Chr.10: 43175931-43175933 on Build GRCh38
rs370983026 Chr.10: 43175935-43175935 on Build GRCh38
rs761018259 Chr.10: 43176004-43176004 on Build GRCh38
rs144838728 Chr.10: 43175956-43175956 on Build GRCh38
rs369366976 Chr.10: 43175890-43175890 on Build GRCh38
rs778999753 Chr.10: 43176065-43176065 on Build GRCh38
rs748714844 Chr.10: 43175943-43175943 on Build GRCh38
rs61579799 Chr.10: 43175931-43175931 on Build GRCh38
rs541263227 Chr.10: 43176182-43176182 on Build GRCh38
rs978733725 Chr.10: 43176293-43176293 on Build GRCh38
rs569123492 Chr.10: 43176246-43176246 on Build GRCh38
rs750859874 Chr.10: 43176037-43176037 on Build GRCh38
rs758990532 Chr.10: 43176109-43176109 on Build GRCh38
rs750804921 Chr.10: 43175923-43175923 on Build GRCh38
rs756771391 Chr.10: 43176063-43176063 on Build GRCh38
rs866113646 Chr.10: 43175953-43175954 on Build GRCh38
rs535659827 Chr.10: 43175893-43175893 on Build GRCh38
rs772125021 Chr.10: 43176083-43176083 on Build GRCh38
rs1043134599 Chr.10: 43175966-43175966 on Build GRCh38
rs776815858 Chr.10: 43176012-43176012 on Build GRCh38
rs758699810 Chr.10: 43175931-43175931 on Build GRCh38
rs200933777 Chr.10: 43176055-43176055 on Build GRCh38
rs995234250 Chr.10: 43176202-43176202 on Build GRCh38
rs976134311 Chr.10: 43176011-43176011 on Build GRCh38
rs398075109 Chr.10: 43175942-43175942 on Build GRCh38
rs549914724 Chr.10: 43175886-43175886 on Build GRCh38
rs116694433 Chr.10: 43175951-43175951 on Build GRCh38
rs543115498 Chr.10: 43175932-43175933 on Build GRCh38
rs1014235770 Chr.10: 43176178-43176178 on Build GRCh38
rs182228813 Chr.10: 43175897-43175897 on Build GRCh38
rs375579259 Chr.10: 43176092-43176092 on Build GRCh38
rs943950127 Chr.10: 43175962-43175962 on Build GRCh38
rs780932234 Chr.10: 43175930-43175931 on Build GRCh38
rs758398160 Chr.10: 43175914-43175918 on Build GRCh38
rs375074382 Chr.10: 43175921-43175921 on Build GRCh38
rs777390040 Chr.10: 43175939-43175939 on Build GRCh38
rs755369944 Chr.10: 43175972-43175972 on Build GRCh38
rs993104240 Chr.10: 43176236-43176236 on Build GRCh38
rs938157624 Chr.10: 43176100-43176100 on Build GRCh38
rs200528769 Chr.10: 43175934-43175934 on Build GRCh38
rs751847006 Chr.10: 43176053-43176053 on Build GRCh38
rs555463146 Chr.10: 43176161-43176161 on Build GRCh38
rs749517811 Chr.10: 43176022-43176022 on Build GRCh38
rs963237525 Chr.10: 43176199-43176199 on Build GRCh38
rs1034106787 Chr.10: 43176224-43176224 on Build GRCh38

View Full Product Details