Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68910201-68910724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX50
Entrez Gene ID
79009
Gene Name
DEAD-box helicase 50
Gene Aliases
GU2, GUB, RH-II/GuB, mcdrh
UniGene
Hs.522984
Gene Chromosome Location
Chr.10: 68901278-68956312 on Build GRCh38
Transcripts
NM_024045.1 XM_005270148.2 XM_011540144.2 XM_017016626.1 XM_017016627.1 XM_017016628.1 XM_017016629.1 XM_017016630.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3985704 c.428C>A p.P143H Chr.10: 68910350-68910350 on Build GRCh38
4993796 c.460+4T>A p.? Chr.10: 68910386-68910386 on Build GRCh38
6334015 c.395G>T p.R132L Chr.10: 68910317-68910317 on Build GRCh38
2159659 c.449A>C p.K150T Chr.10: 68910371-68910371 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs568970686 Chr.10: 68910533-68910534 on Build GRCh38
rs997243365 Chr.10: 68910505-68910505 on Build GRCh38
rs374841303 Chr.10: 68910317-68910317 on Build GRCh38
rs373943363 Chr.10: 68910288-68910288 on Build GRCh38
rs185236038 Chr.10: 68910273-68910273 on Build GRCh38
rs10823269 Chr.10: 68910603-68910603 on Build GRCh38
rs904103290 Chr.10: 68910694-68910694 on Build GRCh38
rs779511938 Chr.10: 68910275-68910275 on Build GRCh38
rs919119547 Chr.10: 68910651-68910651 on Build GRCh38
rs540263131 Chr.10: 68910640-68910640 on Build GRCh38
rs969014871 Chr.10: 68910512-68910512 on Build GRCh38
rs969374303 Chr.10: 68910633-68910633 on Build GRCh38
rs747857927 Chr.10: 68910592-68910592 on Build GRCh38
rs576801884 Chr.10: 68910246-68910246 on Build GRCh38
rs763856954 Chr.10: 68910344-68910344 on Build GRCh38
rs113782027 Chr.10: 68910474-68910490 on Build GRCh38
rs1007655552 Chr.10: 68910551-68910551 on Build GRCh38
rs546668621 Chr.10: 68910520-68910520 on Build GRCh38
rs758818246 Chr.10: 68910285-68910285 on Build GRCh38
rs1036184957 Chr.10: 68910459-68910459 on Build GRCh38
rs190884371 Chr.10: 68910436-68910436 on Build GRCh38
rs751261926 Chr.10: 68910396-68910407 on Build GRCh38
rs980012003 Chr.10: 68910330-68910330 on Build GRCh38
rs780421609 Chr.10: 68910401-68910401 on Build GRCh38
rs909123024 Chr.10: 68910454-68910454 on Build GRCh38
rs373829270 Chr.10: 68910386-68910386 on Build GRCh38
rs753340061 Chr.10: 68910263-68910263 on Build GRCh38
rs761682138 Chr.10: 68910408-68910410 on Build GRCh38
rs539231765 Chr.10: 68910290-68910290 on Build GRCh38
rs778703795 Chr.10: 68910445-68910445 on Build GRCh38
rs971851660 Chr.10: 68910642-68910642 on Build GRCh38
rs560042723 Chr.10: 68910545-68910545 on Build GRCh38
rs781544917 Chr.10: 68910429-68910429 on Build GRCh38
rs370670333 Chr.10: 68910352-68910352 on Build GRCh38
rs543949939 Chr.10: 68910447-68910447 on Build GRCh38
rs41305008 Chr.10: 68910611-68910611 on Build GRCh38
rs148713144 Chr.10: 68910316-68910316 on Build GRCh38
rs532950128 Chr.10: 68910508-68910508 on Build GRCh38
rs1018488350 Chr.10: 68910560-68910560 on Build GRCh38
rs371419335 Chr.10: 68910477-68910493 on Build GRCh38
rs943016255 Chr.10: 68910687-68910687 on Build GRCh38
rs764711042 Chr.10: 68910363-68910363 on Build GRCh38
rs933167170 Chr.10: 68910662-68910662 on Build GRCh38
rs570820207 Chr.10: 68910490-68910491 on Build GRCh38
rs573102159 Chr.10: 68910336-68910336 on Build GRCh38
rs748305733 Chr.10: 68910431-68910431 on Build GRCh38
rs769760129 Chr.10: 68910324-68910324 on Build GRCh38
rs201568083 Chr.10: 68910397-68910397 on Build GRCh38
rs777933925 Chr.10: 68910434-68910434 on Build GRCh38
rs781253118 Chr.10: 68910318-68910318 on Build GRCh38
rs888758660 Chr.10: 68910542-68910542 on Build GRCh38
rs111554656 Chr.10: 68910690-68910690 on Build GRCh38
rs774489293 Chr.10: 68910333-68910333 on Build GRCh38
rs142163966 Chr.10: 68910563-68910563 on Build GRCh38
rs764406176 Chr.10: 68910276-68910278 on Build GRCh38
rs752959432 Chr.10: 68910667-68910667 on Build GRCh38
rs748303036 Chr.10: 68910321-68910321 on Build GRCh38
rs746960582 Chr.10: 68910637-68910637 on Build GRCh38
rs372283056 Chr.10: 68910433-68910433 on Build GRCh38
rs1044387137 Chr.10: 68910341-68910341 on Build GRCh38
rs548727437 Chr.10: 68910600-68910600 on Build GRCh38
rs370499254 Chr.10: 68910284-68910284 on Build GRCh38
rs564204694 Chr.10: 68910458-68910458 on Build GRCh38
rs376642364 Chr.10: 68910309-68910309 on Build GRCh38
rs1010473733 Chr.10: 68910420-68910420 on Build GRCh38
rs542372355 Chr.10: 68910569-68910569 on Build GRCh38
rs897575484 Chr.10: 68910465-68910465 on Build GRCh38
rs111659638 Chr.10: 68910677-68910677 on Build GRCh38
rs755269536 Chr.10: 68910407-68910407 on Build GRCh38
rs546595995 Chr.10: 68910679-68910679 on Build GRCh38
rs1044333533 Chr.10: 68910655-68910655 on Build GRCh38
rs113171200 Chr.10: 68910555-68910555 on Build GRCh38
rs1001027621 Chr.10: 68910564-68910564 on Build GRCh38
rs1043150523 Chr.10: 68910692-68910692 on Build GRCh38
rs888865527 Chr.10: 68910680-68910680 on Build GRCh38
rs1025079877 Chr.10: 68910614-68910614 on Build GRCh38
rs759491146 Chr.10: 68910335-68910335 on Build GRCh38
rs997130019 Chr.10: 68910231-68910231 on Build GRCh38
rs912388361 Chr.10: 68910591-68910591 on Build GRCh38
rs780335896 Chr.10: 68910295-68910295 on Build GRCh38
rs557615978 Chr.10: 68910323-68910323 on Build GRCh38
rs368086619 Chr.10: 68910315-68910315 on Build GRCh38
rs981186119 Chr.10: 68910635-68910635 on Build GRCh38
rs1029991607 Chr.10: 68910241-68910241 on Build GRCh38
rs753397169 Chr.10: 68910348-68910348 on Build GRCh38
rs79596082 Chr.10: 68910402-68910402 on Build GRCh38
rs769996550 Chr.10: 68910476-68910476 on Build GRCh38
rs200538672 Chr.10: 68910245-68910245 on Build GRCh38

View Full Product Details