Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68956248-68956754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX21
Entrez Gene ID
9188
Gene Name
DEAD-box helicase 21
Gene Aliases
GUA, GURDB, RH-II/GU, RH-II/GuA
UniGene
Hs.223141
Gene Chromosome Location
Chr.10: 68956123-68985069 on Build GRCh38
Transcripts
NM_001256910.1 NM_004728.3 XM_011540336.1 XM_017016910.1 XM_017016626.1

Gene Symbol
DDX50
Entrez Gene ID
79009
Gene Name
DEAD-box helicase 50
Gene Aliases
GU2, GUB, RH-II/GuB, mcdrh
UniGene
Hs.522984
Gene Chromosome Location
Chr.10: 68901278-68956312 on Build GRCh38
Transcripts
NM_001256910.1 NM_004728.3 XM_011540336.1 XM_017016910.1 XM_017016626.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6040736 c.76C>G p.Q26E Chr.10: 68956301-68956301 on Build GRCh38
919837 c.67C>G p.L23V Chr.10: 68956292-68956292 on Build GRCh38
7152962 c.71G>A p.R24Q Chr.10: 68956296-68956296 on Build GRCh38
4015336 c.57A>G p.K19K Chr.10: 68956282-68956282 on Build GRCh38
7264436 c.51_52insA p.G20fs*20 Chr.10: 68956276-68956277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1026633655 Chr.10: 68956681-68956681 on Build GRCh38
rs1019406506 Chr.10: 68956384-68956384 on Build GRCh38
rs903096327 Chr.10: 68956335-68956335 on Build GRCh38
rs892741532 Chr.10: 68956587-68956587 on Build GRCh38
rs1025044131 Chr.10: 68956413-68956413 on Build GRCh38
rs17556220 Chr.10: 68956305-68956305 on Build GRCh38
rs368924310 Chr.10: 68956484-68956484 on Build GRCh38
rs919192502 Chr.10: 68956473-68956473 on Build GRCh38
rs1045393580 Chr.10: 68956470-68956470 on Build GRCh38
rs774156390 Chr.10: 68956518-68956518 on Build GRCh38
rs544582582 Chr.10: 68956594-68956594 on Build GRCh38
rs748041969 Chr.10: 68956527-68956527 on Build GRCh38
rs539932340 Chr.10: 68956433-68956433 on Build GRCh38
rs369746986 Chr.10: 68956333-68956333 on Build GRCh38
rs1007948041 Chr.10: 68956560-68956560 on Build GRCh38
rs1007163982 Chr.10: 68956300-68956300 on Build GRCh38
rs987259990 Chr.10: 68956708-68956708 on Build GRCh38
rs532406881 Chr.10: 68956380-68956380 on Build GRCh38
rs924370835 Chr.10: 68956724-68956724 on Build GRCh38
rs369038483 Chr.10: 68956356-68956356 on Build GRCh38
rs759184404 Chr.10: 68956695-68956695 on Build GRCh38
rs531195964 Chr.10: 68956550-68956550 on Build GRCh38
rs916968675 Chr.10: 68956510-68956510 on Build GRCh38
rs780836636 Chr.10: 68956319-68956319 on Build GRCh38
rs1055485609 Chr.10: 68956503-68956503 on Build GRCh38
rs547099416 Chr.10: 68956717-68956717 on Build GRCh38
rs139727835 Chr.10: 68956539-68956539 on Build GRCh38
rs905275652 Chr.10: 68956630-68956630 on Build GRCh38
rs1026479101 Chr.10: 68956626-68956626 on Build GRCh38
rs115031368 Chr.10: 68956496-68956496 on Build GRCh38
rs972354683 Chr.10: 68956465-68956465 on Build GRCh38
rs1042701570 Chr.10: 68956537-68956537 on Build GRCh38
rs577762748 Chr.10: 68956354-68956354 on Build GRCh38
rs1002248631 Chr.10: 68956642-68956642 on Build GRCh38
rs936966260 Chr.10: 68956501-68956501 on Build GRCh38
rs963678814 Chr.10: 68956658-68956658 on Build GRCh38
rs111678632 Chr.10: 68956477-68956477 on Build GRCh38
rs553487021 Chr.10: 68956457-68956457 on Build GRCh38
rs959662251 Chr.10: 68956664-68956664 on Build GRCh38
rs754763018 Chr.10: 68956306-68956306 on Build GRCh38
rs969488470 Chr.10: 68956422-68956422 on Build GRCh38
rs373860935 Chr.10: 68956334-68956334 on Build GRCh38
rs542516610 Chr.10: 68956534-68956534 on Build GRCh38
rs949833106 Chr.10: 68956513-68956513 on Build GRCh38
rs766279960 Chr.10: 68956273-68956273 on Build GRCh38
rs746297181 Chr.10: 68956359-68956359 on Build GRCh38
rs755613026 Chr.10: 68956337-68956337 on Build GRCh38
rs966134351 Chr.10: 68956400-68956400 on Build GRCh38
rs760247173 Chr.10: 68956574-68956574 on Build GRCh38
rs770400499 Chr.10: 68956636-68956636 on Build GRCh38
rs963411232 Chr.10: 68956459-68956459 on Build GRCh38
rs112803606 Chr.10: 68956478-68956478 on Build GRCh38
rs910291506 Chr.10: 68956526-68956526 on Build GRCh38
rs993603048 Chr.10: 68956617-68956617 on Build GRCh38
rs895571815 Chr.10: 68956292-68956292 on Build GRCh38
rs778784216 Chr.10: 68956553-68956553 on Build GRCh38
rs533606217 Chr.10: 68956711-68956711 on Build GRCh38
rs776200635 Chr.10: 68956649-68956649 on Build GRCh38
rs972107210 Chr.10: 68956665-68956665 on Build GRCh38
rs79554134 Chr.10: 68956270-68956270 on Build GRCh38
rs557549736 Chr.10: 68956648-68956648 on Build GRCh38
rs193010366 Chr.10: 68956583-68956583 on Build GRCh38
rs991550470 Chr.10: 68956670-68956670 on Build GRCh38
rs980759947 Chr.10: 68956450-68956450 on Build GRCh38
rs985481205 Chr.10: 68956451-68956451 on Build GRCh38
rs746080411 Chr.10: 68956360-68956360 on Build GRCh38
rs773122993 Chr.10: 68956565-68956565 on Build GRCh38
rs1053088220 Chr.10: 68956582-68956582 on Build GRCh38
rs777290418 Chr.10: 68956340-68956340 on Build GRCh38
rs1025181506 Chr.10: 68956605-68956605 on Build GRCh38
rs1054746362 Chr.10: 68956540-68956540 on Build GRCh38
rs5030896 Chr.10: 68956350-68956350 on Build GRCh38
rs972194781 Chr.10: 68956430-68956430 on Build GRCh38

View Full Product Details