Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68959668-68960202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX21
Entrez Gene ID
9188
Gene Name
DEAD-box helicase 21
Gene Aliases
GUA, GURDB, RH-II/GU, RH-II/GuA
UniGene
Hs.223141
Gene Chromosome Location
Chr.10: 68956123-68985069 on Build GRCh38
Transcripts
NM_001256910.1 NM_004728.3 XM_011540336.1 XM_017016910.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3867663 c.465T>C p.P155P Chr.10: 68960183-68960183 on Build GRCh38
5522481 c.157C>T p.P53S Chr.10: 68959875-68959875 on Build GRCh38
3981091 c.323C>A p.S108Y Chr.10: 68960041-68960041 on Build GRCh38
6641614 c.282T>C p.S94S Chr.10: 68960000-68960000 on Build GRCh38
5522482 c.156C>T p.F52F Chr.10: 68959874-68959874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35813389 Chr.10: 68959893-68959893 on Build GRCh38
rs780414967 Chr.10: 68959923-68959923 on Build GRCh38
rs965161089 Chr.10: 68959839-68959839 on Build GRCh38
rs374126084 Chr.10: 68960005-68960005 on Build GRCh38
rs753458616 Chr.10: 68959891-68959891 on Build GRCh38
rs147861232 Chr.10: 68960044-68960044 on Build GRCh38
rs779827842 Chr.10: 68959759-68959760 on Build GRCh38
rs754069395 Chr.10: 68959814-68959814 on Build GRCh38
rs141331906 Chr.10: 68960056-68960056 on Build GRCh38
rs766352017 Chr.10: 68959751-68959751 on Build GRCh38
rs1023196442 Chr.10: 68960025-68960025 on Build GRCh38
rs760908575 Chr.10: 68959791-68959791 on Build GRCh38
rs1057352187 Chr.10: 68960071-68960071 on Build GRCh38
rs570130822 Chr.10: 68960063-68960063 on Build GRCh38
rs201516552 Chr.10: 68959922-68959922 on Build GRCh38
rs776819535 Chr.10: 68959798-68959798 on Build GRCh38
rs768961518 Chr.10: 68959948-68959948 on Build GRCh38
rs776373179 Chr.10: 68960119-68960119 on Build GRCh38
rs777262760 Chr.10: 68959877-68959877 on Build GRCh38
rs767422709 Chr.10: 68959973-68959973 on Build GRCh38
rs773099188 Chr.10: 68960061-68960062 on Build GRCh38
rs868449260 Chr.10: 68959903-68959903 on Build GRCh38
rs773330252 Chr.10: 68960082-68960082 on Build GRCh38
rs987940502 Chr.10: 68960155-68960155 on Build GRCh38
rs370418699 Chr.10: 68959975-68959975 on Build GRCh38
rs946615430 Chr.10: 68959892-68959892 on Build GRCh38
rs772116635 Chr.10: 68960106-68960106 on Build GRCh38
rs989549982 Chr.10: 68960002-68960002 on Build GRCh38
rs143526452 Chr.10: 68960148-68960148 on Build GRCh38
rs780392199 Chr.10: 68960049-68960049 on Build GRCh38
rs779581058 Chr.10: 68959912-68959912 on Build GRCh38
rs988360681 Chr.10: 68959887-68959887 on Build GRCh38
rs1043613610 Chr.10: 68959902-68959902 on Build GRCh38
rs749994989 Chr.10: 68960167-68960167 on Build GRCh38
rs756818495 Chr.10: 68959907-68959907 on Build GRCh38
rs751953810 Chr.10: 68960178-68960178 on Build GRCh38
rs766968332 Chr.10: 68960174-68960174 on Build GRCh38
rs894561847 Chr.10: 68960130-68960130 on Build GRCh38
rs747472832 Chr.10: 68960055-68960055 on Build GRCh38
rs952824475 Chr.10: 68959812-68959812 on Build GRCh38
rs372610547 Chr.10: 68959920-68959920 on Build GRCh38
rs753378539 Chr.10: 68959998-68959998 on Build GRCh38
rs1004098608 Chr.10: 68959817-68959817 on Build GRCh38
rs146119299 Chr.10: 68960026-68960026 on Build GRCh38
rs759331255 Chr.10: 68959833-68959833 on Build GRCh38
rs760385541 Chr.10: 68959986-68959986 on Build GRCh38
rs367544427 Chr.10: 68960115-68960115 on Build GRCh38
rs756828616 Chr.10: 68960022-68960022 on Build GRCh38
rs373676336 Chr.10: 68960095-68960095 on Build GRCh38
rs1023647530 Chr.10: 68959774-68959774 on Build GRCh38
rs938816389 Chr.10: 68960057-68960057 on Build GRCh38
rs187776549 Chr.10: 68959795-68959795 on Build GRCh38
rs945047119 Chr.10: 68960006-68960006 on Build GRCh38
rs775855456 Chr.10: 68959775-68959775 on Build GRCh38
rs974385695 Chr.10: 68959743-68959743 on Build GRCh38
rs776712482 Chr.10: 68959950-68959950 on Build GRCh38
rs752315891 Chr.10: 68959852-68959852 on Build GRCh38
rs755841241 Chr.10: 68959872-68959872 on Build GRCh38
rs759421230 Chr.10: 68959967-68959967 on Build GRCh38
rs144879525 Chr.10: 68959821-68959821 on Build GRCh38
rs201461790 Chr.10: 68960172-68960172 on Build GRCh38
rs750986998 Chr.10: 68960034-68960034 on Build GRCh38
rs775280306 Chr.10: 68960108-68960108 on Build GRCh38
rs200379451 Chr.10: 68959995-68959995 on Build GRCh38
rs374486646 Chr.10: 68959932-68959932 on Build GRCh38
rs781489820 Chr.10: 68960074-68960074 on Build GRCh38
rs976578762 Chr.10: 68959865-68959865 on Build GRCh38
rs865876480 Chr.10: 68960050-68960050 on Build GRCh38
rs868098850 Chr.10: 68959945-68959945 on Build GRCh38
rs766369835 Chr.10: 68959811-68959811 on Build GRCh38
rs767261297 Chr.10: 68959849-68959849 on Build GRCh38
rs762141510 Chr.10: 68959796-68959796 on Build GRCh38
rs760571144 Chr.10: 68960064-68960072 on Build GRCh38
rs375850935 Chr.10: 68959962-68959962 on Build GRCh38
rs747312279 Chr.10: 68959931-68959931 on Build GRCh38
rs763734795 Chr.10: 68960116-68960116 on Build GRCh38
rs748329959 Chr.10: 68960076-68960076 on Build GRCh38
rs894589933 Chr.10: 68959716-68959716 on Build GRCh38
rs771729798 Chr.10: 68959850-68959852 on Build GRCh38
rs924879102 Chr.10: 68960033-68960033 on Build GRCh38
rs141991731 Chr.10: 68959965-68959965 on Build GRCh38
rs762991895 Chr.10: 68960100-68960100 on Build GRCh38
rs372263328 Chr.10: 68960166-68960166 on Build GRCh38
rs761766702 Chr.10: 68959802-68959802 on Build GRCh38

View Full Product Details