Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 69630161-69630662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf35
Entrez Gene ID
219738
Gene Name
chromosome 10 open reading frame 35
Gene Aliases
-
UniGene
Hs.522992
Gene Chromosome Location
Chr.10: 69630241-69633599 on Build GRCh38
Transcripts
NM_145306.2 XM_005269606.1 XM_005269608.3 XM_017015836.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7233323 c.1A>G p.M1V Chr.10: 69630637-69630637 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs967142977 Chr.10: 69630215-69630215 on Build GRCh38
rs115566752 Chr.10: 69630271-69630271 on Build GRCh38
rs958349127 Chr.10: 69630329-69630329 on Build GRCh38
rs889641806 Chr.10: 69630422-69630422 on Build GRCh38
rs897091582 Chr.10: 69630250-69630250 on Build GRCh38
rs901295169 Chr.10: 69630485-69630485 on Build GRCh38
rs1017427312 Chr.10: 69630262-69630262 on Build GRCh38
rs902488940 Chr.10: 69630368-69630368 on Build GRCh38
rs149318502 Chr.10: 69630284-69630284 on Build GRCh38
rs957165207 Chr.10: 69630232-69630232 on Build GRCh38
rs965369564 Chr.10: 69630286-69630286 on Build GRCh38
rs969760743 Chr.10: 69630380-69630380 on Build GRCh38
rs894010138 Chr.10: 69630533-69630533 on Build GRCh38
rs969561492 Chr.10: 69630552-69630552 on Build GRCh38
rs573622418 Chr.10: 69630465-69630465 on Build GRCh38
rs922946953 Chr.10: 69630274-69630274 on Build GRCh38
rs557432881 Chr.10: 69630476-69630476 on Build GRCh38
rs931251674 Chr.10: 69630412-69630412 on Build GRCh38
rs1048266193 Chr.10: 69630276-69630276 on Build GRCh38
rs916755709 Chr.10: 69630370-69630370 on Build GRCh38
rs928291826 Chr.10: 69630193-69630193 on Build GRCh38
rs879744890 Chr.10: 69630636-69630637 on Build GRCh38
rs1036967224 Chr.10: 69630231-69630231 on Build GRCh38
rs979859756 Chr.10: 69630382-69630382 on Build GRCh38
rs975994569 Chr.10: 69630458-69630458 on Build GRCh38
rs999506729 Chr.10: 69630297-69630297 on Build GRCh38
rs182427343 Chr.10: 69630599-69630599 on Build GRCh38
rs1013878298 Chr.10: 69630183-69630183 on Build GRCh38
rs542076531 Chr.10: 69630461-69630461 on Build GRCh38
rs879816390 Chr.10: 69630634-69630634 on Build GRCh38
rs989950802 Chr.10: 69630244-69630244 on Build GRCh38
rs1032395916 Chr.10: 69630404-69630404 on Build GRCh38
rs1041091787 Chr.10: 69630463-69630463 on Build GRCh38
rs972418810 Chr.10: 69630399-69630399 on Build GRCh38
rs1019545766 Chr.10: 69630285-69630285 on Build GRCh38
rs1048670727 Chr.10: 69630414-69630414 on Build GRCh38
rs964594374 Chr.10: 69630265-69630265 on Build GRCh38
rs552845903 Chr.10: 69630517-69630517 on Build GRCh38
rs949602284 Chr.10: 69630549-69630549 on Build GRCh38
rs909979021 Chr.10: 69630449-69630449 on Build GRCh38
rs575588831 Chr.10: 69630537-69630537 on Build GRCh38
rs916722500 Chr.10: 69630547-69630547 on Build GRCh38
rs1011163931 Chr.10: 69630544-69630544 on Build GRCh38
rs1046607196 Chr.10: 69630612-69630612 on Build GRCh38
rs964542132 Chr.10: 69630454-69630454 on Build GRCh38
rs1031659018 Chr.10: 69630307-69630307 on Build GRCh38
rs574621041 Chr.10: 69630405-69630405 on Build GRCh38
rs918268695 Chr.10: 69630613-69630613 on Build GRCh38
rs1035423238 Chr.10: 69630582-69630582 on Build GRCh38
rs938307081 Chr.10: 69630388-69630388 on Build GRCh38
rs972772769 Chr.10: 69630610-69630610 on Build GRCh38
rs959809286 Chr.10: 69630584-69630584 on Build GRCh38
rs368040246 Chr.10: 69630529-69630529 on Build GRCh38
rs1024193738 Chr.10: 69630551-69630551 on Build GRCh38
rs999587949 Chr.10: 69630217-69630217 on Build GRCh38
rs977096 Chr.10: 69630637-69630637 on Build GRCh38
rs751396007 Chr.10: 69630204-69630204 on Build GRCh38
rs746237706 Chr.10: 69630597-69630597 on Build GRCh38
rs1947741 Chr.10: 69630636-69630636 on Build GRCh38
rs928260670 Chr.10: 69630384-69630384 on Build GRCh38
rs541143828 Chr.10: 69630510-69630510 on Build GRCh38
rs2812579 Chr.10: 69630354-69630354 on Build GRCh38
rs950374488 Chr.10: 69630508-69630508 on Build GRCh38
rs949649153 Chr.10: 69630345-69630345 on Build GRCh38
rs10998885 Chr.10: 69630186-69630186 on Build GRCh38
rs990598459 Chr.10: 69630536-69630536 on Build GRCh38
rs931196774 Chr.10: 69630630-69630630 on Build GRCh38
rs982299008 Chr.10: 69630562-69630562 on Build GRCh38
rs938548767 Chr.10: 69630210-69630210 on Build GRCh38
rs1017371573 Chr.10: 69630453-69630453 on Build GRCh38
rs889831042 Chr.10: 69630283-69630283 on Build GRCh38
rs908964540 Chr.10: 69630292-69630292 on Build GRCh38

View Full Product Details