Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70774097-70774590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBATA
Entrez Gene ID
219793
Gene Name
thymus, brain and testes associated
Gene Aliases
C10orf27, SPATIAL
UniGene
Hs.386698
Gene Chromosome Location
Chr.10: 70771237-70785419 on Build GRCh38
Transcripts
NM_001318241.1 NM_001318242.1 NM_001318243.1 NM_152710.3 NR_134531.1 NR_134532.1 NR_134533.1 NR_134534.1 XM_017015845.1 XM_017015846.1 XM_017015847.1 XM_017015848.1 XM_017015849.1 XM_017015852.1 XM_017015853.1 XM_017015854.1 XM_017015855.1 XM_017015856.1 XM_017015857.1 XM_017015858.1 XM_017015859.1 XM_017015860.1 XM_017015861.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6797084 c.886C>T p.P296S Chr.10: 70774244-70774244 on Build GRCh38
1702600 c.830C>T p.P277L Chr.10: 70774300-70774300 on Build GRCh38
6869751 c.858C>T p.L286L Chr.10: 70774272-70774272 on Build GRCh38
5104571 c.917+9C>T p.? Chr.10: 70774204-70774204 on Build GRCh38
1675324 c.778G>T p.D260Y Chr.10: 70774352-70774352 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932206848 Chr.10: 70774489-70774489 on Build GRCh38
rs767210101 Chr.10: 70774243-70774243 on Build GRCh38
rs983555148 Chr.10: 70774424-70774424 on Build GRCh38
rs897387630 Chr.10: 70774385-70774385 on Build GRCh38
rs751152974 Chr.10: 70774219-70774219 on Build GRCh38
rs748772627 Chr.10: 70774208-70774208 on Build GRCh38
rs945736129 Chr.10: 70774532-70774532 on Build GRCh38
rs1034081403 Chr.10: 70774434-70774434 on Build GRCh38
rs7077672 Chr.10: 70774154-70774154 on Build GRCh38
rs541674352 Chr.10: 70774167-70774167 on Build GRCh38
rs745877492 Chr.10: 70774169-70774169 on Build GRCh38
rs749163580 Chr.10: 70774287-70774287 on Build GRCh38
rs778931572 Chr.10: 70774390-70774390 on Build GRCh38
rs528836640 Chr.10: 70774122-70774122 on Build GRCh38
rs762764506 Chr.10: 70774329-70774329 on Build GRCh38
rs935825388 Chr.10: 70774566-70774566 on Build GRCh38
rs1019965224 Chr.10: 70774499-70774499 on Build GRCh38
rs78222976 Chr.10: 70774128-70774128 on Build GRCh38
rs747162561 Chr.10: 70774301-70774301 on Build GRCh38
rs990782526 Chr.10: 70774188-70774188 on Build GRCh38
rs753705911 Chr.10: 70774373-70774373 on Build GRCh38
rs748457552 Chr.10: 70774324-70774324 on Build GRCh38
rs762468616 Chr.10: 70774211-70774211 on Build GRCh38
rs564432340 Chr.10: 70774130-70774130 on Build GRCh38
rs762292411 Chr.10: 70774200-70774200 on Build GRCh38
rs1024920391 Chr.10: 70774417-70774417 on Build GRCh38
rs137964156 Chr.10: 70774168-70774168 on Build GRCh38
rs765801087 Chr.10: 70774203-70774203 on Build GRCh38
rs768710623 Chr.10: 70774313-70774313 on Build GRCh38
rs752445725 Chr.10: 70774245-70774245 on Build GRCh38
rs970726708 Chr.10: 70774418-70774418 on Build GRCh38
rs376538725 Chr.10: 70774463-70774463 on Build GRCh38
rs775696888 Chr.10: 70774179-70774179 on Build GRCh38
rs7078118 Chr.10: 70774456-70774456 on Build GRCh38
rs942233577 Chr.10: 70774277-70774277 on Build GRCh38
rs200052414 Chr.10: 70774322-70774322 on Build GRCh38
rs371171934 Chr.10: 70774183-70774183 on Build GRCh38
rs143316854 Chr.10: 70774292-70774292 on Build GRCh38
rs571786251 Chr.10: 70774487-70774487 on Build GRCh38
rs555950093 Chr.10: 70774420-70774420 on Build GRCh38
rs188288543 Chr.10: 70774468-70774468 on Build GRCh38
rs944027096 Chr.10: 70774367-70774367 on Build GRCh38
rs551882872 Chr.10: 70774129-70774129 on Build GRCh38
rs192679281 Chr.10: 70774523-70774523 on Build GRCh38
rs371170608 Chr.10: 70774226-70774226 on Build GRCh38
rs372755762 Chr.10: 70774163-70774163 on Build GRCh38
rs1034714372 Chr.10: 70774175-70774175 on Build GRCh38
rs539891487 Chr.10: 70774326-70774326 on Build GRCh38
rs141983416 Chr.10: 70774196-70774196 on Build GRCh38
rs1000113995 Chr.10: 70774158-70774158 on Build GRCh38
rs897439934 Chr.10: 70774153-70774153 on Build GRCh38
rs754558853 Chr.10: 70774238-70774238 on Build GRCh38
rs372705159 Chr.10: 70774372-70774372 on Build GRCh38
rs373435028 Chr.10: 70774515-70774515 on Build GRCh38
rs533241310 Chr.10: 70774346-70774346 on Build GRCh38
rs922188046 Chr.10: 70774296-70774296 on Build GRCh38
rs773860097 Chr.10: 70774204-70774204 on Build GRCh38
rs751554301 Chr.10: 70774396-70774396 on Build GRCh38
rs529407684 Chr.10: 70774181-70774181 on Build GRCh38
rs772054555 Chr.10: 70774173-70774173 on Build GRCh38
rs1012174147 Chr.10: 70774412-70774412 on Build GRCh38
rs780744914 Chr.10: 70774542-70774542 on Build GRCh38
rs771513454 Chr.10: 70774338-70774338 on Build GRCh38
rs1034229918 Chr.10: 70774393-70774393 on Build GRCh38
rs376260477 Chr.10: 70774248-70774248 on Build GRCh38
rs1028618481 Chr.10: 70774506-70774506 on Build GRCh38
rs911555838 Chr.10: 70774522-70774522 on Build GRCh38
rs757119632 Chr.10: 70774382-70774382 on Build GRCh38
rs36005481 Chr.10: 70774547-70774548 on Build GRCh38
rs773771976 Chr.10: 70774333-70774333 on Build GRCh38
rs566171083 Chr.10: 70774263-70774263 on Build GRCh38
rs150078967 Chr.10: 70774347-70774347 on Build GRCh38
rs531488986 Chr.10: 70774134-70774134 on Build GRCh38
rs537312540 Chr.10: 70774400-70774400 on Build GRCh38
rs373624773 Chr.10: 70774298-70774298 on Build GRCh38
rs139405249 Chr.10: 70774349-70774349 on Build GRCh38
rs377256258 Chr.10: 70774544-70774544 on Build GRCh38
rs888604055 Chr.10: 70774421-70774421 on Build GRCh38
rs749889988 Chr.10: 70774547-70774547 on Build GRCh38
rs904272497 Chr.10: 70774557-70774557 on Build GRCh38
rs907988632 Chr.10: 70774432-70774432 on Build GRCh38
rs763368087 Chr.10: 70774335-70774335 on Build GRCh38
rs368653790 Chr.10: 70774464-70774464 on Build GRCh38

View Full Product Details