Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 71446179-71446722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH23
Entrez Gene ID
64072
Gene Name
cadherin-related 23
Gene Aliases
CDHR23, USH1D
UniGene
Hs.656032
Gene Chromosome Location
Chr.10: 71396934-71815947 on Build GRCh38
Transcripts
NM_001171930.1 NM_001171931.1 NM_001171932.1 NM_022124.5 NM_052836.3 XM_006717940.3 XM_006717942.3 XM_011540039.2 XM_011540042.2 XM_011540043.2 XM_011540044.2 XM_011540045.2 XM_011540048.2 XM_011540049.2 XM_011540051.2 XM_017016499.1 XM_017016500.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5768762 c.100C>T p.H34Y Chr.10: 71446350-71446350 on Build GRCh38
3867731 c.130G>A p.E44K Chr.10: 71446380-71446380 on Build GRCh38
3382977 c.264C>T p.S88S Chr.10: 71446379-71446379 on Build GRCh38
6417440 c.71A>T p.Q24L Chr.10: 71446321-71446321 on Build GRCh38
7139592 c.105C>A p.F35L Chr.10: 71446355-71446355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766291656 Chr.10: 71446351-71446351 on Build GRCh38
rs370760723 Chr.10: 71446329-71446329 on Build GRCh38
rs727504613 Chr.10: 71446387-71446387 on Build GRCh38
rs142456469 Chr.10: 71446315-71446315 on Build GRCh38
rs1005656005 Chr.10: 71446594-71446595 on Build GRCh38
rs373826012 Chr.10: 71446472-71446472 on Build GRCh38
rs780679219 Chr.10: 71446428-71446428 on Build GRCh38
rs901468434 Chr.10: 71446543-71446543 on Build GRCh38
rs371310149 Chr.10: 71446422-71446422 on Build GRCh38
rs543947136 Chr.10: 71446267-71446267 on Build GRCh38
rs769381881 Chr.10: 71446433-71446433 on Build GRCh38
rs765567124 Chr.10: 71446405-71446405 on Build GRCh38
rs201925753 Chr.10: 71446379-71446379 on Build GRCh38
rs763659234 Chr.10: 71446402-71446402 on Build GRCh38
rs575030372 Chr.10: 71446562-71446562 on Build GRCh38
rs1040049438 Chr.10: 71446538-71446538 on Build GRCh38
rs746220827 Chr.10: 71446286-71446286 on Build GRCh38
rs375566405 Chr.10: 71446377-71446377 on Build GRCh38
rs935906056 Chr.10: 71446217-71446217 on Build GRCh38
rs1017177424 Chr.10: 71446594-71446594 on Build GRCh38
rs1047443844 Chr.10: 71446554-71446554 on Build GRCh38
rs924457478 Chr.10: 71446426-71446426 on Build GRCh38
rs182368040 Chr.10: 71446593-71446593 on Build GRCh38
rs367953860 Chr.10: 71446295-71446295 on Build GRCh38
rs889721196 Chr.10: 71446451-71446451 on Build GRCh38
rs187159287 Chr.10: 71446475-71446475 on Build GRCh38
rs557193960 Chr.10: 71446602-71446602 on Build GRCh38
rs12770610 Chr.10: 71446530-71446530 on Build GRCh38
rs190087532 Chr.10: 71446548-71446548 on Build GRCh38
rs755540063 Chr.10: 71446427-71446427 on Build GRCh38
rs757609099 Chr.10: 71446282-71446282 on Build GRCh38
rs372660833 Chr.10: 71446401-71446401 on Build GRCh38
rs956069480 Chr.10: 71446352-71446352 on Build GRCh38
rs756171646 Chr.10: 71446270-71446270 on Build GRCh38
rs896862264 Chr.10: 71446224-71446224 on Build GRCh38
rs999274149 Chr.10: 71446536-71446536 on Build GRCh38
rs776170967 Chr.10: 71446317-71446317 on Build GRCh38
rs918697176 Chr.10: 71446694-71446694 on Build GRCh38
rs972781642 Chr.10: 71446638-71446638 on Build GRCh38
rs534252168 Chr.10: 71446404-71446404 on Build GRCh38
rs747663591 Chr.10: 71446432-71446432 on Build GRCh38
rs772762752 Chr.10: 71446444-71446444 on Build GRCh38
rs113798012 Chr.10: 71446478-71446478 on Build GRCh38
rs772569888 Chr.10: 71446289-71446289 on Build GRCh38
rs754089636 Chr.10: 71446280-71446280 on Build GRCh38
rs956178846 Chr.10: 71446552-71446552 on Build GRCh38
rs900885333 Chr.10: 71446531-71446531 on Build GRCh38
rs762669711 Chr.10: 71446348-71446348 on Build GRCh38
rs554264957 Chr.10: 71446596-71446596 on Build GRCh38
rs575567480 Chr.10: 71446609-71446609 on Build GRCh38
rs536563942 Chr.10: 71446678-71446678 on Build GRCh38
rs761684829 Chr.10: 71446589-71446589 on Build GRCh38
rs769504706 Chr.10: 71446330-71446330 on Build GRCh38
rs764254196 Chr.10: 71446388-71446388 on Build GRCh38
rs115158212 Chr.10: 71446421-71446421 on Build GRCh38
rs990090244 Chr.10: 71446473-71446473 on Build GRCh38
rs566545152 Chr.10: 71446523-71446523 on Build GRCh38
rs990174419 Chr.10: 71446378-71446378 on Build GRCh38
rs535954072 Chr.10: 71446586-71446586 on Build GRCh38
rs765951640 Chr.10: 71446278-71446278 on Build GRCh38
rs1024311219 Chr.10: 71446631-71446631 on Build GRCh38
rs79513610 Chr.10: 71446274-71446274 on Build GRCh38
rs772995753 Chr.10: 71446346-71446346 on Build GRCh38
rs995134101 Chr.10: 71446239-71446239 on Build GRCh38
rs780443284 Chr.10: 71446291-71446291 on Build GRCh38
rs1048291491 Chr.10: 71446431-71446431 on Build GRCh38
rs993621392 Chr.10: 71446364-71446364 on Build GRCh38
rs1036783146 Chr.10: 71446416-71446416 on Build GRCh38
rs1054345210 Chr.10: 71446223-71446223 on Build GRCh38
rs1006530352 Chr.10: 71446512-71446512 on Build GRCh38
rs762876228 Chr.10: 71446414-71446414 on Build GRCh38
rs753962913 Chr.10: 71446629-71446629 on Build GRCh38
rs200685398 Chr.10: 71446283-71446283 on Build GRCh38
rs375882842 Chr.10: 71446418-71446418 on Build GRCh38
rs750084299 Chr.10: 71446299-71446299 on Build GRCh38
rs971780263 Chr.10: 71446660-71446660 on Build GRCh38

View Full Product Details