Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 71645739-71646244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH23
Entrez Gene ID
64072
Gene Name
cadherin-related 23
Gene Aliases
CDHR23, USH1D
UniGene
Hs.656032
Gene Chromosome Location
Chr.10: 71396934-71815947 on Build GRCh38
Transcripts
NM_001171930.1 NM_001171931.1 NM_022124.5 NM_052836.3 XM_006717940.3 XM_006717942.3 XM_011540039.2 XM_011540042.2 XM_011540043.2 XM_011540044.2 XM_011540045.2 XM_011540046.2 XM_011540047.2 XM_011540048.2 XM_011540049.2 XM_011540051.2 XM_017016499.1 XM_017016500.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2160953 c.1348G>A p.V450I Chr.10: 71645903-71645903 on Build GRCh38
1348903 c.1256G>A p.R419Q Chr.10: 71645931-71645931 on Build GRCh38
2160952 c.1228G>A p.V410I Chr.10: 71645903-71645903 on Build GRCh38
7269730 c.1361C>T p.A454V Chr.10: 71645916-71645916 on Build GRCh38
6551031 c.1242G>T p.A414A Chr.10: 71645917-71645917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757456114 Chr.10: 71645967-71645967 on Build GRCh38
rs771275265 Chr.10: 71645831-71645831 on Build GRCh38
rs776232014 Chr.10: 71646008-71646008 on Build GRCh38
rs993003646 Chr.10: 71645808-71645808 on Build GRCh38
rs975434291 Chr.10: 71646094-71646094 on Build GRCh38
rs903771460 Chr.10: 71646204-71646204 on Build GRCh38
rs772522793 Chr.10: 71645914-71645914 on Build GRCh38
rs763742877 Chr.10: 71645854-71645854 on Build GRCh38
rs1018406764 Chr.10: 71646101-71646101 on Build GRCh38
rs373168635 Chr.10: 71645895-71645895 on Build GRCh38
rs377441168 Chr.10: 71645902-71645902 on Build GRCh38
rs775021846 Chr.10: 71645852-71645852 on Build GRCh38
rs34974340 Chr.10: 71645986-71645987 on Build GRCh38
rs575019489 Chr.10: 71646067-71646067 on Build GRCh38
rs532687616 Chr.10: 71646010-71646010 on Build GRCh38
rs750369111 Chr.10: 71645811-71645811 on Build GRCh38
rs752156210 Chr.10: 71645775-71645775 on Build GRCh38
rs893967639 Chr.10: 71646075-71646075 on Build GRCh38
rs193192069 Chr.10: 71646097-71646097 on Build GRCh38
rs761623919 Chr.10: 71645869-71645869 on Build GRCh38
rs562546644 Chr.10: 71646038-71646038 on Build GRCh38
rs772474417 Chr.10: 71645843-71645843 on Build GRCh38
rs376657497 Chr.10: 71645915-71645915 on Build GRCh38
rs769297838 Chr.10: 71645992-71645992 on Build GRCh38
rs370014467 Chr.10: 71645888-71645888 on Build GRCh38
rs780628787 Chr.10: 71645974-71645974 on Build GRCh38
rs375312983 Chr.10: 71645908-71645908 on Build GRCh38
rs188376296 Chr.10: 71645972-71645972 on Build GRCh38
rs769991253 Chr.10: 71646087-71646087 on Build GRCh38
rs775050967 Chr.10: 71646016-71646016 on Build GRCh38
rs774139031 Chr.10: 71646018-71646018 on Build GRCh38
rs538941658 Chr.10: 71646211-71646211 on Build GRCh38
rs758272167 Chr.10: 71645813-71645813 on Build GRCh38
rs571318482 Chr.10: 71646209-71646209 on Build GRCh38
rs374419277 Chr.10: 71645924-71645924 on Build GRCh38
rs534810179 Chr.10: 71645826-71645826 on Build GRCh38
rs984049246 Chr.10: 71646146-71646146 on Build GRCh38
rs918596431 Chr.10: 71645911-71645911 on Build GRCh38
rs998695785 Chr.10: 71646116-71646116 on Build GRCh38
rs377492133 Chr.10: 71645885-71645885 on Build GRCh38
rs577849058 Chr.10: 71645841-71645841 on Build GRCh38
rs1024456111 Chr.10: 71646046-71646046 on Build GRCh38
rs758305358 Chr.10: 71645881-71645881 on Build GRCh38
rs200103711 Chr.10: 71646021-71646021 on Build GRCh38
rs186887035 Chr.10: 71645816-71645816 on Build GRCh38
rs747452182 Chr.10: 71645978-71645978 on Build GRCh38
rs3214768 Chr.10: 71646216-71646217 on Build GRCh38
rs750193646 Chr.10: 71645933-71645933 on Build GRCh38
rs769964522 Chr.10: 71645819-71645819 on Build GRCh38
rs111825773 Chr.10: 71646039-71646039 on Build GRCh38
rs903488588 Chr.10: 71645991-71645991 on Build GRCh38
rs763867901 Chr.10: 71645780-71645780 on Build GRCh38
rs762624678 Chr.10: 71645943-71645943 on Build GRCh38
rs747106189 Chr.10: 71645839-71645839 on Build GRCh38
rs557243019 Chr.10: 71646221-71646221 on Build GRCh38
rs371212430 Chr.10: 71645917-71645917 on Build GRCh38
rs986606399 Chr.10: 71646133-71646133 on Build GRCh38
rs372340806 Chr.10: 71645959-71645959 on Build GRCh38
rs937702926 Chr.10: 71646216-71646216 on Build GRCh38
rs945296076 Chr.10: 71646192-71646192 on Build GRCh38
rs2305207 Chr.10: 71645960-71645960 on Build GRCh38
rs528720730 Chr.10: 71645953-71645953 on Build GRCh38
rs781398004 Chr.10: 71645939-71645939 on Build GRCh38
rs761319618 Chr.10: 71645916-71645916 on Build GRCh38
rs192730794 Chr.10: 71645872-71645872 on Build GRCh38
rs61851983 Chr.10: 71646112-71646112 on Build GRCh38
rs553029568 Chr.10: 71645827-71645827 on Build GRCh38
rs368817860 Chr.10: 71645925-71645925 on Build GRCh38
rs909841173 Chr.10: 71646179-71646179 on Build GRCh38
rs371302682 Chr.10: 71646140-71646140 on Build GRCh38
rs530034524 Chr.10: 71646044-71646044 on Build GRCh38
rs561032891 Chr.10: 71645931-71645931 on Build GRCh38
rs751486021 Chr.10: 71645954-71645954 on Build GRCh38
rs367744918 Chr.10: 71646020-71646020 on Build GRCh38
rs1016951100 Chr.10: 71646072-71646072 on Build GRCh38
rs755776524 Chr.10: 71645777-71645777 on Build GRCh38
rs397517304 Chr.10: 71645896-71645896 on Build GRCh38
rs892398847 Chr.10: 71645947-71645947 on Build GRCh38
rs370549448 Chr.10: 71645903-71645903 on Build GRCh38
rs554597140 Chr.10: 71645790-71645790 on Build GRCh38
rs951623307 Chr.10: 71646058-71646058 on Build GRCh38
rs2305208 Chr.10: 71646197-71646197 on Build GRCh38
rs778878494 Chr.10: 71645773-71645773 on Build GRCh38
rs772719809 Chr.10: 71646016-71646016 on Build GRCh38
rs534545913 Chr.10: 71646139-71646139 on Build GRCh38
rs373674747 Chr.10: 71645966-71645966 on Build GRCh38
rs369803887 Chr.10: 71645858-71645858 on Build GRCh38
rs1043380204 Chr.10: 71645988-71645988 on Build GRCh38
rs375085534 Chr.10: 71645807-71645807 on Build GRCh38
rs185105210 Chr.10: 71645875-71645875 on Build GRCh38
rs367782695 Chr.10: 71646129-71646129 on Build GRCh38
rs764949139 Chr.10: 71645830-71645863 on Build GRCh38
rs397840008 Chr.10: 71646217-71646218 on Build GRCh38
rs746146461 Chr.10: 71645968-71645968 on Build GRCh38
rs781274193 Chr.10: 71645814-71645814 on Build GRCh38
rs34028302 Chr.10: 71646091-71646092 on Build GRCh38
rs909705325 Chr.10: 71645930-71645930 on Build GRCh38

View Full Product Details