Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 72009489-72009994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST3
Entrez Gene ID
9469
Gene Name
carbohydrate sulfotransferase 3
Gene Aliases
C6ST, C6ST1, HSD
UniGene
Hs.158304
Gene Chromosome Location
Chr.10: 71964362-72013564 on Build GRCh38
Transcripts
NM_004273.4 XM_006718075.3 XM_011540369.2

SNP Detail

SNP ID Chromosome Location
rs1001717157 Chr.10: 72009552-72009552 on Build GRCh38
rs996673640 Chr.10: 72009790-72009790 on Build GRCh38
rs964815410 Chr.10: 72009576-72009576 on Build GRCh38
rs544195789 Chr.10: 72009797-72009797 on Build GRCh38
rs933667281 Chr.10: 72009867-72009867 on Build GRCh38
rs34828900 Chr.10: 72009626-72009627 on Build GRCh38
rs756598951 Chr.10: 72009671-72009671 on Build GRCh38
rs201456562 Chr.10: 72009816-72009816 on Build GRCh38
rs954701706 Chr.10: 72009615-72009615 on Build GRCh38
rs745509297 Chr.10: 72009974-72009974 on Build GRCh38
rs985281879 Chr.10: 72009624-72009624 on Build GRCh38
rs916903244 Chr.10: 72009740-72009740 on Build GRCh38
rs886047166 Chr.10: 72009524-72009524 on Build GRCh38
rs909525435 Chr.10: 72009972-72009972 on Build GRCh38
rs886047169 Chr.10: 72009727-72009727 on Build GRCh38
rs889570871 Chr.10: 72009724-72009724 on Build GRCh38
rs534540774 Chr.10: 72009933-72009933 on Build GRCh38
rs1020693097 Chr.10: 72009868-72009868 on Build GRCh38
rs4148942 Chr.10: 72009601-72009601 on Build GRCh38
rs535090592 Chr.10: 72009856-72009857 on Build GRCh38
rs755041484 Chr.10: 72009619-72009619 on Build GRCh38
rs529839491 Chr.10: 72009934-72009934 on Build GRCh38
rs958699475 Chr.10: 72009971-72009971 on Build GRCh38
rs373515105 Chr.10: 72009656-72009656 on Build GRCh38
rs76808415 Chr.10: 72009918-72009918 on Build GRCh38
rs998286732 Chr.10: 72009689-72009689 on Build GRCh38
rs538070743 Chr.10: 72009967-72009967 on Build GRCh38
rs775020468 Chr.10: 72009961-72009961 on Build GRCh38
rs1006427623 Chr.10: 72009562-72009562 on Build GRCh38
rs993066875 Chr.10: 72009921-72009921 on Build GRCh38
rs925868127 Chr.10: 72009625-72009625 on Build GRCh38
rs981435017 Chr.10: 72009948-72009948 on Build GRCh38
rs1030379309 Chr.10: 72009802-72009802 on Build GRCh38
rs937102794 Chr.10: 72009674-72009674 on Build GRCh38
rs927390810 Chr.10: 72009654-72009654 on Build GRCh38
rs758894650 Chr.10: 72009735-72009735 on Build GRCh38
rs902114662 Chr.10: 72009866-72009866 on Build GRCh38
rs997908276 Chr.10: 72009592-72009592 on Build GRCh38
rs886047165 Chr.10: 72009513-72009513 on Build GRCh38
rs886047168 Chr.10: 72009715-72009715 on Build GRCh38
rs76062174 Chr.10: 72009897-72009897 on Build GRCh38
rs927134523 Chr.10: 72009902-72009902 on Build GRCh38
rs937443692 Chr.10: 72009796-72009796 on Build GRCh38
rs895452223 Chr.10: 72009521-72009521 on Build GRCh38
rs4148945 Chr.10: 72009832-72009832 on Build GRCh38
rs1045788383 Chr.10: 72009649-72009650 on Build GRCh38
rs971760727 Chr.10: 72009944-72009944 on Build GRCh38
rs577339997 Chr.10: 72009957-72009957 on Build GRCh38
rs4747236 Chr.10: 72009627-72009627 on Build GRCh38
rs1037893512 Chr.10: 72009777-72009777 on Build GRCh38
rs886047167 Chr.10: 72009562-72009562 on Build GRCh38
rs886047170 Chr.10: 72009875-72009875 on Build GRCh38
rs886719426 Chr.10: 72009770-72009770 on Build GRCh38
rs4148944 Chr.10: 72009785-72009785 on Build GRCh38
rs577247765 Chr.10: 72009600-72009600 on Build GRCh38
rs80269486 Chr.10: 72009896-72009896 on Build GRCh38
rs1006461774 Chr.10: 72009771-72009771 on Build GRCh38
rs866928391 Chr.10: 72009683-72009683 on Build GRCh38
rs536272249 Chr.10: 72009544-72009544 on Build GRCh38
rs1029347183 Chr.10: 72009705-72009705 on Build GRCh38
rs116999241 Chr.10: 72009572-72009572 on Build GRCh38
rs528427679 Chr.10: 72009737-72009737 on Build GRCh38
rs1006670007 Chr.10: 72009731-72009731 on Build GRCh38
rs765379561 Chr.10: 72009632-72009632 on Build GRCh38
rs556197350 Chr.10: 72009969-72009969 on Build GRCh38
rs372345396 Chr.10: 72009838-72009839 on Build GRCh38
rs4148943 Chr.10: 72009749-72009749 on Build GRCh38
rs562405951 Chr.10: 72009870-72009870 on Build GRCh38
rs60924680 Chr.10: 72009841-72009841 on Build GRCh38
rs1020183023 Chr.10: 72009942-72009942 on Build GRCh38
rs989141637 Chr.10: 72009627-72009627 on Build GRCh38
rs757821498 Chr.10: 72009856-72009856 on Build GRCh38
rs115617050 Chr.10: 72009559-72009559 on Build GRCh38
rs984883743 Chr.10: 72009824-72009824 on Build GRCh38
rs1014260218 Chr.10: 72009534-72009534 on Build GRCh38
rs187692473 Chr.10: 72009655-72009655 on Build GRCh38
rs778280355 Chr.10: 72009747-72009747 on Build GRCh38
rs560847494 Chr.10: 72009672-72009672 on Build GRCh38
rs148125052 Chr.10: 72009561-72009561 on Build GRCh38
rs906007154 Chr.10: 72009538-72009538 on Build GRCh38

View Full Product Details