Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 72010970-72011508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST3
Entrez Gene ID
9469
Gene Name
carbohydrate sulfotransferase 3
Gene Aliases
C6ST, C6ST1, HSD
UniGene
Hs.158304
Gene Chromosome Location
Chr.10: 71964362-72013564 on Build GRCh38
Transcripts
NM_004273.4 XM_006718075.3 XM_011540369.2

SNP Detail

SNP ID Chromosome Location
rs1038004520 Chr.10: 72011124-72011124 on Build GRCh38
rs768431224 Chr.10: 72011151-72011152 on Build GRCh38
rs982011982 Chr.10: 72011482-72011482 on Build GRCh38
rs182900996 Chr.10: 72011369-72011369 on Build GRCh38
rs542056241 Chr.10: 72011427-72011427 on Build GRCh38
rs908522906 Chr.10: 72011059-72011059 on Build GRCh38
rs893918063 Chr.10: 72011397-72011397 on Build GRCh38
rs571119719 Chr.10: 72011039-72011039 on Build GRCh38
rs886047178 Chr.10: 72011314-72011315 on Build GRCh38
rs982775712 Chr.10: 72011256-72011256 on Build GRCh38
rs144248670 Chr.10: 72011326-72011326 on Build GRCh38
rs977140771 Chr.10: 72011015-72011018 on Build GRCh38
rs115489836 Chr.10: 72011189-72011189 on Build GRCh38
rs187078225 Chr.10: 72011031-72011031 on Build GRCh38
rs893414790 Chr.10: 72011370-72011370 on Build GRCh38
rs890458768 Chr.10: 72011168-72011168 on Build GRCh38
rs768318754 Chr.10: 72011293-72011293 on Build GRCh38
rs916015046 Chr.10: 72011002-72011002 on Build GRCh38
rs991246419 Chr.10: 72011056-72011058 on Build GRCh38
rs1000522642 Chr.10: 72011366-72011366 on Build GRCh38
rs748988264 Chr.10: 72011200-72011200 on Build GRCh38
rs940040640 Chr.10: 72011301-72011301 on Build GRCh38
rs573559460 Chr.10: 72011445-72011445 on Build GRCh38
rs746330162 Chr.10: 72011006-72011006 on Build GRCh38
rs932565615 Chr.10: 72011316-72011316 on Build GRCh38
rs555301573 Chr.10: 72011188-72011188 on Build GRCh38
rs967028862 Chr.10: 72011250-72011250 on Build GRCh38
rs577193207 Chr.10: 72011324-72011324 on Build GRCh38
rs890933998 Chr.10: 72011325-72011325 on Build GRCh38
rs147227559 Chr.10: 72011214-72011214 on Build GRCh38
rs143743853 Chr.10: 72011131-72011131 on Build GRCh38
rs1023116442 Chr.10: 72011287-72011287 on Build GRCh38
rs915719326 Chr.10: 72011061-72011061 on Build GRCh38
rs773949378 Chr.10: 72011298-72011298 on Build GRCh38
rs1049567104 Chr.10: 72011157-72011157 on Build GRCh38
rs957495780 Chr.10: 72010994-72010994 on Build GRCh38
rs771840365 Chr.10: 72011337-72011337 on Build GRCh38
rs114457501 Chr.10: 72011297-72011297 on Build GRCh38
rs531918105 Chr.10: 72011115-72011115 on Build GRCh38
rs1013146677 Chr.10: 72011407-72011407 on Build GRCh38
rs1035322851 Chr.10: 72011274-72011274 on Build GRCh38
rs943390469 Chr.10: 72011382-72011382 on Build GRCh38
rs779081972 Chr.10: 72011033-72011037 on Build GRCh38
rs527242888 Chr.10: 72011017-72011017 on Build GRCh38
rs1033264754 Chr.10: 72011208-72011208 on Build GRCh38
rs919869114 Chr.10: 72011307-72011307 on Build GRCh38
rs974060429 Chr.10: 72011303-72011303 on Build GRCh38
rs747937392 Chr.10: 72011327-72011327 on Build GRCh38
rs755645339 Chr.10: 72011140-72011140 on Build GRCh38
rs915688099 Chr.10: 72011314-72011315 on Build GRCh38
rs191889682 Chr.10: 72011062-72011062 on Build GRCh38
rs902691502 Chr.10: 72011454-72011454 on Build GRCh38
rs1039075430 Chr.10: 72011383-72011383 on Build GRCh38
rs886047177 Chr.10: 72011127-72011127 on Build GRCh38
rs371988923 Chr.10: 72011196-72011196 on Build GRCh38
rs140272473 Chr.10: 72011311-72011311 on Build GRCh38
rs886047176 Chr.10: 72011063-72011063 on Build GRCh38
rs890893894 Chr.10: 72011183-72011183 on Build GRCh38
rs1042524555 Chr.10: 72011437-72011437 on Build GRCh38
rs554201020 Chr.10: 72011238-72011238 on Build GRCh38
rs1023570647 Chr.10: 72011422-72011422 on Build GRCh38
rs1021609861 Chr.10: 72011207-72011207 on Build GRCh38
rs1038252711 Chr.10: 72011067-72011067 on Build GRCh38
rs929289924 Chr.10: 72011132-72011132 on Build GRCh38
rs560145890 Chr.10: 72011428-72011428 on Build GRCh38
rs556516377 Chr.10: 72011340-72011340 on Build GRCh38
rs941893093 Chr.10: 72011091-72011091 on Build GRCh38
rs1050042966 Chr.10: 72011320-72011320 on Build GRCh38
rs991671222 Chr.10: 72010998-72010998 on Build GRCh38
rs938374261 Chr.10: 72011044-72011045 on Build GRCh38
rs566046898 Chr.10: 72011279-72011279 on Build GRCh38
rs999065896 Chr.10: 72011206-72011206 on Build GRCh38
rs971909460 Chr.10: 72011292-72011292 on Build GRCh38
rs939935847 Chr.10: 72011063-72011063 on Build GRCh38

View Full Product Details