Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 72216119-72216616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC16
Entrez Gene ID
119504
Gene Name
anaphase promoting complex subunit 16
Gene Aliases
APC16, C10orf104, CENP-27, MSAG, bA570G20.3
UniGene
Hs.426296
Gene Chromosome Location
Chr.10: 72216000-72235860 on Build GRCh38
Transcripts
NM_001242546.1 NM_001242547.1 NM_001242548.1 NM_173473.3 NR_038391.1 NR_038392.1 NM_001198799.2 NM_001198800.2 NR_045564.1 XM_006717873.3 XM_011539841.2 XM_011539842.2 XM_017016291.1 XM_017016292.1 XM_017016293.1 XM_017016294.1 XM_017016295.1 XM_017016297.1 XM_017016298.1 XM_017016299.1 XM_017016300.1 XM_017016301.1 XM_017016302.1

Gene Symbol
ASCC1
Entrez Gene ID
51008
Gene Name
activating signal cointegrator 1 complex subunit 1
Gene Aliases
ASC1p50, CGI-18, SMABF2, p50
UniGene
Hs.500007
Gene Chromosome Location
Chr.10: 72096032-72217134 on Build GRCh38
Transcripts
NM_001242546.1 NM_001242547.1 NM_001242548.1 NM_173473.3 NR_038391.1 NR_038392.1 NM_001198799.2 NM_001198800.2 NR_045564.1 XM_006717873.3 XM_011539841.2 XM_011539842.2 XM_017016291.1 XM_017016292.1 XM_017016293.1 XM_017016294.1 XM_017016295.1 XM_017016297.1 XM_017016298.1 XM_017016299.1 XM_017016300.1 XM_017016301.1 XM_017016302.1

SNP Detail

SNP ID Chromosome Location
rs532484903 Chr.10: 72216273-72216273 on Build GRCh38
rs66848140 Chr.10: 72216494-72216495 on Build GRCh38
rs893795868 Chr.10: 72216542-72216542 on Build GRCh38
rs530731099 Chr.10: 72216303-72216303 on Build GRCh38
rs1050924801 Chr.10: 72216397-72216397 on Build GRCh38
rs928211195 Chr.10: 72216347-72216347 on Build GRCh38
rs1020166201 Chr.10: 72216298-72216323 on Build GRCh38
rs1020872382 Chr.10: 72216295-72216295 on Build GRCh38
rs74844751 Chr.10: 72216483-72216483 on Build GRCh38
rs73276918 Chr.10: 72216514-72216514 on Build GRCh38
rs959746579 Chr.10: 72216319-72216319 on Build GRCh38
rs899561919 Chr.10: 72216374-72216374 on Build GRCh38
rs897642830 Chr.10: 72216277-72216277 on Build GRCh38
rs1010981601 Chr.10: 72216582-72216582 on Build GRCh38
rs902619661 Chr.10: 72216463-72216463 on Build GRCh38
rs540734293 Chr.10: 72216245-72216245 on Build GRCh38
rs144621123 Chr.10: 72216341-72216341 on Build GRCh38
rs567128943 Chr.10: 72216342-72216342 on Build GRCh38
rs940996871 Chr.10: 72216362-72216362 on Build GRCh38
rs1039027928 Chr.10: 72216537-72216537 on Build GRCh38
rs919124879 Chr.10: 72216393-72216393 on Build GRCh38
rs143967411 Chr.10: 72216481-72216482 on Build GRCh38
rs983704047 Chr.10: 72216355-72216355 on Build GRCh38
rs573425489 Chr.10: 72216493-72216495 on Build GRCh38
rs1051565979 Chr.10: 72216434-72216434 on Build GRCh38
rs1037555966 Chr.10: 72216270-72216270 on Build GRCh38
rs915673246 Chr.10: 72216243-72216243 on Build GRCh38
rs938346939 Chr.10: 72216237-72216237 on Build GRCh38
rs886789802 Chr.10: 72216166-72216166 on Build GRCh38
rs932421541 Chr.10: 72216392-72216392 on Build GRCh38
rs959618471 Chr.10: 72216348-72216348 on Build GRCh38
rs918094331 Chr.10: 72216346-72216346 on Build GRCh38
rs899613651 Chr.10: 72216238-72216238 on Build GRCh38
rs1038080625 Chr.10: 72216365-72216365 on Build GRCh38
rs191655357 Chr.10: 72216279-72216279 on Build GRCh38
rs929578887 Chr.10: 72216156-72216156 on Build GRCh38
rs149733377 Chr.10: 72216275-72216275 on Build GRCh38
rs565419206 Chr.10: 72216257-72216257 on Build GRCh38
rs890958093 Chr.10: 72216404-72216404 on Build GRCh38
rs377433738 Chr.10: 72216234-72216234 on Build GRCh38
rs890375456 Chr.10: 72216495-72216495 on Build GRCh38
rs908173354 Chr.10: 72216359-72216359 on Build GRCh38
rs372919139 Chr.10: 72216221-72216221 on Build GRCh38
rs1052106954 Chr.10: 72216289-72216289 on Build GRCh38
rs752292902 Chr.10: 72216201-72216201 on Build GRCh38
rs929548949 Chr.10: 72216352-72216352 on Build GRCh38
rs113170950 Chr.10: 72216382-72216382 on Build GRCh38
rs543314461 Chr.10: 72216185-72216185 on Build GRCh38
rs754419295 Chr.10: 72216301-72216301 on Build GRCh38
rs1009406643 Chr.10: 72216412-72216412 on Build GRCh38
rs1020821451 Chr.10: 72216501-72216501 on Build GRCh38
rs982759036 Chr.10: 72216227-72216227 on Build GRCh38
rs914042333 Chr.10: 72216213-72216213 on Build GRCh38
rs571351175 Chr.10: 72216457-72216457 on Build GRCh38
rs757902333 Chr.10: 72216204-72216204 on Build GRCh38
rs73276916 Chr.10: 72216453-72216453 on Build GRCh38
rs534556356 Chr.10: 72216394-72216394 on Build GRCh38
rs926726764 Chr.10: 72216313-72216313 on Build GRCh38
rs918159388 Chr.10: 72216154-72216154 on Build GRCh38
rs941733303 Chr.10: 72216264-72216264 on Build GRCh38
rs1035096586 Chr.10: 72216474-72216474 on Build GRCh38
rs1046733331 Chr.10: 72216165-72216165 on Build GRCh38
rs955303567 Chr.10: 72216285-72216285 on Build GRCh38
rs929127641 Chr.10: 72216398-72216398 on Build GRCh38
rs1003599276 Chr.10: 72216469-72216469 on Build GRCh38
rs1009458896 Chr.10: 72216294-72216294 on Build GRCh38

View Full Product Details