Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 72233935-72234480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC16
Entrez Gene ID
119504
Gene Name
anaphase promoting complex subunit 16
Gene Aliases
APC16, C10orf104, CENP-27, MSAG, bA570G20.3
UniGene
Hs.426296
Gene Chromosome Location
Chr.10: 72216000-72235860 on Build GRCh38
Transcripts
NM_001242546.1 NM_001242547.1 NM_001242548.1 NM_173473.3 NR_038391.1 NR_038392.1

SNP Detail

SNP ID Chromosome Location
rs201470688 Chr.10: 72234253-72234253 on Build GRCh38
rs565422118 Chr.10: 72234060-72234060 on Build GRCh38
rs373052716 Chr.10: 72234397-72234397 on Build GRCh38
rs914697267 Chr.10: 72233993-72233993 on Build GRCh38
rs997417409 Chr.10: 72234196-72234196 on Build GRCh38
rs900366964 Chr.10: 72234178-72234178 on Build GRCh38
rs576848339 Chr.10: 72234038-72234038 on Build GRCh38
rs761391492 Chr.10: 72234111-72234111 on Build GRCh38
rs1057343639 Chr.10: 72234049-72234049 on Build GRCh38
rs544259551 Chr.10: 72234428-72234428 on Build GRCh38
rs373738531 Chr.10: 72234070-72234070 on Build GRCh38
rs181810221 Chr.10: 72234059-72234059 on Build GRCh38
rs1010628907 Chr.10: 72234445-72234445 on Build GRCh38
rs1017986043 Chr.10: 72234179-72234179 on Build GRCh38
rs938690844 Chr.10: 72234015-72234015 on Build GRCh38
rs7922264 Chr.10: 72234425-72234425 on Build GRCh38
rs983159017 Chr.10: 72234388-72234388 on Build GRCh38
rs889020958 Chr.10: 72234043-72234043 on Build GRCh38
rs1055763711 Chr.10: 72234058-72234058 on Build GRCh38
rs1010246682 Chr.10: 72234256-72234256 on Build GRCh38
rs112124969 Chr.10: 72234338-72234338 on Build GRCh38
rs915939324 Chr.10: 72234310-72234310 on Build GRCh38
rs539074465 Chr.10: 72234069-72234069 on Build GRCh38
rs891721037 Chr.10: 72234442-72234442 on Build GRCh38
rs77300823 Chr.10: 72234408-72234408 on Build GRCh38
rs186440920 Chr.10: 72234430-72234430 on Build GRCh38
rs576692978 Chr.10: 72234296-72234296 on Build GRCh38
rs199668653 Chr.10: 72234009-72234009 on Build GRCh38
rs535266289 Chr.10: 72234009-72234009 on Build GRCh38
rs1022089505 Chr.10: 72234265-72234265 on Build GRCh38
rs964291249 Chr.10: 72234190-72234190 on Build GRCh38
rs933175677 Chr.10: 72234429-72234429 on Build GRCh38
rs1021429209 Chr.10: 72234251-72234251 on Build GRCh38
rs1021852418 Chr.10: 72234216-72234216 on Build GRCh38
rs5786065 Chr.10: 72234023-72234024 on Build GRCh38
rs545433712 Chr.10: 72234455-72234455 on Build GRCh38
rs938949044 Chr.10: 72234044-72234044 on Build GRCh38
rs960150748 Chr.10: 72234302-72234302 on Build GRCh38
rs977493199 Chr.10: 72234011-72234011 on Build GRCh38
rs551001968 Chr.10: 72234353-72234353 on Build GRCh38
rs927451834 Chr.10: 72234309-72234309 on Build GRCh38
rs60558719 Chr.10: 72234117-72234117 on Build GRCh38
rs1007894724 Chr.10: 72234135-72234135 on Build GRCh38
rs968848420 Chr.10: 72234287-72234287 on Build GRCh38
rs1047925849 Chr.10: 72234067-72234067 on Build GRCh38
rs532807368 Chr.10: 72234309-72234310 on Build GRCh38
rs1006445896 Chr.10: 72234077-72234077 on Build GRCh38
rs1051972245 Chr.10: 72234372-72234373 on Build GRCh38
rs4293052 Chr.10: 72234310-72234310 on Build GRCh38
rs928928955 Chr.10: 72234377-72234377 on Build GRCh38
rs58974704 Chr.10: 72234008-72234008 on Build GRCh38
rs980648315 Chr.10: 72234288-72234288 on Build GRCh38
rs763795400 Chr.10: 72234068-72234068 on Build GRCh38
rs573088922 Chr.10: 72233980-72233980 on Build GRCh38
rs977440757 Chr.10: 72234268-72234268 on Build GRCh38
rs1011513663 Chr.10: 72234207-72234207 on Build GRCh38
rs956018689 Chr.10: 72234225-72234225 on Build GRCh38
rs1037970948 Chr.10: 72234328-72234328 on Build GRCh38
rs1011878578 Chr.10: 72234452-72234452 on Build GRCh38
rs908928946 Chr.10: 72234402-72234402 on Build GRCh38
rs1038827761 Chr.10: 72234427-72234427 on Build GRCh38
rs967846291 Chr.10: 72234010-72234010 on Build GRCh38
rs898013170 Chr.10: 72234071-72234071 on Build GRCh38
rs990381200 Chr.10: 72234008-72234009 on Build GRCh38
rs927526452 Chr.10: 72234013-72234013 on Build GRCh38
rs993889474 Chr.10: 72234101-72234101 on Build GRCh38
rs113517546 Chr.10: 72234008-72234008 on Build GRCh38
rs1030293543 Chr.10: 72234197-72234197 on Build GRCh38

View Full Product Details