Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73139231-73139733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECD
Entrez Gene ID
11319
Gene Name
ecdysoneless cell cycle regulator
Gene Aliases
GCR2, HSGT1, SGT1
UniGene
Hs.631822
Gene Chromosome Location
Chr.10: 73134524-73168095 on Build GRCh38
Transcripts
NM_001135752.1 NM_001135753.1 NM_007265.2 NR_024203.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5816672 c.1334-2A>T p.? Chr.10: 73139497-73139497 on Build GRCh38
4528033 c.1507G>A p.A503T Chr.10: 73139322-73139322 on Build GRCh38
2161754 c.1336C>T p.Q446* Chr.10: 73139394-73139394 on Build GRCh38
2161761 c.1278A>G p.I426M Chr.10: 73139686-73139686 on Build GRCh38
685392 c.1401C>T p.H467H Chr.10: 73139329-73139329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139627042 Chr.10: 73139653-73139653 on Build GRCh38
rs755484131 Chr.10: 73139630-73139630 on Build GRCh38
rs766720035 Chr.10: 73139617-73139617 on Build GRCh38
rs141641524 Chr.10: 73139467-73139467 on Build GRCh38
rs776899584 Chr.10: 73139590-73139590 on Build GRCh38
rs768185082 Chr.10: 73139555-73139555 on Build GRCh38
rs761936712 Chr.10: 73139606-73139606 on Build GRCh38
rs773708177 Chr.10: 73139424-73139424 on Build GRCh38
rs1051735819 Chr.10: 73139548-73139548 on Build GRCh38
rs34865820 Chr.10: 73139410-73139410 on Build GRCh38
rs369177164 Chr.10: 73139694-73139694 on Build GRCh38
rs777418264 Chr.10: 73139648-73139648 on Build GRCh38
rs375783049 Chr.10: 73139519-73139519 on Build GRCh38
rs567071975 Chr.10: 73139676-73139676 on Build GRCh38
rs3736517 Chr.10: 73139353-73139353 on Build GRCh38
rs764141288 Chr.10: 73139521-73139522 on Build GRCh38
rs770576759 Chr.10: 73139528-73139528 on Build GRCh38
rs971224908 Chr.10: 73139707-73139707 on Build GRCh38
rs945633938 Chr.10: 73139560-73139560 on Build GRCh38
rs775500009 Chr.10: 73139677-73139677 on Build GRCh38
rs1007524995 Chr.10: 73139428-73139428 on Build GRCh38
rs770616157 Chr.10: 73139658-73139658 on Build GRCh38
rs752847696 Chr.10: 73139296-73139296 on Build GRCh38
rs7083418 Chr.10: 73139637-73139637 on Build GRCh38
rs763460587 Chr.10: 73139374-73139378 on Build GRCh38
rs780621697 Chr.10: 73139460-73139460 on Build GRCh38
rs774599264 Chr.10: 73139702-73139702 on Build GRCh38
rs745653828 Chr.10: 73139675-73139675 on Build GRCh38
rs149336255 Chr.10: 73139698-73139698 on Build GRCh38
rs746259542 Chr.10: 73139320-73139320 on Build GRCh38
rs767991255 Chr.10: 73139392-73139392 on Build GRCh38
rs7083532 Chr.10: 73139670-73139670 on Build GRCh38
rs772984243 Chr.10: 73139291-73139291 on Build GRCh38
rs772016482 Chr.10: 73139546-73139546 on Build GRCh38
rs374769397 Chr.10: 73139666-73139666 on Build GRCh38
rs777368436 Chr.10: 73139513-73139513 on Build GRCh38
rs773420599 Chr.10: 73139687-73139687 on Build GRCh38
rs765072176 Chr.10: 73139289-73139289 on Build GRCh38
rs373470186 Chr.10: 73139293-73139293 on Build GRCh38
rs370377456 Chr.10: 73139537-73139537 on Build GRCh38
rs76482336 Chr.10: 73139586-73139586 on Build GRCh38
rs770936832 Chr.10: 73139421-73139421 on Build GRCh38
rs200280694 Chr.10: 73139608-73139608 on Build GRCh38
rs757436416 Chr.10: 73139585-73139586 on Build GRCh38
rs779489054 Chr.10: 73139343-73139343 on Build GRCh38
rs762168022 Chr.10: 73139442-73139442 on Build GRCh38
rs781685876 Chr.10: 73139631-73139631 on Build GRCh38
rs756112205 Chr.10: 73139302-73139302 on Build GRCh38
rs75751868 Chr.10: 73139587-73139587 on Build GRCh38
rs748249410 Chr.10: 73139681-73139681 on Build GRCh38
rs546818557 Chr.10: 73139586-73139586 on Build GRCh38
rs958559470 Chr.10: 73139316-73139316 on Build GRCh38
rs759024075 Chr.10: 73139361-73139361 on Build GRCh38
rs35652947 Chr.10: 73139422-73139422 on Build GRCh38
rs755558331 Chr.10: 73139640-73139640 on Build GRCh38
rs574287013 Chr.10: 73139400-73139400 on Build GRCh38
rs764140915 Chr.10: 73139432-73139432 on Build GRCh38
rs749091074 Chr.10: 73139526-73139526 on Build GRCh38
rs1036705005 Chr.10: 73139367-73139367 on Build GRCh38
rs148267944 Chr.10: 73139710-73139710 on Build GRCh38
rs750635055 Chr.10: 73139695-73139697 on Build GRCh38
rs746532921 Chr.10: 73139387-73139387 on Build GRCh38
rs796987845 Chr.10: 73139586-73139587 on Build GRCh38
rs778614213 Chr.10: 73139672-73139672 on Build GRCh38
rs763253432 Chr.10: 73139616-73139616 on Build GRCh38
rs1036822310 Chr.10: 73139662-73139662 on Build GRCh38
rs759459596 Chr.10: 73139538-73139538 on Build GRCh38
rs150500888 Chr.10: 73139474-73139474 on Build GRCh38
rs758769408 Chr.10: 73139457-73139457 on Build GRCh38
rs776189860 Chr.10: 73139395-73139395 on Build GRCh38
rs372478574 Chr.10: 73139263-73139263 on Build GRCh38
rs200978167 Chr.10: 73139462-73139462 on Build GRCh38
rs372951554 Chr.10: 73139620-73139620 on Build GRCh38
rs780604680 Chr.10: 73139484-73139484 on Build GRCh38
rs750615935 Chr.10: 73139615-73139615 on Build GRCh38
rs747630787 Chr.10: 73139485-73139485 on Build GRCh38
rs193279143 Chr.10: 73139568-73139568 on Build GRCh38
rs563153737 Chr.10: 73139507-73139507 on Build GRCh38
rs747860932 Chr.10: 73139397-73139397 on Build GRCh38
rs376462298 Chr.10: 73139413-73139413 on Build GRCh38
rs765534407 Chr.10: 73139452-73139452 on Build GRCh38
rs143345132 Chr.10: 73139310-73139310 on Build GRCh38
rs552703905 Chr.10: 73139586-73139587 on Build GRCh38
rs3736518 Chr.10: 73139376-73139376 on Build GRCh38
rs753949042 Chr.10: 73139438-73139438 on Build GRCh38

View Full Product Details