Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73230039-73230581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM149B1
Entrez Gene ID
317662
Gene Name
family with sequence similarity 149 member B1
Gene Aliases
KIAA0974
UniGene
Hs.408577
Gene Chromosome Location
Chr.10: 73168090-73242181 on Build GRCh38
Transcripts
NM_173348.1 XM_005269744.2 XM_005269745.2 XM_005269747.3 XM_011539737.2 XM_017016164.1 XM_017016165.1 XM_017016166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3935157 c.1109C>T p.S370F Chr.10: 73230507-73230507 on Build GRCh38
920260 c.1100G>T p.R367I Chr.10: 73230498-73230498 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs541467792 Chr.10: 73230520-73230520 on Build GRCh38
rs770467331 Chr.10: 73230159-73230159 on Build GRCh38
rs924112337 Chr.10: 73230527-73230527 on Build GRCh38
rs762661001 Chr.10: 73230417-73230417 on Build GRCh38
rs777058485 Chr.10: 73230139-73230139 on Build GRCh38
rs746991722 Chr.10: 73230083-73230083 on Build GRCh38
rs920863487 Chr.10: 73230145-73230145 on Build GRCh38
rs554018296 Chr.10: 73230466-73230466 on Build GRCh38
rs770670664 Chr.10: 73230426-73230426 on Build GRCh38
rs370741909 Chr.10: 73230535-73230535 on Build GRCh38
rs761296673 Chr.10: 73230460-73230460 on Build GRCh38
rs1042202954 Chr.10: 73230269-73230269 on Build GRCh38
rs769582146 Chr.10: 73230377-73230377 on Build GRCh38
rs750681131 Chr.10: 73230526-73230526 on Build GRCh38
rs773054125 Chr.10: 73230390-73230390 on Build GRCh38
rs753919394 Chr.10: 73230491-73230491 on Build GRCh38
rs929053927 Chr.10: 73230070-73230070 on Build GRCh38
rs12570053 Chr.10: 73230369-73230369 on Build GRCh38
rs117551545 Chr.10: 73230106-73230106 on Build GRCh38
rs751907996 Chr.10: 73230556-73230556 on Build GRCh38
rs376112730 Chr.10: 73230347-73230347 on Build GRCh38
rs895121685 Chr.10: 73230554-73230554 on Build GRCh38
rs773317134 Chr.10: 73230269-73230269 on Build GRCh38
rs142562964 Chr.10: 73230536-73230536 on Build GRCh38
rs944720767 Chr.10: 73230515-73230515 on Build GRCh38
rs570895031 Chr.10: 73230249-73230249 on Build GRCh38
rs187928157 Chr.10: 73230092-73230092 on Build GRCh38
rs975381180 Chr.10: 73230492-73230492 on Build GRCh38
rs200156581 Chr.10: 73230485-73230485 on Build GRCh38
rs973622534 Chr.10: 73230131-73230131 on Build GRCh38
rs987227775 Chr.10: 73230422-73230422 on Build GRCh38
rs893963210 Chr.10: 73230162-73230162 on Build GRCh38
rs762443646 Chr.10: 73230130-73230132 on Build GRCh38
rs117484669 Chr.10: 73230552-73230552 on Build GRCh38
rs747590949 Chr.10: 73230375-73230375 on Build GRCh38
rs762715318 Chr.10: 73230278-73230278 on Build GRCh38
rs1042437742 Chr.10: 73230203-73230204 on Build GRCh38
rs893753426 Chr.10: 73230337-73230337 on Build GRCh38
rs535762300 Chr.10: 73230464-73230464 on Build GRCh38
rs766614712 Chr.10: 73230389-73230389 on Build GRCh38
rs746363058 Chr.10: 73230152-73230152 on Build GRCh38
rs986386413 Chr.10: 73230509-73230509 on Build GRCh38
rs902317177 Chr.10: 73230277-73230277 on Build GRCh38
rs946928920 Chr.10: 73230168-73230168 on Build GRCh38
rs866426573 Chr.10: 73230489-73230489 on Build GRCh38
rs538333879 Chr.10: 73230253-73230253 on Build GRCh38
rs182471816 Chr.10: 73230514-73230514 on Build GRCh38
rs528386107 Chr.10: 73230362-73230362 on Build GRCh38
rs890527446 Chr.10: 73230240-73230240 on Build GRCh38
rs911552577 Chr.10: 73230465-73230465 on Build GRCh38
rs774172961 Chr.10: 73230457-73230457 on Build GRCh38
rs1053612718 Chr.10: 73230309-73230309 on Build GRCh38
rs370009319 Chr.10: 73230088-73230088 on Build GRCh38
rs767041717 Chr.10: 73230477-73230477 on Build GRCh38
rs1026171003 Chr.10: 73230416-73230416 on Build GRCh38
rs761340545 Chr.10: 73230065-73230068 on Build GRCh38
rs535305882 Chr.10: 73230099-73230100 on Build GRCh38
rs1023329738 Chr.10: 73230075-73230075 on Build GRCh38
rs895184473 Chr.10: 73230297-73230297 on Build GRCh38
rs1034942618 Chr.10: 73230279-73230279 on Build GRCh38
rs775955555 Chr.10: 73230244-73230244 on Build GRCh38
rs189809341 Chr.10: 73230110-73230110 on Build GRCh38

View Full Product Details