Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73772178-73772678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT11
Entrez Gene ID
170384
Gene Name
fucosyltransferase 11
Gene Aliases
-
UniGene
Hs.588854
Gene Chromosome Location
Chr.10: 73772175-73780251 on Build GRCh38
Transcripts
NM_001284194.1 NM_173540.2 XM_006717656.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5192995 c.176C>A p.T59K Chr.10: 73772509-73772509 on Build GRCh38
6667863 c.225G>T p.L75F Chr.10: 73772558-73772558 on Build GRCh38
1207659 c.331G>T p.A111S Chr.10: 73772664-73772664 on Build GRCh38
4491412 c.37C>T p.L13L Chr.10: 73772370-73772370 on Build GRCh38
1603717 c.132C>T p.G44G Chr.10: 73772465-73772465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775846461 Chr.10: 73772497-73772497 on Build GRCh38
rs912544456 Chr.10: 73772389-73772389 on Build GRCh38
rs759791115 Chr.10: 73772381-73772382 on Build GRCh38
rs752384561 Chr.10: 73772641-73772641 on Build GRCh38
rs774510919 Chr.10: 73772414-73772414 on Build GRCh38
rs1029878183 Chr.10: 73772479-73772479 on Build GRCh38
rs751161820 Chr.10: 73772476-73772476 on Build GRCh38
rs747954635 Chr.10: 73772266-73772266 on Build GRCh38
rs776768279 Chr.10: 73772404-73772404 on Build GRCh38
rs1016827827 Chr.10: 73772269-73772269 on Build GRCh38
rs774026945 Chr.10: 73772629-73772629 on Build GRCh38
rs768669478 Chr.10: 73772352-73772352 on Build GRCh38
rs997119920 Chr.10: 73772277-73772277 on Build GRCh38
rs778591459 Chr.10: 73772341-73772341 on Build GRCh38
rs573139041 Chr.10: 73772324-73772324 on Build GRCh38
rs760704696 Chr.10: 73772246-73772246 on Build GRCh38
rs929512962 Chr.10: 73772633-73772633 on Build GRCh38
rs929350435 Chr.10: 73772413-73772413 on Build GRCh38
rs1020741605 Chr.10: 73772526-73772526 on Build GRCh38
rs759490259 Chr.10: 73772422-73772422 on Build GRCh38
rs749980928 Chr.10: 73772472-73772472 on Build GRCh38
rs376925741 Chr.10: 73772475-73772475 on Build GRCh38
rs144665547 Chr.10: 73772394-73772394 on Build GRCh38
rs1040302036 Chr.10: 73772260-73772260 on Build GRCh38
rs775337085 Chr.10: 73772343-73772343 on Build GRCh38
rs748095383 Chr.10: 73772407-73772407 on Build GRCh38
rs926755592 Chr.10: 73772219-73772219 on Build GRCh38
rs566605385 Chr.10: 73772493-73772493 on Build GRCh38
rs754890483 Chr.10: 73772477-73772477 on Build GRCh38
rs748797775 Chr.10: 73772339-73772339 on Build GRCh38
rs1013087310 Chr.10: 73772468-73772468 on Build GRCh38
rs1009708454 Chr.10: 73772518-73772518 on Build GRCh38
rs770046212 Chr.10: 73772615-73772615 on Build GRCh38
rs765685814 Chr.10: 73772465-73772465 on Build GRCh38
rs763378577 Chr.10: 73772642-73772642 on Build GRCh38
rs758094835 Chr.10: 73772392-73772392 on Build GRCh38
rs763140146 Chr.10: 73772353-73772353 on Build GRCh38
rs766857859 Chr.10: 73772625-73772625 on Build GRCh38
rs753135999 Chr.10: 73772359-73772359 on Build GRCh38
rs754698293 Chr.10: 73772575-73772575 on Build GRCh38
rs777241876 Chr.10: 73772271-73772271 on Build GRCh38
rs752004522 Chr.10: 73772238-73772238 on Build GRCh38
rs552207359 Chr.10: 73772650-73772650 on Build GRCh38
rs895454030 Chr.10: 73772464-73772464 on Build GRCh38
rs201902076 Chr.10: 73772471-73772471 on Build GRCh38
rs777004460 Chr.10: 73772520-73772520 on Build GRCh38
rs150554210 Chr.10: 73772308-73772309 on Build GRCh38
rs756038975 Chr.10: 73772652-73772652 on Build GRCh38
rs373328220 Chr.10: 73772460-73772460 on Build GRCh38
rs765008856 Chr.10: 73772438-73772446 on Build GRCh38
rs758675651 Chr.10: 73772213-73772213 on Build GRCh38
rs761158918 Chr.10: 73772433-73772433 on Build GRCh38
rs141139329 Chr.10: 73772454-73772454 on Build GRCh38
rs775358428 Chr.10: 73772306-73772306 on Build GRCh38
rs771182717 Chr.10: 73772411-73772411 on Build GRCh38
rs757166506 Chr.10: 73772661-73772661 on Build GRCh38
rs112283826 Chr.10: 73772259-73772259 on Build GRCh38
rs754034769 Chr.10: 73772340-73772341 on Build GRCh38
rs770596883 Chr.10: 73772273-73772273 on Build GRCh38
rs1006205780 Chr.10: 73772448-73772448 on Build GRCh38
rs767286729 Chr.10: 73772632-73772632 on Build GRCh38
rs16930888 Chr.10: 73772467-73772467 on Build GRCh38
rs777535393 Chr.10: 73772512-73772512 on Build GRCh38
rs755910269 Chr.10: 73772582-73772582 on Build GRCh38
rs767779747 Chr.10: 73772423-73772423 on Build GRCh38
rs973264220 Chr.10: 73772243-73772243 on Build GRCh38
rs778213051 Chr.10: 73772338-73772338 on Build GRCh38
rs1020049172 Chr.10: 73772617-73772617 on Build GRCh38
rs947633313 Chr.10: 73772268-73772268 on Build GRCh38
rs947499308 Chr.10: 73772416-73772416 on Build GRCh38
rs763087190 Chr.10: 73772317-73772317 on Build GRCh38
rs919934442 Chr.10: 73772245-73772245 on Build GRCh38
rs773838671 Chr.10: 73772618-73772618 on Build GRCh38
rs993598864 Chr.10: 73772249-73772249 on Build GRCh38
rs762585364 Chr.10: 73772551-73772551 on Build GRCh38
rs369598879 Chr.10: 73772382-73772382 on Build GRCh38
rs776768898 Chr.10: 73772606-73772606 on Build GRCh38
rs1012645014 Chr.10: 73772307-73772307 on Build GRCh38
rs959814627 Chr.10: 73772208-73772208 on Build GRCh38
rs771317724 Chr.10: 73772276-73772276 on Build GRCh38
rs896601025 Chr.10: 73772241-73772241 on Build GRCh38
rs765738963 Chr.10: 73772535-73772535 on Build GRCh38
rs918100226 Chr.10: 73772408-73772408 on Build GRCh38
rs547952525 Chr.10: 73772487-73772487 on Build GRCh38
rs1044015389 Chr.10: 73772203-73772203 on Build GRCh38
rs527288970 Chr.10: 73772609-73772609 on Build GRCh38
rs763339729 Chr.10: 73772621-73772621 on Build GRCh38
rs924891714 Chr.10: 73772453-73772453 on Build GRCh38
rs561462538 Chr.10: 73772318-73772318 on Build GRCh38
rs540662503 Chr.10: 73772325-73772325 on Build GRCh38
rs979663485 Chr.10: 73772366-73772366 on Build GRCh38
rs750842372 Chr.10: 73772539-73772539 on Build GRCh38
rs756823985 Chr.10: 73772365-73772365 on Build GRCh38
rs762126054 Chr.10: 73772534-73772534 on Build GRCh38
rs565233785 Chr.10: 73772346-73772346 on Build GRCh38
rs747043863 Chr.10: 73772605-73772605 on Build GRCh38
rs535161593 Chr.10: 73772224-73772224 on Build GRCh38
rs17853514 Chr.10: 73772484-73772484 on Build GRCh38
rs542777283 Chr.10: 73772308-73772308 on Build GRCh38
rs998967500 Chr.10: 73772285-73772285 on Build GRCh38
rs749176743 Chr.10: 73772591-73772591 on Build GRCh38
rs756695386 Chr.10: 73772340-73772340 on Build GRCh38
rs769879936 Chr.10: 73772485-73772485 on Build GRCh38
rs921389364 Chr.10: 73772649-73772649 on Build GRCh38
rs979130240 Chr.10: 73772572-73772572 on Build GRCh38
rs773801572 Chr.10: 73772245-73772245 on Build GRCh38
rs750557083 Chr.10: 73772316-73772316 on Build GRCh38
rs376115376 Chr.10: 73772323-73772323 on Build GRCh38
rs372603690 Chr.10: 73772530-73772530 on Build GRCh38
rs377084510 Chr.10: 73772579-73772579 on Build GRCh38
rs1029413122 Chr.10: 73772279-73772279 on Build GRCh38
rs763763816 Chr.10: 73772654-73772654 on Build GRCh38
rs760633525 Chr.10: 73772309-73772309 on Build GRCh38
rs768581295 Chr.10: 73772242-73772242 on Build GRCh38
rs751267685 Chr.10: 73772565-73772565 on Build GRCh38
rs1022766584 Chr.10: 73772313-73772313 on Build GRCh38
rs774348364 Chr.10: 73772251-73772251 on Build GRCh38
rs747899391 Chr.10: 73772483-73772483 on Build GRCh38
rs371592507 Chr.10: 73772515-73772515 on Build GRCh38
rs759337165 Chr.10: 73772296-73772296 on Build GRCh38
rs562756247 Chr.10: 73772370-73772370 on Build GRCh38
rs774751416 Chr.10: 73772354-73772354 on Build GRCh38
rs754380315 Chr.10: 73772458-73772458 on Build GRCh38
rs967694416 Chr.10: 73772555-73772555 on Build GRCh38
rs912669939 Chr.10: 73772256-73772256 on Build GRCh38
rs780949404 Chr.10: 73772577-73772577 on Build GRCh38
rs754326953 Chr.10: 73772364-73772364 on Build GRCh38
rs1035354948 Chr.10: 73772294-73772294 on Build GRCh38
rs1043298198 Chr.10: 73772274-73772274 on Build GRCh38
rs768514766 Chr.10: 73772399-73772399 on Build GRCh38
rs140951394 Chr.10: 73772361-73772361 on Build GRCh38
rs777824644 Chr.10: 73772586-73772586 on Build GRCh38
rs771885234 Chr.10: 73772304-73772304 on Build GRCh38
rs779973216 Chr.10: 73772603-73772603 on Build GRCh38
rs137937533 Chr.10: 73772356-73772356 on Build GRCh38
rs370513558 Chr.10: 73772595-73772595 on Build GRCh38
rs772030306 Chr.10: 73772495-73772495 on Build GRCh38
rs959856238 Chr.10: 73772584-73772584 on Build GRCh38
rs753860541 Chr.10: 73772660-73772660 on Build GRCh38
rs781109251 Chr.10: 73772481-73772481 on Build GRCh38
rs771805340 Chr.10: 73772342-73772342 on Build GRCh38
rs778366352 Chr.10: 73772376-73772376 on Build GRCh38
rs1043245694 Chr.10: 73772447-73772447 on Build GRCh38
rs750718104 Chr.10: 73772205-73772205 on Build GRCh38
rs756666315 Chr.10: 73772336-73772336 on Build GRCh38
rs775517466 Chr.10: 73772430-73772430 on Build GRCh38
rs148202686 Chr.10: 73772396-73772396 on Build GRCh38

View Full Product Details