Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73773057-73773599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT11
Entrez Gene ID
170384
Gene Name
fucosyltransferase 11
Gene Aliases
-
UniGene
Hs.588854
Gene Chromosome Location
Chr.10: 73772175-73780251 on Build GRCh38
Transcripts
NM_001284194.1 NM_173540.2 XM_006717656.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6551055 c.709G>T p.V237L Chr.10: 73773190-73773190 on Build GRCh38
7423320 c.1000G>T p.A334S Chr.10: 73773481-73773481 on Build GRCh38
4573822 c.854G>A p.C285Y Chr.10: 73773335-73773335 on Build GRCh38
1207660 c.860A>G p.D287G Chr.10: 73773341-73773341 on Build GRCh38
1627611 c.818G>T p.R273L Chr.10: 73773299-73773299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761191382 Chr.10: 73773092-73773092 on Build GRCh38
rs922070686 Chr.10: 73773256-73773256 on Build GRCh38
rs145493843 Chr.10: 73773331-73773331 on Build GRCh38
rs773541592 Chr.10: 73773091-73773106 on Build GRCh38
rs772632063 Chr.10: 73773346-73773346 on Build GRCh38
rs748393861 Chr.10: 73773090-73773092 on Build GRCh38
rs751248050 Chr.10: 73773365-73773365 on Build GRCh38
rs748477561 Chr.10: 73773548-73773548 on Build GRCh38
rs145929404 Chr.10: 73773091-73773122 on Build GRCh38
rs750140528 Chr.10: 73773238-73773238 on Build GRCh38
rs756662019 Chr.10: 73773149-73773149 on Build GRCh38
rs186668800 Chr.10: 73773144-73773144 on Build GRCh38
rs758514536 Chr.10: 73773520-73773520 on Build GRCh38
rs749786366 Chr.10: 73773575-73773575 on Build GRCh38
rs140577849 Chr.10: 73773517-73773517 on Build GRCh38
rs766147046 Chr.10: 73773359-73773359 on Build GRCh38
rs1003680084 Chr.10: 73773113-73773144 on Build GRCh38
rs765986547 Chr.10: 73773452-73773452 on Build GRCh38
rs775862106 Chr.10: 73773225-73773225 on Build GRCh38
rs5017445 Chr.10: 73773128-73773128 on Build GRCh38
rs751592475 Chr.10: 73773532-73773532 on Build GRCh38
rs762610347 Chr.10: 73773437-73773437 on Build GRCh38
rs555255735 Chr.10: 73773107-73773122 on Build GRCh38
rs373281050 Chr.10: 73773542-73773542 on Build GRCh38
rs770185021 Chr.10: 73773560-73773560 on Build GRCh38
rs375283537 Chr.10: 73773467-73773467 on Build GRCh38
rs769057290 Chr.10: 73773410-73773410 on Build GRCh38
rs925089966 Chr.10: 73773360-73773360 on Build GRCh38
rs764002062 Chr.10: 73773475-73773476 on Build GRCh38
rs894932729 Chr.10: 73773382-73773382 on Build GRCh38
rs372216492 Chr.10: 73773300-73773300 on Build GRCh38
rs763995269 Chr.10: 73773492-73773492 on Build GRCh38
rs776884425 Chr.10: 73773418-73773418 on Build GRCh38
rs1014729208 Chr.10: 73773540-73773540 on Build GRCh38
rs757403129 Chr.10: 73773279-73773279 on Build GRCh38
rs375782601 Chr.10: 73773483-73773483 on Build GRCh38
rs1029490294 Chr.10: 73773554-73773554 on Build GRCh38
rs1005068742 Chr.10: 73773516-73773516 on Build GRCh38
rs762614735 Chr.10: 73773447-73773447 on Build GRCh38
rs759737803 Chr.10: 73773228-73773229 on Build GRCh38
rs746755098 Chr.10: 73773180-73773180 on Build GRCh38
rs1046032261 Chr.10: 73773461-73773461 on Build GRCh38
rs777698056 Chr.10: 73773398-73773398 on Build GRCh38
rs199683305 Chr.10: 73773106-73773106 on Build GRCh38
rs778213770 Chr.10: 73773566-73773566 on Build GRCh38
rs765314050 Chr.10: 73773309-73773309 on Build GRCh38
rs770210709 Chr.10: 73773429-73773429 on Build GRCh38
rs1042344567 Chr.10: 73773330-73773330 on Build GRCh38
rs753248700 Chr.10: 73773081-73773081 on Build GRCh38
rs772229925 Chr.10: 73773220-73773220 on Build GRCh38
rs781411981 Chr.10: 73773547-73773547 on Build GRCh38
rs542457778 Chr.10: 73773182-73773182 on Build GRCh38
rs767394724 Chr.10: 73773381-73773381 on Build GRCh38
rs367955675 Chr.10: 73773234-73773234 on Build GRCh38
rs776754893 Chr.10: 73773173-73773174 on Build GRCh38
rs755878847 Chr.10: 73773486-73773486 on Build GRCh38
rs753349249 Chr.10: 73773145-73773145 on Build GRCh38
rs770907888 Chr.10: 73773200-73773200 on Build GRCh38
rs778878172 Chr.10: 73773287-73773287 on Build GRCh38
rs781339977 Chr.10: 73773270-73773270 on Build GRCh38
rs750041284 Chr.10: 73773158-73773158 on Build GRCh38
rs755247784 Chr.10: 73773339-73773339 on Build GRCh38
rs145993159 Chr.10: 73773237-73773237 on Build GRCh38
rs544522646 Chr.10: 73773125-73773125 on Build GRCh38
rs765727957 Chr.10: 73773340-73773340 on Build GRCh38
rs369235771 Chr.10: 73773484-73773484 on Build GRCh38
rs745919565 Chr.10: 73773291-73773291 on Build GRCh38
rs111981276 Chr.10: 73773156-73773156 on Build GRCh38
rs368549080 Chr.10: 73773219-73773219 on Build GRCh38
rs869156028 Chr.10: 73773090-73773122 on Build GRCh38
rs759653463 Chr.10: 73773210-73773210 on Build GRCh38
rs755225852 Chr.10: 73773535-73773535 on Build GRCh38
rs780085558 Chr.10: 73773523-73773523 on Build GRCh38
rs770964525 Chr.10: 73773094-73773106 on Build GRCh38
rs1031793988 Chr.10: 73773124-73773124 on Build GRCh38
rs573097374 Chr.10: 73773337-73773337 on Build GRCh38
rs371450426 Chr.10: 73773229-73773229 on Build GRCh38
rs753809265 Chr.10: 73773493-73773493 on Build GRCh38
rs752485865 Chr.10: 73773390-73773390 on Build GRCh38
rs758253957 Chr.10: 73773171-73773171 on Build GRCh38
rs996336698 Chr.10: 73773543-73773543 on Build GRCh38
rs749403827 Chr.10: 73773197-73773197 on Build GRCh38
rs761217754 Chr.10: 73773090-73773091 on Build GRCh38
rs147269045 Chr.10: 73773333-73773333 on Build GRCh38
rs12766368 Chr.10: 73773112-73773112 on Build GRCh38
rs748001224 Chr.10: 73773278-73773278 on Build GRCh38
rs369685993 Chr.10: 73773318-73773318 on Build GRCh38
rs771511865 Chr.10: 73773576-73773576 on Build GRCh38
rs954868203 Chr.10: 73773268-73773268 on Build GRCh38
rs200368257 Chr.10: 73773302-73773302 on Build GRCh38
rs570485259 Chr.10: 73773440-73773440 on Build GRCh38
rs779816526 Chr.10: 73773248-73773248 on Build GRCh38
rs954781184 Chr.10: 73773571-73773571 on Build GRCh38
rs151328037 Chr.10: 73773393-73773393 on Build GRCh38
rs781211696 Chr.10: 73773196-73773196 on Build GRCh38
rs751513911 Chr.10: 73773505-73773505 on Build GRCh38
rs751984942 Chr.10: 73773207-73773207 on Build GRCh38
rs745947432 Chr.10: 73773400-73773400 on Build GRCh38
rs754766038 Chr.10: 73773264-73773264 on Build GRCh38
rs780321328 Chr.10: 73773407-73773407 on Build GRCh38
rs757332561 Chr.10: 73773514-73773516 on Build GRCh38
rs76859274 Chr.10: 73773329-73773329 on Build GRCh38
rs750110872 Chr.10: 73773401-73773401 on Build GRCh38
rs147181018 Chr.10: 73773236-73773236 on Build GRCh38
rs148722357 Chr.10: 73773352-73773352 on Build GRCh38
rs779974034 Chr.10: 73773175-73773175 on Build GRCh38
rs767823444 Chr.10: 73773245-73773245 on Build GRCh38
rs560448773 Chr.10: 73773419-73773419 on Build GRCh38
rs142253283 Chr.10: 73773366-73773366 on Build GRCh38
rs751407484 Chr.10: 73773251-73773251 on Build GRCh38
rs907653213 Chr.10: 73773324-73773324 on Build GRCh38
rs199580732 Chr.10: 73773574-73773574 on Build GRCh38
rs747201298 Chr.10: 73773408-73773408 on Build GRCh38
rs764597275 Chr.10: 73773233-73773233 on Build GRCh38

View Full Product Details