Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 74120371-74120915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3M1
Entrez Gene ID
26985
Gene Name
adaptor related protein complex 3 mu 1 subunit
Gene Aliases
-
UniGene
Hs.500104
Gene Chromosome Location
Chr.10: 74120257-74151085 on Build GRCh38
Transcripts
NM_001320263.1 NM_001320264.1 NM_001320265.1 NM_012095.5 NM_207012.3 NR_135191.1

SNP Detail

SNP ID Chromosome Location
rs926787684 Chr.10: 74120690-74120690 on Build GRCh38
rs10824075 Chr.10: 74120496-74120496 on Build GRCh38
rs938283141 Chr.10: 74120396-74120396 on Build GRCh38
rs1057152919 Chr.10: 74120738-74120738 on Build GRCh38
rs976406141 Chr.10: 74120571-74120571 on Build GRCh38
rs958556703 Chr.10: 74120879-74120879 on Build GRCh38
rs774842891 Chr.10: 74120611-74120611 on Build GRCh38
rs552542751 Chr.10: 74120525-74120525 on Build GRCh38
rs549984308 Chr.10: 74120610-74120610 on Build GRCh38
rs1034195426 Chr.10: 74120538-74120538 on Build GRCh38
rs934265946 Chr.10: 74120678-74120678 on Build GRCh38
rs1054234328 Chr.10: 74120783-74120783 on Build GRCh38
rs558853921 Chr.10: 74120773-74120773 on Build GRCh38
rs547897998 Chr.10: 74120415-74120415 on Build GRCh38
rs1030701191 Chr.10: 74120841-74120841 on Build GRCh38
rs1047456459 Chr.10: 74120777-74120777 on Build GRCh38
rs577108423 Chr.10: 74120810-74120810 on Build GRCh38
rs997432493 Chr.10: 74120826-74120826 on Build GRCh38
rs959443245 Chr.10: 74120562-74120562 on Build GRCh38
rs771508580 Chr.10: 74120467-74120467 on Build GRCh38
rs887498713 Chr.10: 74120802-74120802 on Build GRCh38
rs182701684 Chr.10: 74120737-74120737 on Build GRCh38
rs564244232 Chr.10: 74120625-74120627 on Build GRCh38
rs374345967 Chr.10: 74120663-74120663 on Build GRCh38
rs964632855 Chr.10: 74120564-74120564 on Build GRCh38
rs4746174 Chr.10: 74120594-74120594 on Build GRCh38
rs547139795 Chr.10: 74120457-74120457 on Build GRCh38
rs532870275 Chr.10: 74120616-74120616 on Build GRCh38
rs993200488 Chr.10: 74120445-74120445 on Build GRCh38
rs942407812 Chr.10: 74120746-74120746 on Build GRCh38
rs368382481 Chr.10: 74120809-74120809 on Build GRCh38
rs1020593267 Chr.10: 74120480-74120480 on Build GRCh38
rs12416053 Chr.10: 74120734-74120734 on Build GRCh38
rs763337128 Chr.10: 74120740-74120740 on Build GRCh38
rs979574911 Chr.10: 74120668-74120668 on Build GRCh38
rs6480717 Chr.10: 74120666-74120666 on Build GRCh38
rs570755460 Chr.10: 74120574-74120574 on Build GRCh38
rs566898771 Chr.10: 74120760-74120760 on Build GRCh38
rs907080731 Chr.10: 74120819-74120819 on Build GRCh38
rs1010295038 Chr.10: 74120788-74120788 on Build GRCh38
rs186973198 Chr.10: 74120813-74120813 on Build GRCh38
rs533941154 Chr.10: 74120761-74120761 on Build GRCh38
rs914019117 Chr.10: 74120603-74120603 on Build GRCh38
rs79916214 Chr.10: 74120878-74120878 on Build GRCh38
rs765541839 Chr.10: 74120452-74120453 on Build GRCh38
rs1017866998 Chr.10: 74120521-74120521 on Build GRCh38
rs972775536 Chr.10: 74120609-74120609 on Build GRCh38
rs892903561 Chr.10: 74120787-74120787 on Build GRCh38
rs892829177 Chr.10: 74120447-74120447 on Build GRCh38
rs527598135 Chr.10: 74120515-74120515 on Build GRCh38
rs1056095720 Chr.10: 74120398-74120398 on Build GRCh38
rs772713179 Chr.10: 74120481-74120481 on Build GRCh38
rs955390746 Chr.10: 74120580-74120580 on Build GRCh38
rs896150718 Chr.10: 74120751-74120751 on Build GRCh38
rs1054119962 Chr.10: 74120404-74120404 on Build GRCh38

View Full Product Details