Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 74121609-74122105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3M1
Entrez Gene ID
26985
Gene Name
adaptor related protein complex 3 mu 1 subunit
Gene Aliases
-
UniGene
Hs.500104
Gene Chromosome Location
Chr.10: 74120257-74151085 on Build GRCh38
Transcripts
NM_001320263.1 NM_001320264.1 NM_001320265.1 NM_012095.5 NM_207012.3 NR_135191.1

SNP Detail

SNP ID Chromosome Location
rs574491065 Chr.10: 74121740-74121740 on Build GRCh38
rs1055494912 Chr.10: 74122043-74122043 on Build GRCh38
rs891177559 Chr.10: 74121815-74121815 on Build GRCh38
rs929130433 Chr.10: 74121755-74121759 on Build GRCh38
rs149869240 Chr.10: 74122010-74122010 on Build GRCh38
rs140398345 Chr.10: 74121790-74121790 on Build GRCh38
rs991106337 Chr.10: 74121707-74121707 on Build GRCh38
rs556487985 Chr.10: 74121652-74121652 on Build GRCh38
rs1021850560 Chr.10: 74121931-74121931 on Build GRCh38
rs569256894 Chr.10: 74121689-74121689 on Build GRCh38
rs567197082 Chr.10: 74121952-74121954 on Build GRCh38
rs919179849 Chr.10: 74121749-74121750 on Build GRCh38
rs1014077244 Chr.10: 74122070-74122070 on Build GRCh38
rs974793128 Chr.10: 74121901-74121901 on Build GRCh38
rs998997623 Chr.10: 74121962-74121962 on Build GRCh38
rs1008350909 Chr.10: 74121833-74121833 on Build GRCh38
rs531648433 Chr.10: 74122004-74122004 on Build GRCh38
rs542051107 Chr.10: 74121655-74121655 on Build GRCh38
rs79682050 Chr.10: 74121654-74121654 on Build GRCh38
rs1025062863 Chr.10: 74121805-74121805 on Build GRCh38
rs1010151386 Chr.10: 74121678-74121678 on Build GRCh38
rs766386841 Chr.10: 74121743-74121743 on Build GRCh38
rs950918363 Chr.10: 74121826-74121826 on Build GRCh38
rs941932668 Chr.10: 74121733-74121733 on Build GRCh38
rs561832167 Chr.10: 74122035-74122035 on Build GRCh38
rs913068739 Chr.10: 74121943-74121943 on Build GRCh38
rs780515181 Chr.10: 74121935-74121939 on Build GRCh38
rs909490052 Chr.10: 74121845-74121845 on Build GRCh38
rs945887246 Chr.10: 74121978-74121978 on Build GRCh38
rs963677651 Chr.10: 74121861-74121861 on Build GRCh38
rs6688 Chr.10: 74121918-74121918 on Build GRCh38
rs780733722 Chr.10: 74121902-74121902 on Build GRCh38
rs769564082 Chr.10: 74122026-74122026 on Build GRCh38
rs1043035474 Chr.10: 74121696-74121696 on Build GRCh38
rs1001302361 Chr.10: 74121721-74121721 on Build GRCh38
rs369534073 Chr.10: 74121651-74121651 on Build GRCh38
rs983663430 Chr.10: 74121841-74121841 on Build GRCh38
rs193218236 Chr.10: 74121717-74121717 on Build GRCh38
rs949630575 Chr.10: 74121983-74121983 on Build GRCh38
rs959777917 Chr.10: 74122017-74122017 on Build GRCh38
rs895155152 Chr.10: 74122061-74122061 on Build GRCh38
rs987254081 Chr.10: 74121919-74121919 on Build GRCh38

View Full Product Details