Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 74128820-74129341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3M1
Entrez Gene ID
26985
Gene Name
adaptor related protein complex 3 mu 1 subunit
Gene Aliases
-
UniGene
Hs.500104
Gene Chromosome Location
Chr.10: 74120257-74151085 on Build GRCh38
Transcripts
NM_001320263.1 NM_001320264.1 NM_001320265.1 NM_012095.5 NM_207012.3 NR_135191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7141515 c.773G>T p.R258L Chr.10: 74129138-74129138 on Build GRCh38
4845527 c.764G>C p.G255A Chr.10: 74129147-74129147 on Build GRCh38
5436285 c.700C>T p.H234Y Chr.10: 74129211-74129211 on Build GRCh38
4508354 c.772C>G p.R258G Chr.10: 74129139-74129139 on Build GRCh38
1183142 c.788G>A p.R263H Chr.10: 74129123-74129123 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747410949 Chr.10: 74129124-74129124 on Build GRCh38
rs747813904 Chr.10: 74129266-74129266 on Build GRCh38
rs142765677 Chr.10: 74129190-74129190 on Build GRCh38
rs774278214 Chr.10: 74129290-74129290 on Build GRCh38
rs746701921 Chr.10: 74129237-74129237 on Build GRCh38
rs750369352 Chr.10: 74129058-74129058 on Build GRCh38
rs772735058 Chr.10: 74129278-74129278 on Build GRCh38
rs754090301 Chr.10: 74129198-74129198 on Build GRCh38
rs762388214 Chr.10: 74129287-74129287 on Build GRCh38
rs772505102 Chr.10: 74129116-74129116 on Build GRCh38
rs754368138 Chr.10: 74129189-74129189 on Build GRCh38
rs919132981 Chr.10: 74129086-74129086 on Build GRCh38
rs756513579 Chr.10: 74129072-74129072 on Build GRCh38
rs563021257 Chr.10: 74128956-74128956 on Build GRCh38
rs769333400 Chr.10: 74129144-74129144 on Build GRCh38
rs766136178 Chr.10: 74129069-74129069 on Build GRCh38
rs772321083 Chr.10: 74129298-74129298 on Build GRCh38
rs202134394 Chr.10: 74129138-74129138 on Build GRCh38
rs761919224 Chr.10: 74129084-74129084 on Build GRCh38
rs190136804 Chr.10: 74129139-74129139 on Build GRCh38
rs139128870 Chr.10: 74128943-74128943 on Build GRCh38
rs951046893 Chr.10: 74129016-74129016 on Build GRCh38
rs757366271 Chr.10: 74129092-74129092 on Build GRCh38
rs200411548 Chr.10: 74129273-74129273 on Build GRCh38
rs552482927 Chr.10: 74129274-74129274 on Build GRCh38
rs912540917 Chr.10: 74128859-74128859 on Build GRCh38
rs149538577 Chr.10: 74129312-74129312 on Build GRCh38
rs780501600 Chr.10: 74129104-74129104 on Build GRCh38
rs1000915401 Chr.10: 74128939-74128939 on Build GRCh38
rs758899537 Chr.10: 74129234-74129234 on Build GRCh38
rs902921197 Chr.10: 74128935-74128935 on Build GRCh38
rs73282407 Chr.10: 74129221-74129221 on Build GRCh38
rs1025252429 Chr.10: 74128999-74128999 on Build GRCh38
rs781728113 Chr.10: 74129265-74129265 on Build GRCh38
rs374572738 Chr.10: 74129199-74129199 on Build GRCh38
rs998982691 Chr.10: 74129310-74129310 on Build GRCh38
rs747309014 Chr.10: 74129244-74129244 on Build GRCh38
rs114441758 Chr.10: 74128873-74128873 on Build GRCh38
rs756286523 Chr.10: 74129077-74129077 on Build GRCh38
rs766332747 Chr.10: 74129184-74129184 on Build GRCh38
rs762912948 Chr.10: 74129174-74129174 on Build GRCh38
rs184938063 Chr.10: 74128955-74128955 on Build GRCh38
rs772642400 Chr.10: 74129152-74129152 on Build GRCh38
rs761437710 Chr.10: 74129140-74129140 on Build GRCh38
rs777990129 Chr.10: 74129078-74129078 on Build GRCh38
rs11000881 Chr.10: 74129197-74129197 on Build GRCh38
rs759185186 Chr.10: 74129238-74129238 on Build GRCh38
rs987726011 Chr.10: 74129209-74129209 on Build GRCh38
rs769136501 Chr.10: 74129256-74129256 on Build GRCh38
rs770794873 Chr.10: 74129289-74129289 on Build GRCh38
rs557849911 Chr.10: 74129049-74129049 on Build GRCh38
rs1042803832 Chr.10: 74128902-74128902 on Build GRCh38
rs758325165 Chr.10: 74129059-74129059 on Build GRCh38
rs1012127912 Chr.10: 74128857-74128857 on Build GRCh38
rs779029562 Chr.10: 74129206-74129206 on Build GRCh38
rs903898752 Chr.10: 74128914-74128914 on Build GRCh38
rs546157956 Chr.10: 74128863-74128863 on Build GRCh38
rs746377295 Chr.10: 74129113-74129113 on Build GRCh38
rs780534913 Chr.10: 74129123-74129123 on Build GRCh38
rs1012605436 Chr.10: 74128990-74128990 on Build GRCh38
rs1023277844 Chr.10: 74129034-74129034 on Build GRCh38
rs1013770688 Chr.10: 74128967-74128967 on Build GRCh38
rs963355638 Chr.10: 74129047-74129047 on Build GRCh38

View Full Product Details