Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 74133949-74134455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3M1
Entrez Gene ID
26985
Gene Name
adaptor related protein complex 3 mu 1 subunit
Gene Aliases
-
UniGene
Hs.500104
Gene Chromosome Location
Chr.10: 74120257-74151085 on Build GRCh38
Transcripts
NM_001320263.1 NM_001320264.1 NM_001320265.1 NM_012095.5 NM_207012.3 NR_135191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6292574 c.505C>T p.R169W Chr.10: 74134105-74134105 on Build GRCh38
1135057 c.464A>T p.D155V Chr.10: 74134146-74134146 on Build GRCh38
1193281 c.501G>T p.W167C Chr.10: 74134109-74134109 on Build GRCh38
301684 c.478G>T p.G160W Chr.10: 74134132-74134132 on Build GRCh38
4496533 c.477C>A p.T159T Chr.10: 74134133-74134133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913615355 Chr.10: 74134051-74134051 on Build GRCh38
rs755498612 Chr.10: 74133990-74133990 on Build GRCh38
rs145365519 Chr.10: 74134132-74134132 on Build GRCh38
rs955624627 Chr.10: 74134279-74134279 on Build GRCh38
rs748928879 Chr.10: 74134016-74134016 on Build GRCh38
rs746895566 Chr.10: 74134150-74134150 on Build GRCh38
rs914507104 Chr.10: 74133973-74133973 on Build GRCh38
rs201763864 Chr.10: 74134202-74134202 on Build GRCh38
rs924000710 Chr.10: 74134045-74134045 on Build GRCh38
rs746656372 Chr.10: 74134184-74134184 on Build GRCh38
rs781090517 Chr.10: 74134160-74134160 on Build GRCh38
rs149906104 Chr.10: 74134414-74134414 on Build GRCh38
rs550880952 Chr.10: 74134299-74134299 on Build GRCh38
rs761055584 Chr.10: 74134201-74134201 on Build GRCh38
rs768675374 Chr.10: 74134155-74134155 on Build GRCh38
rs775713856 Chr.10: 74134226-74134226 on Build GRCh38
rs768322023 Chr.10: 74134075-74134075 on Build GRCh38
rs199587398 Chr.10: 74134231-74134232 on Build GRCh38
rs146857163 Chr.10: 74134067-74134067 on Build GRCh38
rs898778384 Chr.10: 74134139-74134139 on Build GRCh38
rs751729038 Chr.10: 74133984-74133984 on Build GRCh38
rs368987243 Chr.10: 74134203-74134203 on Build GRCh38
rs182488263 Chr.10: 74134401-74134401 on Build GRCh38
rs756956509 Chr.10: 74134019-74134019 on Build GRCh38
rs374316341 Chr.10: 74134105-74134105 on Build GRCh38
rs753141719 Chr.10: 74134104-74134104 on Build GRCh38
rs147357101 Chr.10: 74134046-74134046 on Build GRCh38
rs746045447 Chr.10: 74134207-74134207 on Build GRCh38
rs1047100515 Chr.10: 74134148-74134148 on Build GRCh38
rs562118078 Chr.10: 74134408-74134408 on Build GRCh38
rs967341632 Chr.10: 74134318-74134318 on Build GRCh38
rs763235391 Chr.10: 74134087-74134087 on Build GRCh38
rs776537184 Chr.10: 74134079-74134079 on Build GRCh38
rs774718328 Chr.10: 74134167-74134167 on Build GRCh38
rs766543590 Chr.10: 74133979-74133979 on Build GRCh38
rs946476951 Chr.10: 74134071-74134071 on Build GRCh38
rs757853058 Chr.10: 74134142-74134142 on Build GRCh38
rs778368356 Chr.10: 74134027-74134027 on Build GRCh38
rs770129831 Chr.10: 74134185-74134185 on Build GRCh38
rs1007900236 Chr.10: 74134188-74134188 on Build GRCh38
rs987733149 Chr.10: 74134333-74134333 on Build GRCh38
rs775305009 Chr.10: 74134050-74134050 on Build GRCh38
rs764345053 Chr.10: 74134112-74134112 on Build GRCh38
rs745414174 Chr.10: 74134035-74134035 on Build GRCh38
rs1011207719 Chr.10: 74134268-74134268 on Build GRCh38
rs964275217 Chr.10: 74134190-74134190 on Build GRCh38
rs767998735 Chr.10: 74134100-74134100 on Build GRCh38
rs34222970 Chr.10: 74134135-74134136 on Build GRCh38
rs759699471 Chr.10: 74134098-74134098 on Build GRCh38
rs756437347 Chr.10: 74134003-74134003 on Build GRCh38
rs905048758 Chr.10: 74134091-74134091 on Build GRCh38
rs775688981 Chr.10: 74134187-74134187 on Build GRCh38
rs200283960 Chr.10: 74134080-74134080 on Build GRCh38
rs202202576 Chr.10: 74134009-74134009 on Build GRCh38
rs896410387 Chr.10: 74134145-74134145 on Build GRCh38
rs1043532636 Chr.10: 74134077-74134077 on Build GRCh38
rs767927972 Chr.10: 74133992-74133992 on Build GRCh38
rs529931208 Chr.10: 74134413-74134413 on Build GRCh38
rs140657150 Chr.10: 74134097-74134097 on Build GRCh38
rs779670202 Chr.10: 74134147-74134147 on Build GRCh38
rs543981306 Chr.10: 74134277-74134277 on Build GRCh38
rs766739538 Chr.10: 74134094-74134094 on Build GRCh38
rs747979202 Chr.10: 74134161-74134161 on Build GRCh38
rs771219571 Chr.10: 74134163-74134163 on Build GRCh38
rs754157302 Chr.10: 74134204-74134204 on Build GRCh38
rs142865646 Chr.10: 74134002-74134002 on Build GRCh38
rs997438220 Chr.10: 74134218-74134218 on Build GRCh38
rs191165214 Chr.10: 74134120-74134120 on Build GRCh38
rs139413801 Chr.10: 74134133-74134133 on Build GRCh38

View Full Product Details