Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 74224460-74224976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADK
Entrez Gene ID
132
Gene Name
adenosine kinase
Gene Aliases
AK
UniGene
Hs.656586
Gene Chromosome Location
Chr.10: 74151185-74709303 on Build GRCh38
Transcripts
NM_001123.3 NM_001202449.1 NM_001202450.1 NM_006721.3 XM_017015698.1 XM_017015700.1 XM_017015701.1 XM_017015702.1 XM_017015704.1 XM_017015705.1 XM_017015706.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5786376 c.141-8T>C p.? Chr.10: 74224530-74224530 on Build GRCh38
5115760 c.141-8T>A p.? Chr.10: 74224530-74224530 on Build GRCh38
5014929 c.188A>T p.K63M Chr.10: 74224585-74224585 on Build GRCh38
5661396 c.168C>G p.I56M Chr.10: 74224565-74224565 on Build GRCh38
236958 c.171G>A p.L57L Chr.10: 74224568-74224568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747709429 Chr.10: 74224743-74224743 on Build GRCh38
rs561437086 Chr.10: 74224654-74224654 on Build GRCh38
rs372338209 Chr.10: 74224533-74224533 on Build GRCh38
rs745774033 Chr.10: 74224554-74224554 on Build GRCh38
rs780717078 Chr.10: 74224487-74224488 on Build GRCh38
rs376900767 Chr.10: 74224487-74224487 on Build GRCh38
rs925132470 Chr.10: 74224749-74224749 on Build GRCh38
rs543257384 Chr.10: 74224653-74224653 on Build GRCh38
rs950059997 Chr.10: 74224821-74224821 on Build GRCh38
rs988869601 Chr.10: 74224797-74224797 on Build GRCh38
rs755713661 Chr.10: 74224523-74224523 on Build GRCh38
rs1011774455 Chr.10: 74224768-74224768 on Build GRCh38
rs1045677156 Chr.10: 74224827-74224827 on Build GRCh38
rs185643742 Chr.10: 74224938-74224938 on Build GRCh38
rs116477529 Chr.10: 74224689-74224689 on Build GRCh38
rs1049304110 Chr.10: 74224877-74224877 on Build GRCh38
rs181738631 Chr.10: 74224788-74224788 on Build GRCh38
rs772091738 Chr.10: 74224563-74224563 on Build GRCh38
rs901801653 Chr.10: 74224665-74224665 on Build GRCh38
rs1040783088 Chr.10: 74224640-74224640 on Build GRCh38
rs937630856 Chr.10: 74224854-74224854 on Build GRCh38
rs761937020 Chr.10: 74224862-74224862 on Build GRCh38
rs376565039 Chr.10: 74224635-74224635 on Build GRCh38
rs770072715 Chr.10: 74224602-74224602 on Build GRCh38
rs371415595 Chr.10: 74224494-74224494 on Build GRCh38
rs760137160 Chr.10: 74224505-74224505 on Build GRCh38
rs368122533 Chr.10: 74224529-74224529 on Build GRCh38
rs1005026659 Chr.10: 74224895-74224895 on Build GRCh38
rs935167001 Chr.10: 74224794-74224794 on Build GRCh38
rs375259414 Chr.10: 74224503-74224503 on Build GRCh38
rs778545091 Chr.10: 74224543-74224543 on Build GRCh38
rs953452051 Chr.10: 74224697-74224697 on Build GRCh38
rs12773903 Chr.10: 74224891-74224891 on Build GRCh38
rs913295827 Chr.10: 74224804-74224804 on Build GRCh38
rs771541515 Chr.10: 74224764-74224764 on Build GRCh38
rs1023111390 Chr.10: 74224796-74224796 on Build GRCh38
rs11592294 Chr.10: 74224816-74224816 on Build GRCh38
rs369965648 Chr.10: 74224583-74224583 on Build GRCh38
rs767741490 Chr.10: 74224508-74224508 on Build GRCh38
rs1014684102 Chr.10: 74224903-74224903 on Build GRCh38
rs911146193 Chr.10: 74224633-74224633 on Build GRCh38
rs148236347 Chr.10: 74224614-74224614 on Build GRCh38
rs779676595 Chr.10: 74224589-74224589 on Build GRCh38
rs753135671 Chr.10: 74224522-74224522 on Build GRCh38
rs546706589 Chr.10: 74224724-74224724 on Build GRCh38
rs771109216 Chr.10: 74224617-74224617 on Build GRCh38
rs763127729 Chr.10: 74224492-74224492 on Build GRCh38
rs773670440 Chr.10: 74224538-74224538 on Build GRCh38
rs112701292 Chr.10: 74224843-74224843 on Build GRCh38
rs372962992 Chr.10: 74224547-74224547 on Build GRCh38
rs1003093152 Chr.10: 74224860-74224860 on Build GRCh38
rs746826404 Chr.10: 74224590-74224590 on Build GRCh38
rs754194632 Chr.10: 74224496-74224497 on Build GRCh38
rs773561592 Chr.10: 74224609-74224609 on Build GRCh38
rs751992871 Chr.10: 74224501-74224501 on Build GRCh38
rs985249100 Chr.10: 74224626-74224626 on Build GRCh38

View Full Product Details