Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 77805585-77806090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLG5
Entrez Gene ID
9231
Gene Name
discs large MAGUK scaffold protein 5
Gene Aliases
LP-DLG, P-DLG5, PDLG
UniGene
Hs.652690
Gene Chromosome Location
Chr.10: 77790787-77926590 on Build GRCh38
Transcripts
NM_004747.3 XM_005270276.4 XM_006718056.3 XM_011540341.2 XM_011540342.1 XM_011540344.2 XM_011540345.1 XM_011540346.2 XM_011540347.2 XM_017016913.1 XM_017016914.1 XM_017016915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5837351 c.5069_5070GG>AA p.R1690Q Chr.10: 77805759-77805760 on Build GRCh38
1297457 c.5053C>T p.R1685C Chr.10: 77805776-77805776 on Build GRCh38
3440869 c.4995C>T p.L1665L Chr.10: 77805834-77805834 on Build GRCh38
3440868 c.5099C>T p.S1700F Chr.10: 77805730-77805730 on Build GRCh38
3770695 c.5029A>G p.T1677A Chr.10: 77805800-77805800 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761585268 Chr.10: 77805889-77805889 on Build GRCh38
rs768409732 Chr.10: 77805896-77805896 on Build GRCh38
rs932723417 Chr.10: 77805986-77805986 on Build GRCh38
rs755224782 Chr.10: 77805745-77805745 on Build GRCh38
rs142696061 Chr.10: 77805739-77805739 on Build GRCh38
rs372879805 Chr.10: 77805681-77805681 on Build GRCh38
rs374472842 Chr.10: 77805622-77805622 on Build GRCh38
rs774554661 Chr.10: 77805775-77805775 on Build GRCh38
rs376031429 Chr.10: 77806027-77806027 on Build GRCh38
rs1014341481 Chr.10: 77806039-77806039 on Build GRCh38
rs748281696 Chr.10: 77805680-77805680 on Build GRCh38
rs753147607 Chr.10: 77806026-77806026 on Build GRCh38
rs1024886850 Chr.10: 77806049-77806049 on Build GRCh38
rs779352223 Chr.10: 77805830-77805830 on Build GRCh38
rs773545465 Chr.10: 77805702-77805702 on Build GRCh38
rs996999063 Chr.10: 77805963-77805963 on Build GRCh38
rs375737773 Chr.10: 77805801-77805801 on Build GRCh38
rs373426065 Chr.10: 77805707-77805707 on Build GRCh38
rs1014347485 Chr.10: 77806028-77806028 on Build GRCh38
rs137872892 Chr.10: 77805726-77805726 on Build GRCh38
rs1049859026 Chr.10: 77806010-77806010 on Build GRCh38
rs796704852 Chr.10: 77805925-77805925 on Build GRCh38
rs565102177 Chr.10: 77805933-77805933 on Build GRCh38
rs112842364 Chr.10: 77805723-77805723 on Build GRCh38
rs16935381 Chr.10: 77805771-77805771 on Build GRCh38
rs748404592 Chr.10: 77805761-77805761 on Build GRCh38
rs904750200 Chr.10: 77805999-77805999 on Build GRCh38
rs181618464 Chr.10: 77805971-77805971 on Build GRCh38
rs1023081298 Chr.10: 77806038-77806038 on Build GRCh38
rs1041124203 Chr.10: 77805727-77805727 on Build GRCh38
rs530075256 Chr.10: 77805998-77805998 on Build GRCh38
rs935705165 Chr.10: 77805882-77805882 on Build GRCh38
rs572783968 Chr.10: 77805732-77805732 on Build GRCh38
rs142141850 Chr.10: 77805792-77805792 on Build GRCh38
rs968735442 Chr.10: 77805677-77805677 on Build GRCh38
rs769972568 Chr.10: 77805825-77805825 on Build GRCh38
rs763678413 Chr.10: 77805734-77805734 on Build GRCh38
rs759173936 Chr.10: 77805714-77805714 on Build GRCh38
rs761637735 Chr.10: 77805793-77805793 on Build GRCh38
rs770028831 Chr.10: 77805682-77805682 on Build GRCh38
rs772792300 Chr.10: 77805910-77805918 on Build GRCh38
rs375715904 Chr.10: 77805740-77805740 on Build GRCh38
rs746635695 Chr.10: 77805888-77805888 on Build GRCh38
rs372312813 Chr.10: 77805783-77805783 on Build GRCh38
rs368743919 Chr.10: 77805764-77805764 on Build GRCh38
rs146019787 Chr.10: 77805760-77805760 on Build GRCh38
rs575052345 Chr.10: 77805960-77805960 on Build GRCh38
rs1044456530 Chr.10: 77805642-77805642 on Build GRCh38
rs149797176 Chr.10: 77805844-77805844 on Build GRCh38
rs944444489 Chr.10: 77805891-77805891 on Build GRCh38
rs781403402 Chr.10: 77805676-77805676 on Build GRCh38
rs746864916 Chr.10: 77805735-77805735 on Build GRCh38
rs756951798 Chr.10: 77805629-77805629 on Build GRCh38
rs146366181 Chr.10: 77805808-77805808 on Build GRCh38
rs200067112 Chr.10: 77805803-77805803 on Build GRCh38
rs185398005 Chr.10: 77806011-77806011 on Build GRCh38
rs41274580 Chr.10: 77805813-77805813 on Build GRCh38
rs201688240 Chr.10: 77805710-77805710 on Build GRCh38
rs756426835 Chr.10: 77805809-77805809 on Build GRCh38
rs769060861 Chr.10: 77805675-77805675 on Build GRCh38
rs200998713 Chr.10: 77805773-77805773 on Build GRCh38
rs761351074 Chr.10: 77805901-77805901 on Build GRCh38
rs764014393 Chr.10: 77806011-77806011 on Build GRCh38
rs747130524 Chr.10: 77805674-77805674 on Build GRCh38
rs768618972 Chr.10: 77805615-77805615 on Build GRCh38
rs944501025 Chr.10: 77805955-77805955 on Build GRCh38
rs760136712 Chr.10: 77805733-77805733 on Build GRCh38
rs746396118 Chr.10: 77805839-77805839 on Build GRCh38
rs1015774698 Chr.10: 77806024-77806024 on Build GRCh38
rs760330551 Chr.10: 77805789-77805789 on Build GRCh38
rs757672815 Chr.10: 77805828-77805828 on Build GRCh38
rs771136671 Chr.10: 77805708-77805708 on Build GRCh38
rs563053873 Chr.10: 77805966-77805966 on Build GRCh38
rs538894922 Chr.10: 77805641-77805641 on Build GRCh38
rs758222263 Chr.10: 77805804-77805804 on Build GRCh38
rs2289311 Chr.10: 77805897-77805897 on Build GRCh38
rs911568082 Chr.10: 77805870-77805870 on Build GRCh38
rs370462253 Chr.10: 77806012-77806012 on Build GRCh38
rs970292229 Chr.10: 77806061-77806061 on Build GRCh38
rs775833263 Chr.10: 77805872-77805872 on Build GRCh38
rs772456306 Chr.10: 77805784-77805784 on Build GRCh38

View Full Product Details