Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 79516084-79516595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF5AL1
Entrez Gene ID
143244
Gene Name
eukaryotic translation initiation factor 5A-like 1
Gene Aliases
EIF5AP1, bA342M3.3
UniGene
Hs.104825
Gene Chromosome Location
Chr.10: 79512601-79516440 on Build GRCh38
Transcripts
NM_001099692.1

SNP Detail

SNP ID Chromosome Location
rs138631936 Chr.10: 79516275-79516275 on Build GRCh38
rs918607443 Chr.10: 79516383-79516383 on Build GRCh38
rs143230318 Chr.10: 79516112-79516112 on Build GRCh38
rs535255412 Chr.10: 79516512-79516512 on Build GRCh38
rs533139087 Chr.10: 79516355-79516355 on Build GRCh38
rs1048981669 Chr.10: 79516414-79516414 on Build GRCh38
rs577539589 Chr.10: 79516152-79516152 on Build GRCh38
rs762119446 Chr.10: 79516534-79516535 on Build GRCh38
rs2945443 Chr.10: 79516176-79516176 on Build GRCh38
rs573039314 Chr.10: 79516549-79516549 on Build GRCh38
rs2559785 Chr.10: 79516334-79516334 on Build GRCh38
rs1049346188 Chr.10: 79516113-79516113 on Build GRCh38
rs187847988 Chr.10: 79516354-79516354 on Build GRCh38
rs570005224 Chr.10: 79516430-79516430 on Build GRCh38
rs141772176 Chr.10: 79516220-79516220 on Build GRCh38
rs1007283375 Chr.10: 79516120-79516120 on Build GRCh38
rs997573912 Chr.10: 79516204-79516204 on Build GRCh38
rs1042424943 Chr.10: 79516517-79516517 on Build GRCh38
rs887613949 Chr.10: 79516119-79516119 on Build GRCh38
rs3827877 Chr.10: 79516420-79516420 on Build GRCh38
rs543423006 Chr.10: 79516158-79516158 on Build GRCh38
rs528302009 Chr.10: 79516277-79516277 on Build GRCh38
rs370066815 Chr.10: 79516159-79516159 on Build GRCh38
rs1001141439 Chr.10: 79516267-79516267 on Build GRCh38
rs200690937 Chr.10: 79516187-79516187 on Build GRCh38
rs551238109 Chr.10: 79516306-79516306 on Build GRCh38
rs1038909326 Chr.10: 79516440-79516440 on Build GRCh38
rs929176838 Chr.10: 79516396-79516396 on Build GRCh38
rs149464575 Chr.10: 79516421-79516421 on Build GRCh38
rs1001489190 Chr.10: 79516548-79516548 on Build GRCh38
rs2559786 Chr.10: 79516451-79516451 on Build GRCh38
rs71186300 Chr.10: 79516438-79516438 on Build GRCh38
rs566256317 Chr.10: 79516476-79516476 on Build GRCh38
rs1020647816 Chr.10: 79516209-79516209 on Build GRCh38
rs745443508 Chr.10: 79516436-79516436 on Build GRCh38
rs563198351 Chr.10: 79516172-79516172 on Build GRCh38
rs901580514 Chr.10: 79516502-79516502 on Build GRCh38
rs990982037 Chr.10: 79516325-79516325 on Build GRCh38
rs959539173 Chr.10: 79516279-79516279 on Build GRCh38
rs578204609 Chr.10: 79516540-79516540 on Build GRCh38
rs558457304 Chr.10: 79516533-79516533 on Build GRCh38

View Full Product Details