Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 80432334-80432881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM213A
Entrez Gene ID
84293
Gene Name
family with sequence similarity 213 member A
Gene Aliases
C10orf58, PAMM
UniGene
Hs.500333
Gene Chromosome Location
Chr.10: 80407829-80437115 on Build GRCh38
Transcripts
NM_001243778.1 NM_001243779.1 NM_001243780.1 NM_001243781.1 NM_001243782.1 NM_032333.4 XM_011540265.2 XM_011540266.2 XM_017016775.1

SNP Detail

SNP ID Chromosome Location
rs527573293 Chr.10: 80432637-80432638 on Build GRCh38
rs7087581 Chr.10: 80432431-80432431 on Build GRCh38
rs916444139 Chr.10: 80432528-80432528 on Build GRCh38
rs386371932 Chr.10: 80432655-80432656 on Build GRCh38
rs1014899133 Chr.10: 80432495-80432495 on Build GRCh38
rs1018469337 Chr.10: 80432453-80432453 on Build GRCh38
rs955307087 Chr.10: 80432435-80432435 on Build GRCh38
rs981692523 Chr.10: 80432759-80432759 on Build GRCh38
rs531148258 Chr.10: 80432821-80432821 on Build GRCh38
rs1056672252 Chr.10: 80432668-80432668 on Build GRCh38
rs1140130 Chr.10: 80432562-80432562 on Build GRCh38
rs776790908 Chr.10: 80432664-80432664 on Build GRCh38
rs1006619667 Chr.10: 80432438-80432438 on Build GRCh38
rs556265914 Chr.10: 80432492-80432492 on Build GRCh38
rs951395332 Chr.10: 80432470-80432470 on Build GRCh38
rs567736686 Chr.10: 80432765-80432765 on Build GRCh38
rs114611572 Chr.10: 80432430-80432430 on Build GRCh38
rs927848234 Chr.10: 80432533-80432533 on Build GRCh38
rs890770771 Chr.10: 80432581-80432581 on Build GRCh38
rs1029161381 Chr.10: 80432409-80432409 on Build GRCh38
rs1050470601 Chr.10: 80432854-80432854 on Build GRCh38
rs935259913 Chr.10: 80432843-80432843 on Build GRCh38
rs962911073 Chr.10: 80432655-80432655 on Build GRCh38
rs765303193 Chr.10: 80432497-80432497 on Build GRCh38
rs999414364 Chr.10: 80432548-80432548 on Build GRCh38
rs572027244 Chr.10: 80432833-80432833 on Build GRCh38
rs147576957 Chr.10: 80432630-80432631 on Build GRCh38
rs958720592 Chr.10: 80432768-80432768 on Build GRCh38
rs1043610995 Chr.10: 80432419-80432419 on Build GRCh38
rs980698754 Chr.10: 80432599-80432599 on Build GRCh38
rs541839432 Chr.10: 80432788-80432788 on Build GRCh38
rs1025784058 Chr.10: 80432727-80432727 on Build GRCh38
rs987225801 Chr.10: 80432415-80432415 on Build GRCh38
rs951721721 Chr.10: 80432749-80432749 on Build GRCh38
rs1029688395 Chr.10: 80432498-80432498 on Build GRCh38
rs181570136 Chr.10: 80432422-80432422 on Build GRCh38
rs979240973 Chr.10: 80432825-80432825 on Build GRCh38
rs962860834 Chr.10: 80432459-80432459 on Build GRCh38
rs939374004 Chr.10: 80432534-80432534 on Build GRCh38
rs866050586 Chr.10: 80432500-80432500 on Build GRCh38
rs762736694 Chr.10: 80432631-80432632 on Build GRCh38
rs902429330 Chr.10: 80432539-80432539 on Build GRCh38
rs575945658 Chr.10: 80432636-80432636 on Build GRCh38
rs10712446 Chr.10: 80432656-80432656 on Build GRCh38
rs1063161 Chr.10: 80432369-80432369 on Build GRCh38
rs914904629 Chr.10: 80432576-80432576 on Build GRCh38
rs370261965 Chr.10: 80432635-80432635 on Build GRCh38
rs780011642 Chr.10: 80432496-80432496 on Build GRCh38
rs1007083088 Chr.10: 80432591-80432591 on Build GRCh38
rs1055854720 Chr.10: 80432510-80432510 on Build GRCh38
rs1051221454 Chr.10: 80432559-80432559 on Build GRCh38
rs1004983140 Chr.10: 80432476-80432476 on Build GRCh38
rs58893330 Chr.10: 80432656-80432657 on Build GRCh38
rs926556638 Chr.10: 80432606-80432606 on Build GRCh38
rs772449573 Chr.10: 80432631-80432633 on Build GRCh38
rs766490910 Chr.10: 80432523-80432523 on Build GRCh38
rs991556489 Chr.10: 80432784-80432784 on Build GRCh38
rs773198563 Chr.10: 80432410-80432410 on Build GRCh38
rs895864502 Chr.10: 80432469-80432469 on Build GRCh38
rs923735543 Chr.10: 80432826-80432826 on Build GRCh38
rs995666829 Chr.10: 80432684-80432684 on Build GRCh38
rs551120123 Chr.10: 80432582-80432582 on Build GRCh38
rs974347767 Chr.10: 80432461-80432461 on Build GRCh38
rs555229113 Chr.10: 80432792-80432792 on Build GRCh38
rs568805119 Chr.10: 80432364-80432364 on Build GRCh38
rs757397677 Chr.10: 80432462-80432462 on Build GRCh38

View Full Product Details