Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 86968324-86968822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADIRF
Entrez Gene ID
10974
Gene Name
adipogenesis regulatory factor
Gene Aliases
AFRO, APM2, C10orf116, apM-2
UniGene
Hs.642660
Gene Chromosome Location
Chr.10: 86968431-86970909 on Build GRCh38
Transcripts
NM_006829.2 XM_006717970.3

Gene Symbol
MMRN2
Entrez Gene ID
79812
Gene Name
multimerin 2
Gene Aliases
EMILIN-3, EMILIN3, ENDOGLYX-1
UniGene
Hs.524479
Gene Chromosome Location
Chr.10: 86935540-86971311 on Build GRCh38
Transcripts
NM_006829.2 XM_006717970.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3807877 c.6A>T p.A2A Chr.10: 86968550-86968550 on Build GRCh38
4016420 c.41A>T p.E14V Chr.10: 86968585-86968585 on Build GRCh38
5961348 c.30G>A p.K10K Chr.10: 86968574-86968574 on Build GRCh38
4016421 c.49G>A p.A17T Chr.10: 86968593-86968593 on Build GRCh38
1742717 c.11A>G p.K4R Chr.10: 86968555-86968555 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949827004 Chr.10: 86968673-86968673 on Build GRCh38
rs748361520 Chr.10: 86968626-86968626 on Build GRCh38
rs748922002 Chr.10: 86968639-86968639 on Build GRCh38
rs117748202 Chr.10: 86968489-86968489 on Build GRCh38
rs558739966 Chr.10: 86968799-86968799 on Build GRCh38
rs1027194914 Chr.10: 86968374-86968374 on Build GRCh38
rs989967658 Chr.10: 86968460-86968460 on Build GRCh38
rs759443267 Chr.10: 86968621-86968621 on Build GRCh38
rs768932954 Chr.10: 86968516-86968516 on Build GRCh38
rs866670301 Chr.10: 86968598-86968598 on Build GRCh38
rs770060530 Chr.10: 86968628-86968628 on Build GRCh38
rs781609734 Chr.10: 86968625-86968625 on Build GRCh38
rs767229669 Chr.10: 86968593-86968593 on Build GRCh38
rs372651740 Chr.10: 86968390-86968390 on Build GRCh38
rs368735201 Chr.10: 86968542-86968542 on Build GRCh38
rs566651860 Chr.10: 86968421-86968421 on Build GRCh38
rs996086126 Chr.10: 86968358-86968358 on Build GRCh38
rs763542760 Chr.10: 86968555-86968555 on Build GRCh38
rs568187232 Chr.10: 86968459-86968459 on Build GRCh38
rs745579593 Chr.10: 86968507-86968507 on Build GRCh38
rs769965741 Chr.10: 86968541-86968541 on Build GRCh38
rs368836654 Chr.10: 86968696-86968696 on Build GRCh38
rs750656455 Chr.10: 86968599-86968599 on Build GRCh38
rs949788663 Chr.10: 86968383-86968383 on Build GRCh38
rs1033232430 Chr.10: 86968434-86968434 on Build GRCh38
rs763578902 Chr.10: 86968612-86968612 on Build GRCh38
rs572609779 Chr.10: 86968469-86968469 on Build GRCh38
rs911817095 Chr.10: 86968802-86968802 on Build GRCh38
rs533782792 Chr.10: 86968423-86968423 on Build GRCh38
rs750250871 Chr.10: 86968622-86968622 on Build GRCh38
rs757056024 Chr.10: 86968500-86968500 on Build GRCh38
rs774782856 Chr.10: 86968588-86968588 on Build GRCh38
rs553029011 Chr.10: 86968427-86968427 on Build GRCh38
rs970738022 Chr.10: 86968406-86968406 on Build GRCh38
rs757221181 Chr.10: 86968616-86968616 on Build GRCh38
rs753835670 Chr.10: 86968613-86968613 on Build GRCh38
rs958259273 Chr.10: 86968760-86968760 on Build GRCh38
rs373783757 Chr.10: 86968632-86968632 on Build GRCh38
rs1019517450 Chr.10: 86968792-86968792 on Build GRCh38
rs987226541 Chr.10: 86968801-86968801 on Build GRCh38
rs573367194 Chr.10: 86968656-86968656 on Build GRCh38
rs887287173 Chr.10: 86968701-86968701 on Build GRCh38
rs535571624 Chr.10: 86968470-86968470 on Build GRCh38
rs889368175 Chr.10: 86968758-86968758 on Build GRCh38
rs7922458 Chr.10: 86968410-86968410 on Build GRCh38
rs781226498 Chr.10: 86968521-86968521 on Build GRCh38
rs775891208 Chr.10: 86968775-86968775 on Build GRCh38
rs7476449 Chr.10: 86968545-86968545 on Build GRCh38
rs1041441701 Chr.10: 86968694-86968694 on Build GRCh38
rs1007690447 Chr.10: 86968481-86968481 on Build GRCh38
rs1033723060 Chr.10: 86968729-86968729 on Build GRCh38
rs540855066 Chr.10: 86968681-86968681 on Build GRCh38
rs970935942 Chr.10: 86968724-86968724 on Build GRCh38
rs889293301 Chr.10: 86968445-86968445 on Build GRCh38
rs370326633 Chr.10: 86968620-86968620 on Build GRCh38
rs199705865 Chr.10: 86968505-86968505 on Build GRCh38
rs759761706 Chr.10: 86968589-86968589 on Build GRCh38
rs758092077 Chr.10: 86968623-86968623 on Build GRCh38
rs1025236872 Chr.10: 86968723-86968723 on Build GRCh38
rs943040304 Chr.10: 86968504-86968504 on Build GRCh38
rs568885154 Chr.10: 86968353-86968353 on Build GRCh38
rs1004363263 Chr.10: 86968704-86968704 on Build GRCh38
rs528962300 Chr.10: 86968728-86968728 on Build GRCh38
rs1054712263 Chr.10: 86968725-86968725 on Build GRCh38
rs867762842 Chr.10: 86968677-86968677 on Build GRCh38
rs989606592 Chr.10: 86968772-86968772 on Build GRCh38
rs759286743 Chr.10: 86968548-86968551 on Build GRCh38
rs765192041 Chr.10: 86968702-86968702 on Build GRCh38
rs984016793 Chr.10: 86968596-86968596 on Build GRCh38
rs1051645219 Chr.10: 86968490-86968490 on Build GRCh38
rs376111042 Chr.10: 86968503-86968503 on Build GRCh38
rs776366012 Chr.10: 86968514-86968514 on Build GRCh38
rs748273782 Chr.10: 86968533-86968533 on Build GRCh38
rs895518726 Chr.10: 86968371-86968371 on Build GRCh38
rs1004497195 Chr.10: 86968385-86968385 on Build GRCh38
rs752314799 Chr.10: 86968597-86968597 on Build GRCh38
rs771267919 Chr.10: 86968559-86968559 on Build GRCh38

View Full Product Details