Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 88941145-88941643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTA2
Entrez Gene ID
59
Gene Name
actin, alpha 2, smooth muscle, aorta
Gene Aliases
AAT6, ACTSA, MYMY5
UniGene
Hs.500483
Gene Chromosome Location
Chr.10: 88935074-88991397 on Build GRCh38
Transcripts
NM_001141945.2 NM_001320855.1 NM_001613.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
268569 c.607G>A p.V203I Chr.10: 88941238-88941238 on Build GRCh38
428100 c.554G>A p.R185Q Chr.10: 88941291-88941291 on Build GRCh38
4016483 c.479G>C p.G160A Chr.10: 88941366-88941366 on Build GRCh38
5616483 c.564T>C p.T188T Chr.10: 88941281-88941281 on Build GRCh38
7287172 c.561C>T p.L187L Chr.10: 88941284-88941284 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024676050 Chr.10: 88941594-88941594 on Build GRCh38
rs886038487 Chr.10: 88941254-88941254 on Build GRCh38
rs772473154 Chr.10: 88941292-88941292 on Build GRCh38
rs1013667914 Chr.10: 88941558-88941558 on Build GRCh38
rs746931074 Chr.10: 88941215-88941215 on Build GRCh38
rs1039495261 Chr.10: 88941500-88941500 on Build GRCh38
rs768520604 Chr.10: 88941217-88941217 on Build GRCh38
rs779347351 Chr.10: 88941201-88941201 on Build GRCh38
rs761222556 Chr.10: 88941271-88941271 on Build GRCh38
rs988630652 Chr.10: 88941529-88941529 on Build GRCh38
rs35663318 Chr.10: 88941611-88941612 on Build GRCh38
rs913872906 Chr.10: 88941466-88941466 on Build GRCh38
rs764571918 Chr.10: 88941278-88941278 on Build GRCh38
rs751364401 Chr.10: 88941427-88941427 on Build GRCh38
rs886039303 Chr.10: 88941310-88941310 on Build GRCh38
rs932999387 Chr.10: 88941559-88941559 on Build GRCh38
rs143771352 Chr.10: 88941239-88941239 on Build GRCh38
rs772862676 Chr.10: 88941253-88941253 on Build GRCh38
rs754939086 Chr.10: 88941428-88941428 on Build GRCh38
rs761262130 Chr.10: 88941235-88941236 on Build GRCh38
rs970469775 Chr.10: 88941469-88941469 on Build GRCh38
rs745594878 Chr.10: 88941204-88941204 on Build GRCh38
rs765499730 Chr.10: 88941401-88941401 on Build GRCh38
rs34416895 Chr.10: 88941606-88941607 on Build GRCh38
rs757491338 Chr.10: 88941195-88941195 on Build GRCh38
rs981340302 Chr.10: 88941477-88941477 on Build GRCh38
rs914322656 Chr.10: 88941532-88941532 on Build GRCh38
rs886042811 Chr.10: 88941229-88941229 on Build GRCh38
rs776573747 Chr.10: 88941487-88941487 on Build GRCh38
rs759846557 Chr.10: 88941263-88941263 on Build GRCh38
rs139684702 Chr.10: 88941408-88941408 on Build GRCh38
rs777208799 Chr.10: 88941347-88941347 on Build GRCh38
rs375204799 Chr.10: 88941307-88941307 on Build GRCh38
rs77231346 Chr.10: 88941464-88941464 on Build GRCh38
rs772458406 Chr.10: 88941266-88941266 on Build GRCh38
rs746972765 Chr.10: 88941252-88941252 on Build GRCh38
rs1023004327 Chr.10: 88941465-88941465 on Build GRCh38
rs756108427 Chr.10: 88941184-88941184 on Build GRCh38
rs776191629 Chr.10: 88941222-88941222 on Build GRCh38
rs201681300 Chr.10: 88941337-88941337 on Build GRCh38
rs891270851 Chr.10: 88941335-88941335 on Build GRCh38
rs577706852 Chr.10: 88941447-88941447 on Build GRCh38
rs775874252 Chr.10: 88941270-88941270 on Build GRCh38
rs766926979 Chr.10: 88941336-88941336 on Build GRCh38
rs780995228 Chr.10: 88941430-88941430 on Build GRCh38
rs761805696 Chr.10: 88941368-88941368 on Build GRCh38
rs367895755 Chr.10: 88941557-88941557 on Build GRCh38
rs397516684 Chr.10: 88941238-88941238 on Build GRCh38
rs556350760 Chr.10: 88941437-88941437 on Build GRCh38
rs577958374 Chr.10: 88941568-88941568 on Build GRCh38
rs142963257 Chr.10: 88941544-88941544 on Build GRCh38
rs794728026 Chr.10: 88941367-88941367 on Build GRCh38
rs41284110 Chr.10: 88941551-88941551 on Build GRCh38
rs3816245 Chr.10: 88941407-88941407 on Build GRCh38
rs758287882 Chr.10: 88941208-88941208 on Build GRCh38
rs969667484 Chr.10: 88941394-88941394 on Build GRCh38
rs1038768671 Chr.10: 88941550-88941550 on Build GRCh38
rs201017430 Chr.10: 88941194-88941194 on Build GRCh38
rs891398947 Chr.10: 88941607-88941607 on Build GRCh38
rs933323197 Chr.10: 88941470-88941470 on Build GRCh38
rs41284108 Chr.10: 88941486-88941486 on Build GRCh38
rs759485973 Chr.10: 88941429-88941430 on Build GRCh38
rs753951062 Chr.10: 88941360-88941360 on Build GRCh38
rs936719391 Chr.10: 88941429-88941429 on Build GRCh38
rs112687898 Chr.10: 88941228-88941228 on Build GRCh38
rs886038868 Chr.10: 88941349-88941349 on Build GRCh38
rs779969849 Chr.10: 88941423-88941423 on Build GRCh38
rs1803028 Chr.10: 88941258-88941258 on Build GRCh38
rs387906592 Chr.10: 88941309-88941309 on Build GRCh38
rs1006792819 Chr.10: 88941493-88941493 on Build GRCh38

View Full Product Details