Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 88941648-88942146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTA2
Entrez Gene ID
59
Gene Name
actin, alpha 2, smooth muscle, aorta
Gene Aliases
AAT6, ACTSA, MYMY5
UniGene
Hs.500483
Gene Chromosome Location
Chr.10: 88935074-88991397 on Build GRCh38
Transcripts
NM_001141945.2 NM_001320855.1 NM_001613.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6316950 c.437C>A p.A146D Chr.10: 88941802-88941802 on Build GRCh38
1349684 c.386T>C p.F129S Chr.10: 88941853-88941853 on Build GRCh38
4016484 c.447C>T p.R149R Chr.10: 88941792-88941792 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748641367 Chr.10: 88941914-88941914 on Build GRCh38
rs776477704 Chr.10: 88941713-88941713 on Build GRCh38
rs560586453 Chr.10: 88942020-88942020 on Build GRCh38
rs181698127 Chr.10: 88941838-88941838 on Build GRCh38
rs778438517 Chr.10: 88941906-88941906 on Build GRCh38
rs987593678 Chr.10: 88942084-88942084 on Build GRCh38
rs1013365150 Chr.10: 88941970-88941970 on Build GRCh38
rs752839175 Chr.10: 88941768-88941768 on Build GRCh38
rs532242060 Chr.10: 88941758-88941758 on Build GRCh38
rs975010979 Chr.10: 88942055-88942055 on Build GRCh38
rs1003317801 Chr.10: 88941706-88941706 on Build GRCh38
rs1035596080 Chr.10: 88941708-88941708 on Build GRCh38
rs974440690 Chr.10: 88941755-88941755 on Build GRCh38
rs748855163 Chr.10: 88941880-88941880 on Build GRCh38
rs777243762 Chr.10: 88941873-88941873 on Build GRCh38
rs748785940 Chr.10: 88941711-88941711 on Build GRCh38
rs111234060 Chr.10: 88941870-88941870 on Build GRCh38
rs988206566 Chr.10: 88941702-88941702 on Build GRCh38
rs904877578 Chr.10: 88941985-88941985 on Build GRCh38
rs794728025 Chr.10: 88941793-88941793 on Build GRCh38
rs113752358 Chr.10: 88942095-88942095 on Build GRCh38
rs191977133 Chr.10: 88941828-88941828 on Build GRCh38
rs1002874590 Chr.10: 88941921-88941921 on Build GRCh38
rs553942058 Chr.10: 88942101-88942101 on Build GRCh38
rs141933412 Chr.10: 88941849-88941849 on Build GRCh38
rs761941154 Chr.10: 88941792-88941792 on Build GRCh38
rs777366519 Chr.10: 88941748-88941748 on Build GRCh38
rs762567614 Chr.10: 88941819-88941819 on Build GRCh38
rs368708487 Chr.10: 88941770-88941770 on Build GRCh38
rs1021154735 Chr.10: 88942061-88942061 on Build GRCh38
rs571537330 Chr.10: 88942024-88942024 on Build GRCh38
rs751022454 Chr.10: 88942018-88942018 on Build GRCh38
rs140147046 Chr.10: 88941701-88941701 on Build GRCh38
rs913975511 Chr.10: 88941712-88941712 on Build GRCh38
rs111265233 Chr.10: 88941822-88941822 on Build GRCh38
rs1024628014 Chr.10: 88941978-88941978 on Build GRCh38
rs267602614 Chr.10: 88941832-88941832 on Build GRCh38
rs770619416 Chr.10: 88941757-88941757 on Build GRCh38
rs763870834 Chr.10: 88941907-88941907 on Build GRCh38
rs955996837 Chr.10: 88941710-88941710 on Build GRCh38
rs761016406 Chr.10: 88941768-88941768 on Build GRCh38
rs747344511 Chr.10: 88941907-88941907 on Build GRCh38
rs714887 Chr.10: 88941888-88941888 on Build GRCh38
rs886850657 Chr.10: 88941942-88941942 on Build GRCh38
rs942599042 Chr.10: 88941721-88941721 on Build GRCh38
rs186832257 Chr.10: 88942021-88942021 on Build GRCh38
rs910525345 Chr.10: 88942105-88942105 on Build GRCh38
rs765374178 Chr.10: 88941804-88941804 on Build GRCh38
rs121434526 Chr.10: 88941794-88941794 on Build GRCh38
rs769314559 Chr.10: 88941735-88941735 on Build GRCh38
rs190186807 Chr.10: 88942040-88942040 on Build GRCh38
rs954654477 Chr.10: 88942119-88942119 on Build GRCh38
rs1002427058 Chr.10: 88942004-88942004 on Build GRCh38
rs777091909 Chr.10: 88941913-88941913 on Build GRCh38
rs1023470970 Chr.10: 88941690-88941690 on Build GRCh38
rs758676977 Chr.10: 88941896-88941897 on Build GRCh38
rs767954449 Chr.10: 88942080-88942081 on Build GRCh38
rs751300489 Chr.10: 88941836-88941836 on Build GRCh38
rs79127111 Chr.10: 88941720-88941720 on Build GRCh38
rs765398351 Chr.10: 88941747-88941748 on Build GRCh38
rs780576052 Chr.10: 88942042-88942042 on Build GRCh38
rs368515500 Chr.10: 88942016-88942016 on Build GRCh38
rs925417312 Chr.10: 88941693-88941693 on Build GRCh38
rs374641258 Chr.10: 88941752-88941752 on Build GRCh38
rs932988356 Chr.10: 88941764-88941764 on Build GRCh38
rs147463067 Chr.10: 88941677-88941677 on Build GRCh38
rs781132665 Chr.10: 88941840-88941840 on Build GRCh38
rs752609914 Chr.10: 88941846-88941846 on Build GRCh38
rs1021515294 Chr.10: 88942028-88942028 on Build GRCh38
rs898571837 Chr.10: 88941675-88941675 on Build GRCh38
rs1028722929 Chr.10: 88942073-88942073 on Build GRCh38
rs547203192 Chr.10: 88941771-88941771 on Build GRCh38
rs397516683 Chr.10: 88941820-88941820 on Build GRCh38

View Full Product Details