Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 90915929-90916425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD1
Entrez Gene ID
27063
Gene Name
ankyrin repeat domain 1
Gene Aliases
ALRP, C-193, CARP, CVARP, MCARP, bA320F15.2
UniGene
Hs.448589
Gene Chromosome Location
Chr.10: 90912100-90921275 on Build GRCh38
Transcripts
NM_014391.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4554597 c.625G>A p.G209R Chr.10: 90916197-90916197 on Build GRCh38
4818943 c.646G>A p.D216N Chr.10: 90916176-90916176 on Build GRCh38
5589655 c.590G>A p.G197E Chr.10: 90916232-90916232 on Build GRCh38
6537076 c.613T>C p.L205L Chr.10: 90916209-90916209 on Build GRCh38
4777282 c.626G>C p.G209A Chr.10: 90916196-90916196 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762041559 Chr.10: 90916165-90916165 on Build GRCh38
rs769305030 Chr.10: 90916239-90916239 on Build GRCh38
rs749531136 Chr.10: 90916274-90916275 on Build GRCh38
rs754771221 Chr.10: 90916197-90916197 on Build GRCh38
rs995402973 Chr.10: 90916086-90916086 on Build GRCh38
rs775996412 Chr.10: 90916288-90916288 on Build GRCh38
rs772870044 Chr.10: 90916132-90916132 on Build GRCh38
rs4933200 Chr.10: 90916370-90916370 on Build GRCh38
rs552123265 Chr.10: 90915950-90915950 on Build GRCh38
rs915573585 Chr.10: 90916364-90916364 on Build GRCh38
rs939065182 Chr.10: 90916065-90916065 on Build GRCh38
rs915185433 Chr.10: 90916396-90916396 on Build GRCh38
rs375020388 Chr.10: 90916296-90916296 on Build GRCh38
rs372707936 Chr.10: 90916138-90916138 on Build GRCh38
rs557044291 Chr.10: 90916356-90916356 on Build GRCh38
rs775666061 Chr.10: 90916269-90916270 on Build GRCh38
rs150275749 Chr.10: 90916100-90916100 on Build GRCh38
rs895289299 Chr.10: 90916080-90916080 on Build GRCh38
rs765256800 Chr.10: 90916167-90916167 on Build GRCh38
rs867949780 Chr.10: 90916233-90916233 on Build GRCh38
rs1018915821 Chr.10: 90916284-90916284 on Build GRCh38
rs869025363 Chr.10: 90916210-90916210 on Build GRCh38
rs756517879 Chr.10: 90916380-90916380 on Build GRCh38
rs761254276 Chr.10: 90916140-90916140 on Build GRCh38
rs1004893173 Chr.10: 90916128-90916128 on Build GRCh38
rs1007912054 Chr.10: 90915967-90915967 on Build GRCh38
rs944471086 Chr.10: 90916074-90916079 on Build GRCh38
rs534954006 Chr.10: 90916120-90916120 on Build GRCh38
rs941687322 Chr.10: 90916365-90916365 on Build GRCh38
rs770544020 Chr.10: 90916286-90916286 on Build GRCh38
rs890262786 Chr.10: 90916005-90916005 on Build GRCh38
rs868668600 Chr.10: 90916224-90916224 on Build GRCh38
rs769199158 Chr.10: 90916130-90916130 on Build GRCh38
rs747589860 Chr.10: 90916124-90916124 on Build GRCh38
rs1003651682 Chr.10: 90916290-90916290 on Build GRCh38
rs1037418556 Chr.10: 90916366-90916366 on Build GRCh38
rs797017029 Chr.10: 90916222-90916223 on Build GRCh38
rs992192584 Chr.10: 90916325-90916325 on Build GRCh38
rs964047285 Chr.10: 90916026-90916026 on Build GRCh38
rs994512244 Chr.10: 90916109-90916111 on Build GRCh38
rs781678671 Chr.10: 90916135-90916135 on Build GRCh38
rs745904223 Chr.10: 90916157-90916159 on Build GRCh38
rs750619783 Chr.10: 90916171-90916171 on Build GRCh38
rs779876669 Chr.10: 90916186-90916186 on Build GRCh38
rs538585155 Chr.10: 90916225-90916225 on Build GRCh38
rs397517251 Chr.10: 90916262-90916262 on Build GRCh38
rs769158987 Chr.10: 90916297-90916297 on Build GRCh38
rs751422509 Chr.10: 90916192-90916192 on Build GRCh38
rs112987534 Chr.10: 90916170-90916170 on Build GRCh38
rs1011581984 Chr.10: 90916040-90916040 on Build GRCh38
rs1018271703 Chr.10: 90916024-90916024 on Build GRCh38
rs768491548 Chr.10: 90916313-90916313 on Build GRCh38
rs769897907 Chr.10: 90916253-90916253 on Build GRCh38
rs762541365 Chr.10: 90916136-90916136 on Build GRCh38
rs1055240902 Chr.10: 90916077-90916077 on Build GRCh38
rs758588912 Chr.10: 90916182-90916182 on Build GRCh38
rs1004430284 Chr.10: 90916223-90916223 on Build GRCh38
rs781049118 Chr.10: 90916219-90916219 on Build GRCh38
rs777185169 Chr.10: 90916252-90916252 on Build GRCh38
rs794728974 Chr.10: 90916269-90916269 on Build GRCh38
rs548071940 Chr.10: 90916125-90916125 on Build GRCh38
rs1038770179 Chr.10: 90916078-90916078 on Build GRCh38
rs929073849 Chr.10: 90916376-90916376 on Build GRCh38
rs886680679 Chr.10: 90916106-90916106 on Build GRCh38
rs757357185 Chr.10: 90916166-90916166 on Build GRCh38
rs748104942 Chr.10: 90916238-90916238 on Build GRCh38

View Full Product Details