Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 93073338-93073844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP26A1
Entrez Gene ID
1592
Gene Name
cytochrome P450 family 26 subfamily A member 1
Gene Aliases
CP26, CYP26, P450RAI, P450RAI1
UniGene
Hs.150595
Gene Chromosome Location
Chr.10: 93073475-93077884 on Build GRCh38
Transcripts
NM_057157.2

SNP Detail

SNP ID Chromosome Location
rs530238000 Chr.10: 93073595-93073595 on Build GRCh38
rs111361951 Chr.10: 93073739-93073739 on Build GRCh38
rs932716123 Chr.10: 93073462-93073462 on Build GRCh38
rs977510377 Chr.10: 93073373-93073373 on Build GRCh38
rs111947416 Chr.10: 93073414-93073414 on Build GRCh38
rs763555752 Chr.10: 93073678-93073678 on Build GRCh38
rs886754131 Chr.10: 93073555-93073555 on Build GRCh38
rs1040714452 Chr.10: 93073776-93073776 on Build GRCh38
rs1008188640 Chr.10: 93073489-93073489 on Build GRCh38
rs929497794 Chr.10: 93073755-93073755 on Build GRCh38
rs529345178 Chr.10: 93073503-93073503 on Build GRCh38
rs373431017 Chr.10: 93073688-93073688 on Build GRCh38
rs973635849 Chr.10: 93073725-93073725 on Build GRCh38
rs897179422 Chr.10: 93073767-93073767 on Build GRCh38
rs998916304 Chr.10: 93073636-93073636 on Build GRCh38
rs977742814 Chr.10: 93073552-93073552 on Build GRCh38
rs1012418547 Chr.10: 93073770-93073770 on Build GRCh38
rs867352351 Chr.10: 93073740-93073740 on Build GRCh38
rs920726091 Chr.10: 93073729-93073729 on Build GRCh38
rs970389278 Chr.10: 93073367-93073367 on Build GRCh38
rs750912984 Chr.10: 93073536-93073536 on Build GRCh38
rs533111333 Chr.10: 93073460-93073460 on Build GRCh38
rs943810756 Chr.10: 93073771-93073771 on Build GRCh38
rs768058938 Chr.10: 93073374-93073374 on Build GRCh38
rs756236466 Chr.10: 93073709-93073709 on Build GRCh38
rs915934794 Chr.10: 93073697-93073697 on Build GRCh38
rs553961784 Chr.10: 93073768-93073768 on Build GRCh38
rs931031870 Chr.10: 93073514-93073514 on Build GRCh38
rs546600702 Chr.10: 93073486-93073486 on Build GRCh38
rs1037830579 Chr.10: 93073491-93073491 on Build GRCh38
rs371007952 Chr.10: 93073560-93073560 on Build GRCh38
rs72558182 Chr.10: 93073655-93073655 on Build GRCh38
rs993456733 Chr.10: 93073415-93073415 on Build GRCh38
rs1021432731 Chr.10: 93073622-93073622 on Build GRCh38
rs569396108 Chr.10: 93073494-93073494 on Build GRCh38
rs938714429 Chr.10: 93073381-93073381 on Build GRCh38
rs990585534 Chr.10: 93073360-93073360 on Build GRCh38
rs1024732347 Chr.10: 93073535-93073535 on Build GRCh38
rs549050198 Chr.10: 93073758-93073758 on Build GRCh38
rs959579052 Chr.10: 93073522-93073522 on Build GRCh38
rs147146881 Chr.10: 93073372-93073372 on Build GRCh38
rs968541938 Chr.10: 93073624-93073624 on Build GRCh38
rs1021493005 Chr.10: 93073370-93073370 on Build GRCh38
rs895265692 Chr.10: 93073769-93073769 on Build GRCh38
rs369720016 Chr.10: 93073562-93073562 on Build GRCh38
rs565057473 Chr.10: 93073451-93073451 on Build GRCh38
rs1031770920 Chr.10: 93073642-93073642 on Build GRCh38
rs571971356 Chr.10: 93073792-93073792 on Build GRCh38
rs899503815 Chr.10: 93073788-93073788 on Build GRCh38
rs992822289 Chr.10: 93073692-93073692 on Build GRCh38
rs560154775 Chr.10: 93073500-93073500 on Build GRCh38
rs1037033083 Chr.10: 93073490-93073490 on Build GRCh38
rs553773738 Chr.10: 93073765-93073766 on Build GRCh38
rs936632742 Chr.10: 93073442-93073442 on Build GRCh38
rs1027549742 Chr.10: 93073520-93073520 on Build GRCh38
rs1007907987 Chr.10: 93073558-93073558 on Build GRCh38
rs1049815826 Chr.10: 93073471-93073472 on Build GRCh38
rs970329853 Chr.10: 93073702-93073702 on Build GRCh38
rs1046770255 Chr.10: 93073760-93073760 on Build GRCh38
rs556227964 Chr.10: 93073563-93073563 on Build GRCh38
rs531826710 Chr.10: 93073763-93073763 on Build GRCh38
rs1031060253 Chr.10: 93073807-93073807 on Build GRCh38
rs996799012 Chr.10: 93073612-93073612 on Build GRCh38
rs533727979 Chr.10: 93073822-93073822 on Build GRCh38
rs916150178 Chr.10: 93073447-93073447 on Build GRCh38
rs754097096 Chr.10: 93073713-93073713 on Build GRCh38
rs1011969087 Chr.10: 93073526-93073526 on Build GRCh38

View Full Product Details