Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 93669571-93670105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRA10AC1
Entrez Gene ID
118924
Gene Name
fragile site, folic acid type, rare, fra(10)(q23.3) or fra(10)(q24.2) candidate 1
Gene Aliases
C10orf4, F26C11.1-like, FRA10A
UniGene
Hs.586650
Gene Chromosome Location
Chr.10: 93667883-93702572 on Build GRCh38
Transcripts
NM_145246.4 XM_005269515.4 XM_005269516.4 XM_011539259.2 XM_011539260.2 XM_017015656.1 XM_017015657.1 XM_017015658.1 XM_017015659.1 XM_017015660.1 XM_017015661.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3415367 c.943C>A p.L315I Chr.10: 93669831-93669831 on Build GRCh38
5999336 c.924G>A p.E308E Chr.10: 93669850-93669850 on Build GRCh38
5359760 c.906-7delT p.? Chr.10: 93669875-93669875 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs914573595 Chr.10: 93669977-93669977 on Build GRCh38
rs1012156533 Chr.10: 93669821-93669821 on Build GRCh38
rs1056733896 Chr.10: 93670054-93670054 on Build GRCh38
rs750107108 Chr.10: 93669901-93669901 on Build GRCh38
rs1054627925 Chr.10: 93669792-93669792 on Build GRCh38
rs573490458 Chr.10: 93669771-93669771 on Build GRCh38
rs1008331355 Chr.10: 93669844-93669844 on Build GRCh38
rs951502632 Chr.10: 93669648-93669648 on Build GRCh38
rs768436976 Chr.10: 93669874-93669875 on Build GRCh38
rs747437451 Chr.10: 93669807-93669807 on Build GRCh38
rs764783725 Chr.10: 93669895-93669895 on Build GRCh38
rs116917664 Chr.10: 93669670-93669670 on Build GRCh38
rs987066097 Chr.10: 93669682-93669682 on Build GRCh38
rs548722633 Chr.10: 93669989-93669989 on Build GRCh38
rs200909144 Chr.10: 93669918-93669920 on Build GRCh38
rs139454370 Chr.10: 93669890-93669890 on Build GRCh38
rs749402038 Chr.10: 93669921-93669922 on Build GRCh38
rs186174296 Chr.10: 93669685-93669685 on Build GRCh38
rs764061453 Chr.10: 93669892-93669892 on Build GRCh38
rs777168704 Chr.10: 93669795-93669795 on Build GRCh38
rs947359027 Chr.10: 93669987-93669987 on Build GRCh38
rs957803811 Chr.10: 93669959-93669959 on Build GRCh38
rs1044965992 Chr.10: 93670076-93670076 on Build GRCh38
rs186067421 Chr.10: 93670060-93670060 on Build GRCh38
rs771197579 Chr.10: 93669852-93669852 on Build GRCh38
rs368910560 Chr.10: 93669806-93669806 on Build GRCh38
rs767329113 Chr.10: 93669884-93669884 on Build GRCh38
rs374903280 Chr.10: 93669842-93669842 on Build GRCh38
rs757010573 Chr.10: 93669805-93669805 on Build GRCh38
rs995274223 Chr.10: 93669829-93669829 on Build GRCh38
rs775304754 Chr.10: 93669885-93669885 on Build GRCh38
rs947475165 Chr.10: 93669749-93669749 on Build GRCh38
rs781555095 Chr.10: 93669815-93669815 on Build GRCh38
rs372971841 Chr.10: 93669816-93669816 on Build GRCh38
rs368737198 Chr.10: 93669785-93669785 on Build GRCh38
rs758087279 Chr.10: 93669914-93669914 on Build GRCh38
rs924722347 Chr.10: 93669999-93669999 on Build GRCh38
rs375727424 Chr.10: 93669791-93669791 on Build GRCh38
rs1037024219 Chr.10: 93670058-93670061 on Build GRCh38
rs759778643 Chr.10: 93669873-93669873 on Build GRCh38
rs902365755 Chr.10: 93669763-93669763 on Build GRCh38
rs964681322 Chr.10: 93669961-93669961 on Build GRCh38
rs759287266 Chr.10: 93669927-93669927 on Build GRCh38
rs752960788 Chr.10: 93669696-93669696 on Build GRCh38
rs529066837 Chr.10: 93669968-93669968 on Build GRCh38
rs755408707 Chr.10: 93669813-93669813 on Build GRCh38
rs61866650 Chr.10: 93669872-93669872 on Build GRCh38
rs1023296435 Chr.10: 93669599-93669599 on Build GRCh38
rs114574048 Chr.10: 93669939-93669939 on Build GRCh38
rs181412848 Chr.10: 93669823-93669823 on Build GRCh38
rs773932723 Chr.10: 93669906-93669907 on Build GRCh38
rs1026803713 Chr.10: 93669896-93669896 on Build GRCh38
rs773302193 Chr.10: 93669616-93669616 on Build GRCh38
rs568774079 Chr.10: 93669657-93669657 on Build GRCh38
rs759686636 Chr.10: 93669862-93669862 on Build GRCh38
rs1041758886 Chr.10: 93669751-93669751 on Build GRCh38
rs891110649 Chr.10: 93669833-93669833 on Build GRCh38
rs1025369621 Chr.10: 93669832-93669832 on Build GRCh38
rs557077629 Chr.10: 93669681-93669681 on Build GRCh38
rs566198340 Chr.10: 93669949-93669949 on Build GRCh38
rs189385723 Chr.10: 93669780-93669780 on Build GRCh38
rs774567501 Chr.10: 93669858-93669858 on Build GRCh38
rs751418424 Chr.10: 93669784-93669784 on Build GRCh38
rs545957890 Chr.10: 93669724-93669724 on Build GRCh38
rs1012581406 Chr.10: 93670075-93670075 on Build GRCh38
rs779812005 Chr.10: 93669812-93669813 on Build GRCh38
rs553473100 Chr.10: 93669739-93669739 on Build GRCh38
rs975651355 Chr.10: 93669964-93669964 on Build GRCh38
rs753365003 Chr.10: 93669802-93669802 on Build GRCh38
rs918762049 Chr.10: 93669641-93669641 on Build GRCh38
rs867253750 Chr.10: 93669915-93669917 on Build GRCh38
rs962810460 Chr.10: 93669602-93669602 on Build GRCh38
rs555259865 Chr.10: 93669656-93669656 on Build GRCh38
rs550150744 Chr.10: 93669732-93669732 on Build GRCh38
rs557351709 Chr.10: 93670061-93670061 on Build GRCh38
rs923732383 Chr.10: 93669956-93669956 on Build GRCh38
rs79307310 Chr.10: 93669875-93669875 on Build GRCh38
rs912576445 Chr.10: 93670035-93670035 on Build GRCh38
rs533076242 Chr.10: 93669929-93669929 on Build GRCh38
rs756036020 Chr.10: 93669794-93669794 on Build GRCh38
rs371405598 Chr.10: 93669788-93669788 on Build GRCh38
rs143207481 Chr.10: 93669894-93669894 on Build GRCh38
rs1023731114 Chr.10: 93669822-93669822 on Build GRCh38
rs3765581 Chr.10: 93670000-93670000 on Build GRCh38

View Full Product Details