Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 94687686-94688188 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2C18
Entrez Gene ID
1562
Gene Name
cytochrome P450 family 2 subfamily C member 18
Gene Aliases
CPCI, CYP2C, CYP2C17, P450-6B/29C, P450IIC17
UniGene
Hs.511872
Gene Chromosome Location
Chr.10: 94683494-94736190 on Build GRCh38
Transcripts
NM_000772.2 NM_001128925.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3868109 c.211C>T p.L71L Chr.10: 94687812-94687812 on Build GRCh38
3868110 c.293G>A p.G98E Chr.10: 94687894-94687894 on Build GRCh38
4940687 c.360G>A p.W120* Chr.10: 94688153-94688153 on Build GRCh38
5837517 c.274G>A p.E92K Chr.10: 94687875-94687875 on Build GRCh38
1739671 c.394C>T p.R132W Chr.10: 94688187-94688187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141719908 Chr.10: 94687760-94687760 on Build GRCh38
rs753621976 Chr.10: 94687947-94687947 on Build GRCh38
rs199721510 Chr.10: 94687969-94687969 on Build GRCh38
rs542103551 Chr.10: 94688117-94688117 on Build GRCh38
rs779240819 Chr.10: 94688094-94688094 on Build GRCh38
rs375456478 Chr.10: 94687934-94687934 on Build GRCh38
rs1028427246 Chr.10: 94688061-94688061 on Build GRCh38
rs945957844 Chr.10: 94688044-94688044 on Build GRCh38
rs186174073 Chr.10: 94688005-94688005 on Build GRCh38
rs759747828 Chr.10: 94687825-94687825 on Build GRCh38
rs771430378 Chr.10: 94688083-94688083 on Build GRCh38
rs776558652 Chr.10: 94688131-94688131 on Build GRCh38
rs368264095 Chr.10: 94687840-94687840 on Build GRCh38
rs778402407 Chr.10: 94688078-94688078 on Build GRCh38
rs774569291 Chr.10: 94687821-94687821 on Build GRCh38
rs774677303 Chr.10: 94687949-94687949 on Build GRCh38
rs949351051 Chr.10: 94687850-94687850 on Build GRCh38
rs769142998 Chr.10: 94687789-94687789 on Build GRCh38
rs905420054 Chr.10: 94687912-94687912 on Build GRCh38
rs965130013 Chr.10: 94688051-94688051 on Build GRCh38
rs550117143 Chr.10: 94687869-94687869 on Build GRCh38
rs749917925 Chr.10: 94687848-94687848 on Build GRCh38
rs202113535 Chr.10: 94687795-94687795 on Build GRCh38
rs755878511 Chr.10: 94687738-94687738 on Build GRCh38
rs757917178 Chr.10: 94687865-94687865 on Build GRCh38
rs951644981 Chr.10: 94687937-94687937 on Build GRCh38
rs146239394 Chr.10: 94687794-94687794 on Build GRCh38
rs1002881931 Chr.10: 94687925-94687925 on Build GRCh38
rs576478468 Chr.10: 94687944-94687944 on Build GRCh38
rs371698646 Chr.10: 94687975-94687975 on Build GRCh38
rs763819511 Chr.10: 94687833-94687833 on Build GRCh38
rs770145786 Chr.10: 94687894-94687894 on Build GRCh38
rs566030952 Chr.10: 94687977-94687977 on Build GRCh38
rs553349325 Chr.10: 94688079-94688079 on Build GRCh38
rs371597682 Chr.10: 94688141-94688141 on Build GRCh38
rs748927230 Chr.10: 94687755-94687755 on Build GRCh38
rs117690007 Chr.10: 94688098-94688098 on Build GRCh38
rs775399047 Chr.10: 94688099-94688099 on Build GRCh38
rs757862150 Chr.10: 94687971-94687971 on Build GRCh38
rs556757426 Chr.10: 94688038-94688038 on Build GRCh38
rs754936490 Chr.10: 94687721-94687721 on Build GRCh38
rs767629969 Chr.10: 94687723-94687723 on Build GRCh38
rs773441991 Chr.10: 94687813-94687813 on Build GRCh38
rs775451730 Chr.10: 94687943-94687943 on Build GRCh38
rs201140921 Chr.10: 94687958-94687958 on Build GRCh38
rs200398730 Chr.10: 94687884-94687884 on Build GRCh38
rs528715501 Chr.10: 94687978-94687978 on Build GRCh38
rs748486797 Chr.10: 94687800-94687800 on Build GRCh38
rs201048148 Chr.10: 94688146-94688146 on Build GRCh38
rs868643042 Chr.10: 94687782-94687783 on Build GRCh38
rs372279673 Chr.10: 94687804-94687805 on Build GRCh38
rs750028305 Chr.10: 94688149-94688149 on Build GRCh38
rs751022156 Chr.10: 94688164-94688164 on Build GRCh38
rs767436073 Chr.10: 94687827-94687827 on Build GRCh38
rs764197481 Chr.10: 94687895-94687895 on Build GRCh38
rs367844838 Chr.10: 94687950-94687950 on Build GRCh38
rs778370337 Chr.10: 94687774-94687774 on Build GRCh38
rs979809079 Chr.10: 94687957-94687957 on Build GRCh38
rs143695270 Chr.10: 94687830-94687830 on Build GRCh38
rs568528448 Chr.10: 94687996-94687996 on Build GRCh38
rs778599617 Chr.10: 94687847-94687847 on Build GRCh38
rs763380144 Chr.10: 94687988-94687988 on Build GRCh38
rs761530319 Chr.10: 94688140-94688140 on Build GRCh38
rs182014387 Chr.10: 94687995-94687995 on Build GRCh38
rs750080010 Chr.10: 94687968-94687968 on Build GRCh38
rs190647817 Chr.10: 94688159-94688159 on Build GRCh38
rs539428834 Chr.10: 94688060-94688060 on Build GRCh38
rs762490017 Chr.10: 94688153-94688153 on Build GRCh38
rs768250057 Chr.10: 94687927-94687927 on Build GRCh38
rs755901390 Chr.10: 94687832-94687832 on Build GRCh38
rs770051570 Chr.10: 94687880-94687881 on Build GRCh38
rs766749841 Chr.10: 94687820-94687820 on Build GRCh38
rs1004377636 Chr.10: 94687986-94687986 on Build GRCh38
rs114439291 Chr.10: 94688042-94688042 on Build GRCh38
rs145731435 Chr.10: 94687926-94687926 on Build GRCh38
rs1048521036 Chr.10: 94687932-94687932 on Build GRCh38
rs749739080 Chr.10: 94687896-94687896 on Build GRCh38
rs143341184 Chr.10: 94687787-94687787 on Build GRCh38
rs117111102 Chr.10: 94688163-94688163 on Build GRCh38
rs763247577 Chr.10: 94687818-94687818 on Build GRCh38
rs756868739 Chr.10: 94687843-94687843 on Build GRCh38
rs1052516310 Chr.10: 94687822-94687822 on Build GRCh38
rs563730614 Chr.10: 94687928-94687928 on Build GRCh38
rs752524484 Chr.10: 94687731-94687731 on Build GRCh38
rs142985033 Chr.10: 94688167-94688167 on Build GRCh38
rs927024733 Chr.10: 94687987-94687987 on Build GRCh38
rs375742950 Chr.10: 94688082-94688082 on Build GRCh38
rs754531436 Chr.10: 94688166-94688166 on Build GRCh38
rs777421434 Chr.10: 94687744-94687744 on Build GRCh38
rs372444081 Chr.10: 94687867-94687867 on Build GRCh38
rs375505433 Chr.10: 94687823-94687823 on Build GRCh38
rs41291550 Chr.10: 94687805-94687805 on Build GRCh38
rs938461726 Chr.10: 94688012-94688012 on Build GRCh38
rs748632945 Chr.10: 94687878-94687878 on Build GRCh38

View Full Product Details