Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 94735390-94735893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2C18
Entrez Gene ID
1562
Gene Name
cytochrome P450 family 2 subfamily C member 18
Gene Aliases
CPCI, CYP2C, CYP2C17, P450-6B/29C, P450IIC17
UniGene
Hs.511872
Gene Chromosome Location
Chr.10: 94683494-94736190 on Build GRCh38
Transcripts
NM_000772.2 NM_001128925.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
921732 c.1452G>A p.Q484Q Chr.10: 94735423-94735423 on Build GRCh38
77549 c.1433G>T p.R478L Chr.10: 94735404-94735404 on Build GRCh38
3868112 c.1442C>T p.P481L Chr.10: 94735413-94735413 on Build GRCh38
1970778 c.1461C>T p.F487F Chr.10: 94735432-94735432 on Build GRCh38
4466993 c.1465C>T p.P489S Chr.10: 94735436-94735436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs955384499 Chr.10: 94735589-94735589 on Build GRCh38
rs937125236 Chr.10: 94735674-94735674 on Build GRCh38
rs537972253 Chr.10: 94735611-94735611 on Build GRCh38
rs1002251734 Chr.10: 94735835-94735835 on Build GRCh38
rs901970983 Chr.10: 94735643-94735643 on Build GRCh38
rs566420409 Chr.10: 94735534-94735534 on Build GRCh38
rs980469530 Chr.10: 94735547-94735547 on Build GRCh38
rs895468351 Chr.10: 94735743-94735743 on Build GRCh38
rs74696697 Chr.10: 94735554-94735554 on Build GRCh38
rs41291552 Chr.10: 94735653-94735653 on Build GRCh38
rs763628569 Chr.10: 94735638-94735638 on Build GRCh38
rs573172062 Chr.10: 94735429-94735429 on Build GRCh38
rs764785028 Chr.10: 94735439-94735439 on Build GRCh38
rs115308818 Chr.10: 94735411-94735411 on Build GRCh38
rs1029680666 Chr.10: 94735586-94735586 on Build GRCh38
rs908368775 Chr.10: 94735614-94735614 on Build GRCh38
rs184489530 Chr.10: 94735450-94735450 on Build GRCh38
rs757457974 Chr.10: 94735492-94735492 on Build GRCh38
rs543988958 Chr.10: 94735507-94735507 on Build GRCh38
rs2860840 Chr.10: 94735475-94735475 on Build GRCh38
rs549008165 Chr.10: 94735573-94735573 on Build GRCh38
rs753053283 Chr.10: 94735451-94735451 on Build GRCh38
rs571078555 Chr.10: 94735644-94735644 on Build GRCh38
rs760296935 Chr.10: 94735421-94735421 on Build GRCh38
rs1050452343 Chr.10: 94735647-94735647 on Build GRCh38
rs760471991 Chr.10: 94735417-94735417 on Build GRCh38
rs768235030 Chr.10: 94735425-94735425 on Build GRCh38
rs1042194 Chr.10: 94735727-94735727 on Build GRCh38
rs766476494 Chr.10: 94735441-94735441 on Build GRCh38
rs865843754 Chr.10: 94735469-94735469 on Build GRCh38
rs761182245 Chr.10: 94735436-94735436 on Build GRCh38
rs557566667 Chr.10: 94735616-94735616 on Build GRCh38
rs1040434018 Chr.10: 94735633-94735633 on Build GRCh38
rs1024755294 Chr.10: 94735512-94735512 on Build GRCh38
rs775248533 Chr.10: 94735414-94735414 on Build GRCh38
rs41291554 Chr.10: 94735870-94735870 on Build GRCh38
rs541850421 Chr.10: 94735800-94735800 on Build GRCh38
rs1000068655 Chr.10: 94735516-94735516 on Build GRCh38
rs568847355 Chr.10: 94735584-94735584 on Build GRCh38
rs76680226 Chr.10: 94735798-94735798 on Build GRCh38
rs778090259 Chr.10: 94735476-94735476 on Build GRCh38
rs903109824 Chr.10: 94735813-94735813 on Build GRCh38
rs1042192 Chr.10: 94735527-94735527 on Build GRCh38
rs754066112 Chr.10: 94735487-94735487 on Build GRCh38
rs982993116 Chr.10: 94735582-94735582 on Build GRCh38
rs546657533 Chr.10: 94735758-94735758 on Build GRCh38
rs575629338 Chr.10: 94735859-94735859 on Build GRCh38
rs531076517 Chr.10: 94735494-94735494 on Build GRCh38
rs181997244 Chr.10: 94735830-94735830 on Build GRCh38
rs1001139766 Chr.10: 94735702-94735702 on Build GRCh38
rs931262996 Chr.10: 94735569-94735569 on Build GRCh38
rs3740367 Chr.10: 94735509-94735509 on Build GRCh38
rs772062076 Chr.10: 94735413-94735413 on Build GRCh38
rs190761792 Chr.10: 94735724-94735724 on Build GRCh38
rs777208624 Chr.10: 94735467-94735467 on Build GRCh38
rs1026664380 Chr.10: 94735839-94735839 on Build GRCh38

View Full Product Details