Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 94989148-94989652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2C9
Entrez Gene ID
1559
Gene Name
cytochrome P450 family 2 subfamily C member 9
Gene Aliases
CPC9, CYP2C, CYP2C10, CYPIIC9, P450IIC9
UniGene
Hs.282624
Gene Chromosome Location
Chr.10: 94905867-94989395 on Build GRCh38
Transcripts
NM_000771.3 XM_017015758.1

SNP Detail

SNP ID Chromosome Location
rs981990092 Chr.10: 94989206-94989206 on Build GRCh38
rs952433675 Chr.10: 94989398-94989398 on Build GRCh38
rs544023907 Chr.10: 94989259-94989259 on Build GRCh38
rs59943433 Chr.10: 94989230-94989230 on Build GRCh38
rs9332244 Chr.10: 94989248-94989248 on Build GRCh38
rs935461448 Chr.10: 94989215-94989215 on Build GRCh38
rs148135940 Chr.10: 94989555-94989555 on Build GRCh38
rs183361728 Chr.10: 94989479-94989479 on Build GRCh38
rs985311781 Chr.10: 94989554-94989554 on Build GRCh38
rs756464465 Chr.10: 94989441-94989441 on Build GRCh38
rs548188162 Chr.10: 94989535-94989535 on Build GRCh38
rs897981482 Chr.10: 94989317-94989317 on Build GRCh38
rs989915403 Chr.10: 94989318-94989318 on Build GRCh38
rs74779754 Chr.10: 94989184-94989184 on Build GRCh38
rs1042622891 Chr.10: 94989407-94989407 on Build GRCh38
rs1029618823 Chr.10: 94989539-94989539 on Build GRCh38
rs898037866 Chr.10: 94989596-94989596 on Build GRCh38
rs149476715 Chr.10: 94989305-94989305 on Build GRCh38
rs1009546196 Chr.10: 94989284-94989284 on Build GRCh38
rs752098372 Chr.10: 94989520-94989534 on Build GRCh38
rs906762857 Chr.10: 94989411-94989411 on Build GRCh38
rs28969379 Chr.10: 94989340-94989340 on Build GRCh38
rs985282390 Chr.10: 94989270-94989270 on Build GRCh38
rs955272162 Chr.10: 94989551-94989551 on Build GRCh38
rs563900017 Chr.10: 94989271-94989271 on Build GRCh38
rs911868822 Chr.10: 94989560-94989560 on Build GRCh38
rs560032244 Chr.10: 94989294-94989295 on Build GRCh38
rs113539383 Chr.10: 94989428-94989428 on Build GRCh38
rs762967090 Chr.10: 94989316-94989316 on Build GRCh38
rs575278481 Chr.10: 94989258-94989258 on Build GRCh38
rs972663222 Chr.10: 94989233-94989233 on Build GRCh38
rs931977572 Chr.10: 94989243-94989243 on Build GRCh38
rs923754764 Chr.10: 94989211-94989211 on Build GRCh38
rs541385626 Chr.10: 94989221-94989221 on Build GRCh38
rs1037019490 Chr.10: 94989578-94989579 on Build GRCh38
rs750124064 Chr.10: 94989620-94989620 on Build GRCh38
rs757086207 Chr.10: 94989561-94989561 on Build GRCh38
rs915273830 Chr.10: 94989619-94989619 on Build GRCh38
rs1036562298 Chr.10: 94989310-94989310 on Build GRCh38
rs79463855 Chr.10: 94989492-94989492 on Build GRCh38
rs532714712 Chr.10: 94989276-94989276 on Build GRCh38
rs943446473 Chr.10: 94989304-94989304 on Build GRCh38
rs1017067333 Chr.10: 94989517-94989517 on Build GRCh38
rs945347678 Chr.10: 94989374-94989374 on Build GRCh38
rs531023354 Chr.10: 94989520-94989520 on Build GRCh38
rs956747035 Chr.10: 94989498-94989498 on Build GRCh38
rs1029545134 Chr.10: 94989239-94989239 on Build GRCh38
rs982427523 Chr.10: 94989476-94989476 on Build GRCh38
rs778975376 Chr.10: 94989292-94989292 on Build GRCh38
rs9332245 Chr.10: 94989424-94989424 on Build GRCh38
rs778111094 Chr.10: 94989467-94989467 on Build GRCh38
rs186217659 Chr.10: 94989570-94989570 on Build GRCh38
rs9332243 Chr.10: 94989183-94989183 on Build GRCh38
rs993917188 Chr.10: 94989326-94989326 on Build GRCh38
rs560062313 Chr.10: 94989334-94989334 on Build GRCh38

View Full Product Details